Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41522741-41523241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACO2
Entrez Gene ID
50
Gene Name
aconitase 2
Gene Aliases
ACONM, HEL-S-284, ICRD, OCA8, OPA9
UniGene
Hs.643610
Gene Chromosome Location
Chr.22: 41469125-41528989 on Build GRCh38
Transcripts
NM_001098.2 XM_017028812.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3713738 c.1180G>A p.G394R Chr.22: 41522871-41522871 on Build GRCh38
4164955 c.1153T>A p.C385S Chr.22: 41522844-41522844 on Build GRCh38
6495716 c.1181G>A p.G394E Chr.22: 41522872-41522872 on Build GRCh38
5393747 c.1286G>A p.R429Q Chr.22: 41522977-41522977 on Build GRCh38
727005 c.1290C>A p.D430E Chr.22: 41522981-41522981 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775964844 Chr.22: 41522870-41522870 on Build GRCh38
rs760522590 Chr.22: 41522984-41522984 on Build GRCh38
rs757438868 Chr.22: 41522976-41522976 on Build GRCh38
rs761368190 Chr.22: 41522872-41522872 on Build GRCh38
rs772651853 Chr.22: 41522875-41522875 on Build GRCh38
rs1033072823 Chr.22: 41523085-41523085 on Build GRCh38
rs945290880 Chr.22: 41523067-41523067 on Build GRCh38
rs200176862 Chr.22: 41523005-41523005 on Build GRCh38
rs536970136 Chr.22: 41523094-41523094 on Build GRCh38
rs777705978 Chr.22: 41522807-41522807 on Build GRCh38
rs775555834 Chr.22: 41522873-41522873 on Build GRCh38
rs769212302 Chr.22: 41523125-41523125 on Build GRCh38
rs773888238 Chr.22: 41523026-41523026 on Build GRCh38
rs569969104 Chr.22: 41523074-41523074 on Build GRCh38
rs748906180 Chr.22: 41523197-41523197 on Build GRCh38
rs767544666 Chr.22: 41523038-41523038 on Build GRCh38
rs769544199 Chr.22: 41522874-41522874 on Build GRCh38
rs901736928 Chr.22: 41523061-41523061 on Build GRCh38
rs373355832 Chr.22: 41522824-41522824 on Build GRCh38
rs771501852 Chr.22: 41522822-41522822 on Build GRCh38
rs752600434 Chr.22: 41522962-41522962 on Build GRCh38
rs751518465 Chr.22: 41522918-41522918 on Build GRCh38
rs765632202 Chr.22: 41523022-41523029 on Build GRCh38
rs567132709 Chr.22: 41523195-41523195 on Build GRCh38
rs746348188 Chr.22: 41522981-41522981 on Build GRCh38
rs779185723 Chr.22: 41522977-41522977 on Build GRCh38
rs1033538492 Chr.22: 41522897-41522897 on Build GRCh38
rs747295924 Chr.22: 41523194-41523194 on Build GRCh38
rs753933056 Chr.22: 41522970-41522970 on Build GRCh38
rs1008732741 Chr.22: 41523128-41523128 on Build GRCh38
rs770633599 Chr.22: 41523209-41523209 on Build GRCh38
rs989001844 Chr.22: 41522843-41522843 on Build GRCh38
rs760553974 Chr.22: 41523167-41523167 on Build GRCh38
rs759565200 Chr.22: 41522951-41522951 on Build GRCh38
rs772704469 Chr.22: 41522982-41522982 on Build GRCh38
rs767183173 Chr.22: 41522961-41522961 on Build GRCh38
rs745392648 Chr.22: 41523213-41523213 on Build GRCh38
rs1042307337 Chr.22: 41523068-41523068 on Build GRCh38
rs1001118979 Chr.22: 41523075-41523080 on Build GRCh38
rs752372964 Chr.22: 41523155-41523155 on Build GRCh38
rs1012884919 Chr.22: 41523103-41523103 on Build GRCh38
rs780269429 Chr.22: 41522983-41522983 on Build GRCh38
rs749794087 Chr.22: 41523111-41523111 on Build GRCh38
rs759383944 Chr.22: 41523037-41523037 on Build GRCh38
rs763292122 Chr.22: 41523181-41523182 on Build GRCh38
rs367600242 Chr.22: 41523015-41523015 on Build GRCh38
rs991916590 Chr.22: 41522922-41522922 on Build GRCh38
rs201999799 Chr.22: 41523025-41523025 on Build GRCh38
rs769264305 Chr.22: 41523002-41523002 on Build GRCh38
rs1017819477 Chr.22: 41523132-41523132 on Build GRCh38
rs370500848 Chr.22: 41523190-41523190 on Build GRCh38
rs547526146 Chr.22: 41522869-41522869 on Build GRCh38
rs368067033 Chr.22: 41522963-41522963 on Build GRCh38
rs1044916140 Chr.22: 41522909-41522909 on Build GRCh38
rs930822764 Chr.22: 41522971-41522971 on Build GRCh38
rs971427336 Chr.22: 41523165-41523165 on Build GRCh38
rs760137161 Chr.22: 41523001-41523001 on Build GRCh38
rs147361669 Chr.22: 41522964-41522964 on Build GRCh38
rs375838727 Chr.22: 41523168-41523168 on Build GRCh38
rs766263322 Chr.22: 41522906-41522906 on Build GRCh38
rs773834741 Chr.22: 41523211-41523211 on Build GRCh38
rs765404984 Chr.22: 41523186-41523186 on Build GRCh38
rs747414082 Chr.22: 41522997-41522997 on Build GRCh38
rs551069383 Chr.22: 41523031-41523031 on Build GRCh38
rs775989546 Chr.22: 41523170-41523171 on Build GRCh38
rs1040448716 Chr.22: 41523041-41523041 on Build GRCh38
rs373167398 Chr.22: 41523164-41523164 on Build GRCh38
rs762454725 Chr.22: 41522901-41522901 on Build GRCh38
rs1024521935 Chr.22: 41522933-41522933 on Build GRCh38
rs116964552 Chr.22: 41523178-41523178 on Build GRCh38
rs758774618 Chr.22: 41523189-41523189 on Build GRCh38
rs1024303988 Chr.22: 41523148-41523148 on Build GRCh38
rs754093555 Chr.22: 41523176-41523176 on Build GRCh38
rs772881111 Chr.22: 41523004-41523004 on Build GRCh38
rs369508892 Chr.22: 41522823-41522823 on Build GRCh38
rs756395995 Chr.22: 41522791-41522791 on Build GRCh38
rs1032662436 Chr.22: 41523220-41523220 on Build GRCh38

View Full Product Details