Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 32159278-32159784 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPD1L
Entrez Gene ID
23171
Gene Name
glycerol-3-phosphate dehydrogenase 1-like
Gene Aliases
GPD1-L
UniGene
Hs.82432
Gene Chromosome Location
Chr.3: 32106511-32168715 on Build GRCh38
Transcripts
NM_015141.3 XM_006713068.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4772973 c.855C>A p.T285T Chr.3: 32159570-32159570 on Build GRCh38
1753161 c.894G>T p.K298N Chr.3: 32159609-32159609 on Build GRCh38
4561862 c.901G>A p.G301R Chr.3: 32159616-32159616 on Build GRCh38
345498 c.859G>A p.E287K Chr.3: 32159574-32159574 on Build GRCh38
6097487 c.862G>C p.E288Q Chr.3: 32159577-32159577 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770341590 Chr.3: 32159466-32159466 on Build GRCh38
rs967324039 Chr.3: 32159490-32159490 on Build GRCh38
rs528251869 Chr.3: 32159476-32159476 on Build GRCh38
rs899133652 Chr.3: 32159717-32159717 on Build GRCh38
rs978683261 Chr.3: 32159511-32159511 on Build GRCh38
rs1005139319 Chr.3: 32159657-32159657 on Build GRCh38
rs7616454 Chr.3: 32159420-32159420 on Build GRCh38
rs546424875 Chr.3: 32159479-32159479 on Build GRCh38
rs777762215 Chr.3: 32159535-32159536 on Build GRCh38
rs146838953 Chr.3: 32159362-32159362 on Build GRCh38
rs57228367 Chr.3: 32159480-32159480 on Build GRCh38
rs763090435 Chr.3: 32159565-32159565 on Build GRCh38
rs534373652 Chr.3: 32159631-32159631 on Build GRCh38
rs770457115 Chr.3: 32159686-32159686 on Build GRCh38
rs1029175280 Chr.3: 32159474-32159474 on Build GRCh38
rs757034620 Chr.3: 32159715-32159715 on Build GRCh38
rs569115941 Chr.3: 32159542-32159542 on Build GRCh38
rs200045526 Chr.3: 32159621-32159621 on Build GRCh38
rs926477058 Chr.3: 32159633-32159633 on Build GRCh38
rs997875331 Chr.3: 32159356-32159356 on Build GRCh38
rs939290195 Chr.3: 32159371-32159371 on Build GRCh38
rs55689071 Chr.3: 32159421-32159421 on Build GRCh38
rs545138860 Chr.3: 32159401-32159401 on Build GRCh38
rs370985609 Chr.3: 32159685-32159685 on Build GRCh38
rs72546643 Chr.3: 32159690-32159690 on Build GRCh38
rs575130003 Chr.3: 32159746-32159746 on Build GRCh38
rs889250685 Chr.3: 32159478-32159481 on Build GRCh38
rs567472303 Chr.3: 32159724-32159724 on Build GRCh38
rs534441374 Chr.3: 32159344-32159344 on Build GRCh38
rs746611202 Chr.3: 32159666-32159666 on Build GRCh38
rs759520122 Chr.3: 32159594-32159594 on Build GRCh38
rs67330075 Chr.3: 32159481-32159481 on Build GRCh38
rs375589412 Chr.3: 32159710-32159710 on Build GRCh38
rs994918169 Chr.3: 32159469-32159481 on Build GRCh38
rs886715448 Chr.3: 32159360-32159360 on Build GRCh38
rs1034455416 Chr.3: 32159478-32159478 on Build GRCh38
rs66691298 Chr.3: 32159745-32159745 on Build GRCh38
rs1002066952 Chr.3: 32159402-32159402 on Build GRCh38
rs367951581 Chr.3: 32159699-32159699 on Build GRCh38
rs201275703 Chr.3: 32159640-32159640 on Build GRCh38
rs548662461 Chr.3: 32159537-32159537 on Build GRCh38
rs139760875 Chr.3: 32159689-32159689 on Build GRCh38
rs59329824 Chr.3: 32159481-32159481 on Build GRCh38
rs980454260 Chr.3: 32159632-32159632 on Build GRCh38
rs746059043 Chr.3: 32159653-32159653 on Build GRCh38
rs556282239 Chr.3: 32159352-32159352 on Build GRCh38
rs786204216 Chr.3: 32159662-32159662 on Build GRCh38
rs67330076 Chr.3: 32159481-32159481 on Build GRCh38
rs886767465 Chr.3: 32159616-32159616 on Build GRCh38
rs757152405 Chr.3: 32159617-32159617 on Build GRCh38
rs530148424 Chr.3: 32159546-32159546 on Build GRCh38
rs769123169 Chr.3: 32159674-32159674 on Build GRCh38
rs71622587 Chr.3: 32159480-32159481 on Build GRCh38
rs765782848 Chr.3: 32159548-32159548 on Build GRCh38
rs142955213 Chr.3: 32159712-32159712 on Build GRCh38
rs761124110 Chr.3: 32159534-32159534 on Build GRCh38
rs776874178 Chr.3: 32159675-32159675 on Build GRCh38
rs558410316 Chr.3: 32159429-32159429 on Build GRCh38
rs767716460 Chr.3: 32159703-32159703 on Build GRCh38
rs750536517 Chr.3: 32159549-32159549 on Build GRCh38
rs766733448 Chr.3: 32159566-32159566 on Build GRCh38
rs72546642 Chr.3: 32159591-32159591 on Build GRCh38
rs188138719 Chr.3: 32159593-32159593 on Build GRCh38
rs370423130 Chr.3: 32159481-32159482 on Build GRCh38
rs913235752 Chr.3: 32159485-32159485 on Build GRCh38
rs192905901 Chr.3: 32159612-32159612 on Build GRCh38
rs751812984 Chr.3: 32159585-32159585 on Build GRCh38
rs561286211 Chr.3: 32159447-32159447 on Build GRCh38
rs745614320 Chr.3: 32159584-32159584 on Build GRCh38
rs942568441 Chr.3: 32159532-32159532 on Build GRCh38
rs1009168479 Chr.3: 32159483-32159483 on Build GRCh38
rs963585481 Chr.3: 32159378-32159378 on Build GRCh38
rs112026900 Chr.3: 32159479-32159480 on Build GRCh38
rs546365846 Chr.3: 32159308-32159308 on Build GRCh38
rs925803064 Chr.3: 32159311-32159311 on Build GRCh38
rs942494824 Chr.3: 32159317-32159317 on Build GRCh38
rs143815080 Chr.3: 32159357-32159357 on Build GRCh38
rs372102105 Chr.3: 32159428-32159428 on Build GRCh38
rs1016420337 Chr.3: 32159366-32159366 on Build GRCh38
rs1039460254 Chr.3: 32159321-32159321 on Build GRCh38
rs879071128 Chr.3: 32159489-32159490 on Build GRCh38
rs766539575 Chr.3: 32159693-32159693 on Build GRCh38
rs760432302 Chr.3: 32159526-32159526 on Build GRCh38
rs35234737 Chr.3: 32159480-32159480 on Build GRCh38
rs757663381 Chr.3: 32159548-32159548 on Build GRCh38
rs771520513 Chr.3: 32159644-32159644 on Build GRCh38
rs751879292 Chr.3: 32159694-32159694 on Build GRCh38
rs72546641 Chr.3: 32159527-32159527 on Build GRCh38

View Full Product Details