Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 38136487-38136991 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAA1
Entrez Gene ID
30
Gene Name
acetyl-CoA acyltransferase 1
Gene Aliases
ACAA, PTHIO, THIO
UniGene
Hs.643487
Gene Chromosome Location
Chr.3: 38122710-38137242 on Build GRCh38
Transcripts
NM_001130410.1 NM_001607.3 NR_024024.1 XM_006713122.1 XM_006713123.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6591439 c.240G>A p.P80P Chr.3: 38136617-38136617 on Build GRCh38
6577231 c.81G>C p.L27L Chr.3: 38136955-38136955 on Build GRCh38
7284947 c.172-1G>A p.? Chr.3: 38136686-38136686 on Build GRCh38
6591440 c.260G>A p.C87Y Chr.3: 38136597-38136597 on Build GRCh38
4625082 c.63G>A p.P21P Chr.3: 38136973-38136973 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs983176008 Chr.3: 38136746-38136746 on Build GRCh38
rs548977459 Chr.3: 38136510-38136510 on Build GRCh38
rs374322144 Chr.3: 38136694-38136694 on Build GRCh38
rs192633018 Chr.3: 38136800-38136800 on Build GRCh38
rs761491032 Chr.3: 38136582-38136582 on Build GRCh38
rs749451057 Chr.3: 38136548-38136548 on Build GRCh38
rs906286688 Chr.3: 38136825-38136825 on Build GRCh38
rs757346816 Chr.3: 38136717-38136717 on Build GRCh38
rs775468325 Chr.3: 38136554-38136554 on Build GRCh38
rs776491983 Chr.3: 38136571-38136571 on Build GRCh38
rs915138955 Chr.3: 38136802-38136802 on Build GRCh38
rs1048707074 Chr.3: 38136887-38136887 on Build GRCh38
rs997342033 Chr.3: 38136515-38136515 on Build GRCh38
rs1027006003 Chr.3: 38136735-38136735 on Build GRCh38
rs199521345 Chr.3: 38136546-38136546 on Build GRCh38
rs756284410 Chr.3: 38136643-38136643 on Build GRCh38
rs1041260981 Chr.3: 38136760-38136760 on Build GRCh38
rs545918909 Chr.3: 38136919-38136919 on Build GRCh38
rs867629907 Chr.3: 38136670-38136670 on Build GRCh38
rs375658726 Chr.3: 38136586-38136586 on Build GRCh38
rs1004255722 Chr.3: 38136902-38136902 on Build GRCh38
rs370449813 Chr.3: 38136780-38136780 on Build GRCh38
rs952749430 Chr.3: 38136608-38136608 on Build GRCh38
rs372124585 Chr.3: 38136518-38136518 on Build GRCh38
rs777908503 Chr.3: 38136714-38136714 on Build GRCh38
rs760071041 Chr.3: 38136934-38136934 on Build GRCh38
rs934311705 Chr.3: 38136873-38136873 on Build GRCh38
rs961335542 Chr.3: 38136801-38136801 on Build GRCh38
rs764753369 Chr.3: 38136677-38136677 on Build GRCh38
rs760597468 Chr.3: 38136562-38136562 on Build GRCh38
rs7625290 Chr.3: 38136799-38136799 on Build GRCh38
rs945089040 Chr.3: 38136806-38136806 on Build GRCh38
rs761579214 Chr.3: 38136671-38136671 on Build GRCh38
rs771864589 Chr.3: 38136550-38136550 on Build GRCh38
rs769607248 Chr.3: 38136895-38136895 on Build GRCh38
rs773806251 Chr.3: 38136927-38136927 on Build GRCh38
rs11552536 Chr.3: 38136916-38136916 on Build GRCh38
rs893442512 Chr.3: 38136631-38136631 on Build GRCh38
rs538300595 Chr.3: 38136705-38136705 on Build GRCh38
rs901317535 Chr.3: 38136509-38136509 on Build GRCh38
rs971976307 Chr.3: 38136650-38136650 on Build GRCh38
rs922876523 Chr.3: 38136900-38136900 on Build GRCh38
rs779067061 Chr.3: 38136818-38136818 on Build GRCh38
rs773603850 Chr.3: 38136959-38136959 on Build GRCh38
rs779326089 Chr.3: 38136889-38136898 on Build GRCh38
rs768053072 Chr.3: 38136952-38136952 on Build GRCh38
rs768664247 Chr.3: 38136823-38136823 on Build GRCh38
rs184980517 Chr.3: 38136953-38136953 on Build GRCh38
rs767895580 Chr.3: 38136699-38136699 on Build GRCh38
rs772886344 Chr.3: 38136678-38136678 on Build GRCh38
rs941466063 Chr.3: 38136963-38136963 on Build GRCh38
rs1049869731 Chr.3: 38136938-38136938 on Build GRCh38
rs554314870 Chr.3: 38136770-38136770 on Build GRCh38
rs968470179 Chr.3: 38136861-38136861 on Build GRCh38
rs866279413 Chr.3: 38136972-38136972 on Build GRCh38
rs201015823 Chr.3: 38136761-38136761 on Build GRCh38
rs772795388 Chr.3: 38136899-38136899 on Build GRCh38
rs894624271 Chr.3: 38136850-38136850 on Build GRCh38
rs543905984 Chr.3: 38136763-38136763 on Build GRCh38
rs766926501 Chr.3: 38136712-38136713 on Build GRCh38
rs771023034 Chr.3: 38136655-38136655 on Build GRCh38
rs755405630 Chr.3: 38136604-38136604 on Build GRCh38
rs574307551 Chr.3: 38136781-38136781 on Build GRCh38
rs562996073 Chr.3: 38136753-38136753 on Build GRCh38
rs1042446328 Chr.3: 38136807-38136807 on Build GRCh38
rs775349894 Chr.3: 38136659-38136659 on Build GRCh38
rs371056247 Chr.3: 38136689-38136689 on Build GRCh38
rs757634562 Chr.3: 38136881-38136882 on Build GRCh38
rs939067186 Chr.3: 38136828-38136828 on Build GRCh38
rs746937159 Chr.3: 38136660-38136660 on Build GRCh38
rs749850931 Chr.3: 38136592-38136592 on Build GRCh38
rs568676843 Chr.3: 38136530-38136530 on Build GRCh38
rs756409737 Chr.3: 38136694-38136695 on Build GRCh38
rs753023953 Chr.3: 38136700-38136700 on Build GRCh38
rs997568340 Chr.3: 38136871-38136895 on Build GRCh38
rs566439081 Chr.3: 38136668-38136668 on Build GRCh38
rs767683203 Chr.3: 38136616-38136617 on Build GRCh38
rs888688864 Chr.3: 38136888-38136888 on Build GRCh38
rs778081842 Chr.3: 38136697-38136711 on Build GRCh38
rs915694656 Chr.3: 38136853-38136853 on Build GRCh38
rs758971878 Chr.3: 38136599-38136599 on Build GRCh38
rs151286491 Chr.3: 38136602-38136602 on Build GRCh38
rs924245143 Chr.3: 38136706-38136706 on Build GRCh38
rs1013072830 Chr.3: 38136877-38136877 on Build GRCh38
rs978483635 Chr.3: 38136866-38136866 on Build GRCh38
rs781161149 Chr.3: 38136841-38136841 on Build GRCh38
rs766833478 Chr.3: 38136601-38136601 on Build GRCh38
rs765897432 Chr.3: 38136692-38136692 on Build GRCh38
rs763966209 Chr.3: 38136567-38136567 on Build GRCh38
rs761001248 Chr.3: 38136965-38136965 on Build GRCh38
rs776765709 Chr.3: 38136682-38136682 on Build GRCh38
rs752783349 Chr.3: 38136660-38136661 on Build GRCh38
rs976982132 Chr.3: 38136520-38136520 on Build GRCh38
rs368322732 Chr.3: 38136614-38136614 on Build GRCh38
rs952767273 Chr.3: 38136745-38136745 on Build GRCh38
rs747991817 Chr.3: 38136855-38136855 on Build GRCh38
rs754078350 Chr.3: 38136715-38136715 on Build GRCh38
rs367873181 Chr.3: 38136698-38136698 on Build GRCh38
rs1042161119 Chr.3: 38136648-38136648 on Build GRCh38
rs147670996 Chr.3: 38136632-38136632 on Build GRCh38
rs372415749 Chr.3: 38136822-38136822 on Build GRCh38
rs932836230 Chr.3: 38136901-38136902 on Build GRCh38
rs749367861 Chr.3: 38136652-38136652 on Build GRCh38
rs371014950 Chr.3: 38136547-38136547 on Build GRCh38
rs767897561 Chr.3: 38136843-38136843 on Build GRCh38

View Full Product Details