Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49725305-49725810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IP6K1
Entrez Gene ID
9807
Gene Name
inositol hexakisphosphate kinase 1
Gene Aliases
IHPK1, PiUS
UniGene
Hs.386168
Gene Chromosome Location
Chr.3: 49724295-49786540 on Build GRCh38
Transcripts
NM_001006115.2 NM_001242829.1 NM_153273.3

SNP Detail

SNP ID Chromosome Location
rs1042809772 Chr.3: 49725642-49725642 on Build GRCh38
rs1017344443 Chr.3: 49725522-49725522 on Build GRCh38
rs948428295 Chr.3: 49725614-49725614 on Build GRCh38
rs908372752 Chr.3: 49725734-49725734 on Build GRCh38
rs1009114118 Chr.3: 49725676-49725676 on Build GRCh38
rs919744126 Chr.3: 49725510-49725510 on Build GRCh38
rs931118980 Chr.3: 49725513-49725513 on Build GRCh38
rs987432600 Chr.3: 49725373-49725373 on Build GRCh38
rs982665297 Chr.3: 49725534-49725534 on Build GRCh38
rs950863028 Chr.3: 49725706-49725706 on Build GRCh38
rs868730093 Chr.3: 49725434-49725434 on Build GRCh38
rs887313709 Chr.3: 49725426-49725426 on Build GRCh38
rs1002590491 Chr.3: 49725467-49725467 on Build GRCh38
rs976122982 Chr.3: 49725574-49725575 on Build GRCh38
rs896359532 Chr.3: 49725480-49725480 on Build GRCh38
rs11716895 Chr.3: 49725346-49725346 on Build GRCh38
rs983087112 Chr.3: 49725634-49725634 on Build GRCh38
rs138664819 Chr.3: 49725689-49725689 on Build GRCh38
rs904709152 Chr.3: 49725335-49725335 on Build GRCh38
rs752842006 Chr.3: 49725743-49725743 on Build GRCh38
rs1009886294 Chr.3: 49725761-49725761 on Build GRCh38
rs2131106 Chr.3: 49725695-49725695 on Build GRCh38
rs373392696 Chr.3: 49725587-49725587 on Build GRCh38
rs775559975 Chr.3: 49725622-49725622 on Build GRCh38
rs934232387 Chr.3: 49725596-49725596 on Build GRCh38
rs1021773421 Chr.3: 49725332-49725332 on Build GRCh38
rs774969001 Chr.3: 49725751-49725754 on Build GRCh38
rs538469063 Chr.3: 49725538-49725538 on Build GRCh38
rs543993079 Chr.3: 49725469-49725469 on Build GRCh38
rs1020792501 Chr.3: 49725503-49725503 on Build GRCh38
rs934270871 Chr.3: 49725377-49725377 on Build GRCh38
rs577851636 Chr.3: 49725633-49725633 on Build GRCh38
rs768273617 Chr.3: 49725669-49725669 on Build GRCh38
rs1047637569 Chr.3: 49725405-49725405 on Build GRCh38
rs150149795 Chr.3: 49725650-49725650 on Build GRCh38
rs922968160 Chr.3: 49725583-49725583 on Build GRCh38
rs938543991 Chr.3: 49725737-49725737 on Build GRCh38
rs758609705 Chr.3: 49725747-49725750 on Build GRCh38
rs556037932 Chr.3: 49725345-49725345 on Build GRCh38
rs973470770 Chr.3: 49725509-49725509 on Build GRCh38
rs894161086 Chr.3: 49725591-49725591 on Build GRCh38
rs753608226 Chr.3: 49725707-49725707 on Build GRCh38
rs943040067 Chr.3: 49725342-49725342 on Build GRCh38
rs938650945 Chr.3: 49725550-49725550 on Build GRCh38
rs1020908449 Chr.3: 49725765-49725765 on Build GRCh38
rs1017563189 Chr.3: 49725431-49725431 on Build GRCh38
rs905183197 Chr.3: 49725464-49725464 on Build GRCh38
rs144667181 Chr.3: 49725788-49725789 on Build GRCh38
rs955020718 Chr.3: 49725360-49725360 on Build GRCh38
rs36006366 Chr.3: 49725458-49725459 on Build GRCh38
rs948355338 Chr.3: 49725788-49725788 on Build GRCh38
rs973397032 Chr.3: 49725694-49725694 on Build GRCh38
rs376367401 Chr.3: 49725754-49725754 on Build GRCh38
rs760510267 Chr.3: 49725649-49725649 on Build GRCh38
rs193198247 Chr.3: 49725684-49725684 on Build GRCh38
rs1038445299 Chr.3: 49725450-49725450 on Build GRCh38
rs1009526881 Chr.3: 49725495-49725495 on Build GRCh38
rs559848548 Chr.3: 49725667-49725667 on Build GRCh38

View Full Product Details