Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 51896590-51896964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQCF1
Entrez Gene ID
132141
Gene Name
IQ motif containing F1
Gene Aliases
-
UniGene
Hs.671210
Gene Chromosome Location
Chr.3: 51894876-51903370 on Build GRCh38
Transcripts
NM_152397.2 XM_011533366.2 XM_017005729.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1180583 c.169A>G p.K57E Chr.3: 51896834-51896834 on Build GRCh38
3595827 c.109-1G>A p.? Chr.3: 51896895-51896895 on Build GRCh38
6565764 c.122T>C p.V41A Chr.3: 51896881-51896881 on Build GRCh38
1046760 c.113A>C p.K38T Chr.3: 51896890-51896890 on Build GRCh38
1566705 c.139A>G p.T47A Chr.3: 51896864-51896864 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748290427 Chr.3: 51896853-51896853 on Build GRCh38
rs754921110 Chr.3: 51896848-51896848 on Build GRCh38
rs773458106 Chr.3: 51896870-51896870 on Build GRCh38
rs758083999 Chr.3: 51896714-51896715 on Build GRCh38
rs747232922 Chr.3: 51896873-51896873 on Build GRCh38
rs770385200 Chr.3: 51896787-51896787 on Build GRCh38
rs74953880 Chr.3: 51896713-51896713 on Build GRCh38
rs180886755 Chr.3: 51896694-51896694 on Build GRCh38
rs888094846 Chr.3: 51896694-51896711 on Build GRCh38
rs868278278 Chr.3: 51896727-51896729 on Build GRCh38
rs59897184 Chr.3: 51896729-51896730 on Build GRCh38
rs199884981 Chr.3: 51896886-51896886 on Build GRCh38
rs149100108 Chr.3: 51896713-51896714 on Build GRCh38
rs770695868 Chr.3: 51896697-51896698 on Build GRCh38
rs150817790 Chr.3: 51896712-51896713 on Build GRCh38
rs895903703 Chr.3: 51896790-51896790 on Build GRCh38
rs548708647 Chr.3: 51896761-51896761 on Build GRCh38
rs1029433375 Chr.3: 51896704-51896713 on Build GRCh38
rs185472752 Chr.3: 51896822-51896822 on Build GRCh38
rs756796714 Chr.3: 51896811-51896811 on Build GRCh38
rs1015308821 Chr.3: 51896695-51896696 on Build GRCh38
rs1023234867 Chr.3: 51896690-51896690 on Build GRCh38
rs113385405 Chr.3: 51896697-51896697 on Build GRCh38
rs767231617 Chr.3: 51896815-51896815 on Build GRCh38
rs57859638 Chr.3: 51896834-51896834 on Build GRCh38
rs765585315 Chr.3: 51896804-51896805 on Build GRCh38
rs550823719 Chr.3: 51896695-51896695 on Build GRCh38
rs766985173 Chr.3: 51896925-51896925 on Build GRCh38
rs750061343 Chr.3: 51896931-51896931 on Build GRCh38
rs150592477 Chr.3: 51896878-51896878 on Build GRCh38
rs1032843367 Chr.3: 51896641-51896641 on Build GRCh38
rs1013159061 Chr.3: 51896687-51896688 on Build GRCh38
rs979181098 Chr.3: 51896813-51896813 on Build GRCh38
rs541052636 Chr.3: 51896867-51896868 on Build GRCh38
rs988543551 Chr.3: 51896687-51896687 on Build GRCh38
rs956102143 Chr.3: 51896658-51896658 on Build GRCh38
rs533630965 Chr.3: 51896715-51896715 on Build GRCh38
rs200764623 Chr.3: 51896804-51896804 on Build GRCh38
rs765848156 Chr.3: 51896887-51896887 on Build GRCh38
rs758683485 Chr.3: 51896711-51896712 on Build GRCh38
rs755820253 Chr.3: 51896932-51896932 on Build GRCh38
rs1048713815 Chr.3: 51896712-51896712 on Build GRCh38
rs777197675 Chr.3: 51896882-51896882 on Build GRCh38
rs1057315018 Chr.3: 51896642-51896642 on Build GRCh38
rs753497577 Chr.3: 51896691-51896691 on Build GRCh38
rs974418162 Chr.3: 51896699-51896699 on Build GRCh38
rs562105038 Chr.3: 51896693-51896693 on Build GRCh38
rs766593079 Chr.3: 51896696-51896697 on Build GRCh38
rs200040168 Chr.3: 51896709-51896709 on Build GRCh38
rs751097980 Chr.3: 51896810-51896810 on Build GRCh38
rs761871050 Chr.3: 51896805-51896810 on Build GRCh38
rs540677828 Chr.3: 51896624-51896624 on Build GRCh38
rs772132466 Chr.3: 51896854-51896854 on Build GRCh38
rs763685705 Chr.3: 51896808-51896808 on Build GRCh38
rs953909941 Chr.3: 51896756-51896756 on Build GRCh38
rs773698831 Chr.3: 51896893-51896893 on Build GRCh38
rs374427022 Chr.3: 51896920-51896920 on Build GRCh38
rs755883779 Chr.3: 51896827-51896827 on Build GRCh38
rs1057263564 Chr.3: 51896890-51896890 on Build GRCh38
rs200841297 Chr.3: 51896711-51896711 on Build GRCh38

View Full Product Details