Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 56624396-56624896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM208A
Entrez Gene ID
23272
Gene Name
family with sequence similarity 208 member A
Gene Aliases
C3orf63, RAP140, se89-1
UniGene
Hs.116877
Gene Chromosome Location
Chr.3: 56620132-56683242 on Build GRCh38
Transcripts
NM_001112736.1 NM_015224.3 XM_005264999.1 XM_006713077.2 XM_006713078.2 XM_011533552.2 XM_011533553.2 XM_017006030.1 XM_017006031.1 XM_017006032.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6659338 c.2988A>C p.R996R Chr.3: 56624847-56624847 on Build GRCh38
4002781 c.4120A>G p.I1374V Chr.3: 56624843-56624843 on Build GRCh38
3993251 c.4108C>G p.Q1370E Chr.3: 56624855-56624855 on Build GRCh38
1246832 c.2958C>G p.I986M Chr.3: 56624877-56624877 on Build GRCh38
6298565 c.4203C>A p.F1401L Chr.3: 56624576-56624576 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754049913 Chr.3: 56624627-56624627 on Build GRCh38
rs765959642 Chr.3: 56624434-56624435 on Build GRCh38
rs748184961 Chr.3: 56624658-56624658 on Build GRCh38
rs200302505 Chr.3: 56624647-56624647 on Build GRCh38
rs1013078076 Chr.3: 56624656-56624656 on Build GRCh38
rs769222976 Chr.3: 56624446-56624446 on Build GRCh38
rs945424563 Chr.3: 56624493-56624493 on Build GRCh38
rs780315978 Chr.3: 56624441-56624441 on Build GRCh38
rs1004328166 Chr.3: 56624666-56624666 on Build GRCh38
rs750686031 Chr.3: 56624693-56624693 on Build GRCh38
rs774146404 Chr.3: 56624439-56624440 on Build GRCh38
rs1043835546 Chr.3: 56624499-56624499 on Build GRCh38
rs921118145 Chr.3: 56624741-56624741 on Build GRCh38
rs146998440 Chr.3: 56624688-56624688 on Build GRCh38
rs149212000 Chr.3: 56624487-56624487 on Build GRCh38
rs760660772 Chr.3: 56624849-56624849 on Build GRCh38
rs768301507 Chr.3: 56624813-56624813 on Build GRCh38
rs773544532 Chr.3: 56624466-56624466 on Build GRCh38
rs549135591 Chr.3: 56624431-56624431 on Build GRCh38
rs915144999 Chr.3: 56624798-56624798 on Build GRCh38
rs772518948 Chr.3: 56624557-56624557 on Build GRCh38
rs752502719 Chr.3: 56624505-56624516 on Build GRCh38
rs766137176 Chr.3: 56624853-56624853 on Build GRCh38
rs570857085 Chr.3: 56624568-56624568 on Build GRCh38
rs750258668 Chr.3: 56624606-56624606 on Build GRCh38
rs1017197410 Chr.3: 56624621-56624621 on Build GRCh38
rs975265519 Chr.3: 56624433-56624433 on Build GRCh38
rs962518809 Chr.3: 56624425-56624425 on Build GRCh38
rs759208694 Chr.3: 56624469-56624469 on Build GRCh38
rs766193487 Chr.3: 56624623-56624623 on Build GRCh38
rs755205144 Chr.3: 56624645-56624645 on Build GRCh38
rs764829852 Chr.3: 56624483-56624483 on Build GRCh38
rs140615577 Chr.3: 56624541-56624541 on Build GRCh38
rs779019710 Chr.3: 56624545-56624545 on Build GRCh38
rs924698553 Chr.3: 56624705-56624705 on Build GRCh38
rs903984618 Chr.3: 56624515-56624515 on Build GRCh38
rs773741943 Chr.3: 56624832-56624832 on Build GRCh38
rs957390624 Chr.3: 56624773-56624773 on Build GRCh38
rs753621253 Chr.3: 56624855-56624855 on Build GRCh38
rs942643119 Chr.3: 56624808-56624808 on Build GRCh38
rs867705035 Chr.3: 56624442-56624442 on Build GRCh38
rs376621565 Chr.3: 56624685-56624685 on Build GRCh38
rs762660721 Chr.3: 56624570-56624570 on Build GRCh38
rs201657342 Chr.3: 56624489-56624489 on Build GRCh38
rs542859306 Chr.3: 56624521-56624521 on Build GRCh38
rs762517549 Chr.3: 56624433-56624434 on Build GRCh38
rs752786611 Chr.3: 56624865-56624865 on Build GRCh38
rs2291499 Chr.3: 56624453-56624453 on Build GRCh38
rs555904876 Chr.3: 56624696-56624696 on Build GRCh38
rs757226862 Chr.3: 56624518-56624518 on Build GRCh38
rs770097257 Chr.3: 56624440-56624440 on Build GRCh38
rs543892618 Chr.3: 56624759-56624759 on Build GRCh38
rs867566912 Chr.3: 56624611-56624611 on Build GRCh38
rs2291498 Chr.3: 56624843-56624843 on Build GRCh38
rs202213634 Chr.3: 56624472-56624472 on Build GRCh38
rs775059508 Chr.3: 56624569-56624569 on Build GRCh38
rs751515469 Chr.3: 56624505-56624505 on Build GRCh38
rs772466594 Chr.3: 56624457-56624457 on Build GRCh38
rs372124489 Chr.3: 56624848-56624848 on Build GRCh38
rs781127177 Chr.3: 56624428-56624428 on Build GRCh38
rs140848517 Chr.3: 56624618-56624618 on Build GRCh38
rs759707067 Chr.3: 56624858-56624858 on Build GRCh38
rs780084252 Chr.3: 56624538-56624538 on Build GRCh38
rs747305410 Chr.3: 56624565-56624565 on Build GRCh38
rs773351165 Chr.3: 56624844-56624844 on Build GRCh38
rs1016646021 Chr.3: 56624667-56624667 on Build GRCh38
rs771263695 Chr.3: 56624785-56624785 on Build GRCh38
rs187741245 Chr.3: 56624722-56624722 on Build GRCh38
rs759587986 Chr.3: 56624445-56624447 on Build GRCh38
rs758910183 Chr.3: 56624663-56624663 on Build GRCh38
rs781586004 Chr.3: 56624787-56624787 on Build GRCh38
rs767499232 Chr.3: 56624590-56624590 on Build GRCh38
rs757422290 Chr.3: 56624639-56624639 on Build GRCh38
rs375354208 Chr.3: 56624536-56624536 on Build GRCh38
rs368247313 Chr.3: 56624791-56624791 on Build GRCh38
rs35208241 Chr.3: 56624468-56624469 on Build GRCh38
rs750207156 Chr.3: 56624522-56624522 on Build GRCh38
rs145666615 Chr.3: 56624586-56624586 on Build GRCh38
rs137861082 Chr.3: 56624835-56624835 on Build GRCh38
rs763548119 Chr.3: 56624574-56624574 on Build GRCh38
rs748653314 Chr.3: 56624550-56624550 on Build GRCh38
rs765432735 Chr.3: 56624859-56624859 on Build GRCh38
rs773597905 Chr.3: 56624559-56624559 on Build GRCh38
rs150032629 Chr.3: 56624486-56624486 on Build GRCh38
rs749514396 Chr.3: 56624540-56624540 on Build GRCh38
rs765475646 Chr.3: 56624643-56624651 on Build GRCh38
rs774781087 Chr.3: 56624447-56624447 on Build GRCh38
rs773885716 Chr.3: 56624577-56624577 on Build GRCh38
rs767154706 Chr.3: 56624449-56624450 on Build GRCh38
rs958978776 Chr.3: 56624625-56624625 on Build GRCh38

View Full Product Details