Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 108160634-108161176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT57
Entrez Gene ID
55081
Gene Name
intraflagellar transport 57
Gene Aliases
ESRRBL1, HIPPI, MHS4R2
UniGene
Hs.412196
Gene Chromosome Location
Chr.3: 108160812-108222570 on Build GRCh38
Transcripts
NM_018010.3

SNP Detail

SNP ID Chromosome Location
rs1009241173 Chr.3: 108161044-108161044 on Build GRCh38
rs865825276 Chr.3: 108160727-108160727 on Build GRCh38
rs938819602 Chr.3: 108161003-108161003 on Build GRCh38
rs1009050685 Chr.3: 108160782-108160782 on Build GRCh38
rs576347633 Chr.3: 108161018-108161018 on Build GRCh38
rs556766664 Chr.3: 108160959-108160959 on Build GRCh38
rs1036888452 Chr.3: 108160942-108160942 on Build GRCh38
rs428321 Chr.3: 108161139-108161139 on Build GRCh38
rs948971093 Chr.3: 108160879-108160879 on Build GRCh38
rs974061856 Chr.3: 108160798-108160798 on Build GRCh38
rs554052377 Chr.3: 108160840-108160840 on Build GRCh38
rs892220064 Chr.3: 108161034-108161034 on Build GRCh38
rs368904361 Chr.3: 108161091-108161092 on Build GRCh38
rs544965595 Chr.3: 108161033-108161033 on Build GRCh38
rs927790352 Chr.3: 108160915-108160915 on Build GRCh38
rs533923515 Chr.3: 108160774-108160774 on Build GRCh38
rs762835283 Chr.3: 108160893-108160893 on Build GRCh38
rs527756663 Chr.3: 108161058-108161058 on Build GRCh38
rs190092074 Chr.3: 108160662-108160662 on Build GRCh38
rs541559801 Chr.3: 108161120-108161120 on Build GRCh38
rs552567022 Chr.3: 108160752-108160752 on Build GRCh38
rs889339994 Chr.3: 108160997-108160997 on Build GRCh38
rs564970770 Chr.3: 108161043-108161043 on Build GRCh38
rs62264145 Chr.3: 108160669-108160669 on Build GRCh38
rs867995519 Chr.3: 108160783-108160783 on Build GRCh38
rs1050626098 Chr.3: 108160991-108160991 on Build GRCh38
rs947855665 Chr.3: 108160889-108160889 on Build GRCh38
rs557809974 Chr.3: 108160725-108160725 on Build GRCh38
rs923595282 Chr.3: 108160804-108160804 on Build GRCh38
rs996797876 Chr.3: 108161142-108161142 on Build GRCh38
rs62264146 Chr.3: 108160883-108160883 on Build GRCh38
rs111415950 Chr.3: 108160988-108160988 on Build GRCh38
rs899164319 Chr.3: 108160959-108160959 on Build GRCh38
rs529043097 Chr.3: 108161140-108161140 on Build GRCh38
rs957724616 Chr.3: 108160817-108160817 on Build GRCh38
rs999273773 Chr.3: 108161075-108161075 on Build GRCh38
rs899170157 Chr.3: 108160711-108160711 on Build GRCh38
rs1029241194 Chr.3: 108160742-108160742 on Build GRCh38
rs1028643360 Chr.3: 108161152-108161152 on Build GRCh38
rs965130882 Chr.3: 108160785-108160785 on Build GRCh38
rs940752420 Chr.3: 108160931-108160931 on Build GRCh38
rs967956831 Chr.3: 108161045-108161045 on Build GRCh38
rs767739469 Chr.3: 108160842-108160843 on Build GRCh38
rs574224750 Chr.3: 108160845-108160845 on Build GRCh38
rs371817870 Chr.3: 108161099-108161099 on Build GRCh38
rs545259050 Chr.3: 108160698-108160698 on Build GRCh38
rs957672512 Chr.3: 108161138-108161138 on Build GRCh38

View Full Product Details