Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 121622555-121623059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO40
Entrez Gene ID
51725
Gene Name
F-box protein 40
Gene Aliases
FBX40
UniGene
Hs.272564
Gene Chromosome Location
Chr.3: 121593323-121630292 on Build GRCh38
Transcripts
NM_016298.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3120788 c.1532G>T p.C511F Chr.3: 121622961-121622961 on Build GRCh38
1728708 c.1201C>T p.Q401* Chr.3: 121622630-121622630 on Build GRCh38
6163023 c.1383C>A p.S461R Chr.3: 121622812-121622812 on Build GRCh38
6163025 c.1306G>T p.E436* Chr.3: 121622735-121622735 on Build GRCh38
3120778 c.1267T>C p.F423L Chr.3: 121622696-121622696 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139359257 Chr.3: 121622713-121622713 on Build GRCh38
rs764170628 Chr.3: 121622767-121622767 on Build GRCh38
rs765718437 Chr.3: 121622738-121622738 on Build GRCh38
rs750183156 Chr.3: 121622884-121622884 on Build GRCh38
rs758317619 Chr.3: 121622641-121622641 on Build GRCh38
rs774979444 Chr.3: 121622829-121622829 on Build GRCh38
rs971114864 Chr.3: 121622836-121622836 on Build GRCh38
rs200497241 Chr.3: 121622753-121622753 on Build GRCh38
rs762614859 Chr.3: 121622833-121622833 on Build GRCh38
rs978554717 Chr.3: 121622583-121622583 on Build GRCh38
rs768264149 Chr.3: 121622842-121622842 on Build GRCh38
rs370131239 Chr.3: 121622773-121622773 on Build GRCh38
rs765535473 Chr.3: 121622897-121622897 on Build GRCh38
rs751214057 Chr.3: 121622816-121622816 on Build GRCh38
rs1031472580 Chr.3: 121622852-121622852 on Build GRCh38
rs369070659 Chr.3: 121622712-121622712 on Build GRCh38
rs748888830 Chr.3: 121622965-121622965 on Build GRCh38
rs773685976 Chr.3: 121622686-121622686 on Build GRCh38
rs758487971 Chr.3: 121622900-121622900 on Build GRCh38
rs958185624 Chr.3: 121622875-121622875 on Build GRCh38
rs765814758 Chr.3: 121622813-121622813 on Build GRCh38
rs924069027 Chr.3: 121622587-121622587 on Build GRCh38
rs755024056 Chr.3: 121622682-121622682 on Build GRCh38
rs376716888 Chr.3: 121622702-121622702 on Build GRCh38
rs374623698 Chr.3: 121622700-121622700 on Build GRCh38
rs777764716 Chr.3: 121622930-121622930 on Build GRCh38
rs760342900 Chr.3: 121622607-121622607 on Build GRCh38
rs760584868 Chr.3: 121622978-121622978 on Build GRCh38
rs1015550336 Chr.3: 121622961-121622961 on Build GRCh38
rs1033518212 Chr.3: 121622927-121622927 on Build GRCh38
rs761773313 Chr.3: 121622859-121622859 on Build GRCh38
rs748646171 Chr.3: 121622583-121622583 on Build GRCh38
rs779809283 Chr.3: 121622958-121622958 on Build GRCh38
rs79455541 Chr.3: 121622628-121622628 on Build GRCh38
rs147462954 Chr.3: 121622797-121622797 on Build GRCh38
rs908218069 Chr.3: 121622746-121622746 on Build GRCh38
rs989962397 Chr.3: 121622885-121622885 on Build GRCh38
rs748307138 Chr.3: 121622789-121622789 on Build GRCh38
rs772094100 Chr.3: 121622971-121622971 on Build GRCh38
rs1001984813 Chr.3: 121622910-121622910 on Build GRCh38
rs201421773 Chr.3: 121622724-121622724 on Build GRCh38
rs149716243 Chr.3: 121622979-121622979 on Build GRCh38
rs537164095 Chr.3: 121622957-121622957 on Build GRCh38
rs144524810 Chr.3: 121623006-121623006 on Build GRCh38
rs780485745 Chr.3: 121622665-121622665 on Build GRCh38
rs772991278 Chr.3: 121622973-121622973 on Build GRCh38
rs750877265 Chr.3: 121623036-121623036 on Build GRCh38
rs372208925 Chr.3: 121622850-121622850 on Build GRCh38
rs752921762 Chr.3: 121622988-121622988 on Build GRCh38
rs932945597 Chr.3: 121622985-121622985 on Build GRCh38
rs866996277 Chr.3: 121623004-121623004 on Build GRCh38
rs778879981 Chr.3: 121622786-121622786 on Build GRCh38
rs760364901 Chr.3: 121622891-121622891 on Build GRCh38
rs757714918 Chr.3: 121622933-121622933 on Build GRCh38
rs369423447 Chr.3: 121622685-121622685 on Build GRCh38
rs766068748 Chr.3: 121622613-121622613 on Build GRCh38
rs757256594 Chr.3: 121623008-121623008 on Build GRCh38
rs747440455 Chr.3: 121622689-121622689 on Build GRCh38
rs953829652 Chr.3: 121623018-121623018 on Build GRCh38
rs139593536 Chr.3: 121622969-121622969 on Build GRCh38
rs200075101 Chr.3: 121623005-121623005 on Build GRCh38
rs762374838 Chr.3: 121622586-121622586 on Build GRCh38
rs556294432 Chr.3: 121622968-121622968 on Build GRCh38
rs138289002 Chr.3: 121622593-121622593 on Build GRCh38
rs775019825 Chr.3: 121622725-121622725 on Build GRCh38
rs143267823 Chr.3: 121622774-121622774 on Build GRCh38
rs752093992 Chr.3: 121622932-121622932 on Build GRCh38
rs770079979 Chr.3: 121622832-121622834 on Build GRCh38
rs752583608 Chr.3: 121622635-121622635 on Build GRCh38
rs1012093493 Chr.3: 121622599-121622599 on Build GRCh38
rs375954041 Chr.3: 121622772-121622772 on Build GRCh38
rs374431438 Chr.3: 121622981-121622981 on Build GRCh38
rs970533581 Chr.3: 121622646-121622646 on Build GRCh38
rs867887704 Chr.3: 121622594-121622594 on Build GRCh38
rs781782231 Chr.3: 121623035-121623035 on Build GRCh38
rs943288636 Chr.3: 121622651-121622651 on Build GRCh38
rs781583571 Chr.3: 121622801-121622801 on Build GRCh38
rs143415272 Chr.3: 121622663-121622663 on Build GRCh38
rs143931074 Chr.3: 121622799-121622799 on Build GRCh38
rs764748179 Chr.3: 121622627-121622627 on Build GRCh38
rs377117443 Chr.3: 121622622-121622622 on Build GRCh38
rs886197282 Chr.3: 121622705-121622705 on Build GRCh38
rs1037822357 Chr.3: 121623023-121623023 on Build GRCh38
rs767246396 Chr.3: 121622866-121622866 on Build GRCh38
rs764006927 Chr.3: 121622662-121622662 on Build GRCh38
rs374744484 Chr.3: 121622899-121622899 on Build GRCh38
rs764552176 Chr.3: 121622680-121622680 on Build GRCh38
rs149191160 Chr.3: 121622620-121622620 on Build GRCh38
rs748632933 Chr.3: 121622683-121622683 on Build GRCh38
rs780903030 Chr.3: 121622954-121622954 on Build GRCh38
rs367937919 Chr.3: 121623009-121623009 on Build GRCh38
rs200647871 Chr.3: 121622784-121622784 on Build GRCh38
rs150634995 Chr.3: 121622814-121622814 on Build GRCh38
rs763123334 Chr.3: 121623000-121623000 on Build GRCh38
rs138259890 Chr.3: 121622798-121622798 on Build GRCh38
rs184189985 Chr.3: 121622941-121622941 on Build GRCh38
rs749510221 Chr.3: 121622667-121622667 on Build GRCh38
rs1043441988 Chr.3: 121622883-121622883 on Build GRCh38
rs372367097 Chr.3: 121622756-121622756 on Build GRCh38

View Full Product Details