Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 122103370-122103871 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD86
Entrez Gene ID
942
Gene Name
CD86 molecule
Gene Aliases
B7-2, B7.2, B70, CD28LG2, LAB72
UniGene
Hs.171182
Gene Chromosome Location
Chr.3: 122055362-122121143 on Build GRCh38
Transcripts
NM_001206924.1 NM_001206925.1 NM_006889.4 NM_175862.4 NM_176892.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
254602 c.134C>T p.S45F Chr.3: 122103581-122103581 on Build GRCh38
6511434 c.350C>A p.P117H Chr.3: 122103797-122103797 on Build GRCh38
4945700 c.387A>G p.E129E Chr.3: 122103834-122103834 on Build GRCh38
6623928 c.217G>A p.E73K Chr.3: 122103664-122103664 on Build GRCh38
3845977 c.109G>A p.D37N Chr.3: 122103556-122103556 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370012716 Chr.3: 122103744-122103744 on Build GRCh38
rs761419249 Chr.3: 122103716-122103716 on Build GRCh38
rs773800947 Chr.3: 122103520-122103520 on Build GRCh38
rs184333802 Chr.3: 122103408-122103408 on Build GRCh38
rs747238810 Chr.3: 122103553-122103553 on Build GRCh38
rs753841632 Chr.3: 122103761-122103761 on Build GRCh38
rs1005895615 Chr.3: 122103598-122103598 on Build GRCh38
rs559114894 Chr.3: 122103615-122103615 on Build GRCh38
rs769924675 Chr.3: 122103664-122103664 on Build GRCh38
rs958118063 Chr.3: 122103759-122103759 on Build GRCh38
rs181748813 Chr.3: 122103717-122103717 on Build GRCh38
rs113527548 Chr.3: 122103421-122103421 on Build GRCh38
rs749851519 Chr.3: 122103815-122103815 on Build GRCh38
rs988531590 Chr.3: 122103447-122103447 on Build GRCh38
rs774603499 Chr.3: 122103698-122103698 on Build GRCh38
rs763707192 Chr.3: 122103789-122103789 on Build GRCh38
rs770052627 Chr.3: 122103811-122103811 on Build GRCh38
rs989527514 Chr.3: 122103762-122103762 on Build GRCh38
rs200704712 Chr.3: 122103846-122103846 on Build GRCh38
rs948647409 Chr.3: 122103475-122103475 on Build GRCh38
rs750002559 Chr.3: 122103741-122103741 on Build GRCh38
rs766230228 Chr.3: 122103409-122103409 on Build GRCh38
rs35582595 Chr.3: 122103451-122103452 on Build GRCh38
rs35276006 Chr.3: 122103694-122103694 on Build GRCh38
rs149667487 Chr.3: 122103518-122103518 on Build GRCh38
rs775704428 Chr.3: 122103812-122103812 on Build GRCh38
rs755636106 Chr.3: 122103742-122103742 on Build GRCh38
rs539519506 Chr.3: 122103530-122103530 on Build GRCh38
rs570961036 Chr.3: 122103507-122103507 on Build GRCh38
rs1042991536 Chr.3: 122103482-122103482 on Build GRCh38
rs1031151199 Chr.3: 122103721-122103721 on Build GRCh38
rs969643514 Chr.3: 122103841-122103841 on Build GRCh38
rs557303692 Chr.3: 122103453-122103453 on Build GRCh38
rs759696095 Chr.3: 122103651-122103651 on Build GRCh38
rs752867412 Chr.3: 122103503-122103504 on Build GRCh38
rs771729431 Chr.3: 122103783-122103783 on Build GRCh38
rs915271884 Chr.3: 122103456-122103457 on Build GRCh38
rs139011683 Chr.3: 122103768-122103768 on Build GRCh38
rs769261211 Chr.3: 122103837-122103837 on Build GRCh38
rs778624784 Chr.3: 122103773-122103773 on Build GRCh38
rs947281461 Chr.3: 122103457-122103457 on Build GRCh38
rs751702002 Chr.3: 122103751-122103751 on Build GRCh38
rs780604915 Chr.3: 122103470-122103470 on Build GRCh38
rs916624078 Chr.3: 122103843-122103843 on Build GRCh38
rs1030574173 Chr.3: 122103696-122103696 on Build GRCh38
rs367934963 Chr.3: 122103791-122103791 on Build GRCh38
rs866110754 Chr.3: 122103578-122103578 on Build GRCh38
rs766139636 Chr.3: 122103500-122103502 on Build GRCh38
rs1015889379 Chr.3: 122103606-122103606 on Build GRCh38
rs938518326 Chr.3: 122103534-122103534 on Build GRCh38
rs778676985 Chr.3: 122103545-122103545 on Build GRCh38
rs755571029 Chr.3: 122103509-122103509 on Build GRCh38
rs956833120 Chr.3: 122103445-122103445 on Build GRCh38
rs865917709 Chr.3: 122103617-122103617 on Build GRCh38
rs145491825 Chr.3: 122103780-122103780 on Build GRCh38
rs1047493821 Chr.3: 122103559-122103559 on Build GRCh38
rs763378254 Chr.3: 122103570-122103570 on Build GRCh38
rs371444245 Chr.3: 122103688-122103688 on Build GRCh38
rs771290530 Chr.3: 122103677-122103677 on Build GRCh38
rs753555067 Chr.3: 122103816-122103816 on Build GRCh38
rs376775826 Chr.3: 122103525-122103525 on Build GRCh38
rs931781143 Chr.3: 122103577-122103577 on Build GRCh38
rs561788422 Chr.3: 122103488-122103488 on Build GRCh38
rs916551151 Chr.3: 122103431-122103431 on Build GRCh38
rs146192611 Chr.3: 122103665-122103665 on Build GRCh38
rs553416970 Chr.3: 122103579-122103579 on Build GRCh38
rs1044318780 Chr.3: 122103505-122103505 on Build GRCh38
rs112508906 Chr.3: 122103555-122103555 on Build GRCh38

View Full Product Details