Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 130893904-130894405 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2C1
Entrez Gene ID
27032
Gene Name
ATPase secretory pathway Ca2+ transporting 1
Gene Aliases
ATP2C1A, BCPM, HHD, PMR1, SPCA1, hSPCA1
UniGene
Hs.584884
Gene Chromosome Location
Chr.3: 130850500-131016712 on Build GRCh38
Transcripts
NM_001001485.2 NM_001199179.1 NM_001199180.1 NM_001199181.1 NM_001199182.1 NM_001199183.1 NM_001199184.1 XM_005247354.2 XM_005247355.2 XM_005247356.2 XM_005247358.2 XM_017006164.1

SNP Detail

SNP ID Chromosome Location
rs940799158 Chr.3: 130894321-130894321 on Build GRCh38
rs75314013 Chr.3: 130894017-130894017 on Build GRCh38
rs1045902096 Chr.3: 130894094-130894094 on Build GRCh38
rs185723567 Chr.3: 130894190-130894190 on Build GRCh38
rs918699481 Chr.3: 130894065-130894065 on Build GRCh38
rs921882597 Chr.3: 130894252-130894252 on Build GRCh38
rs1033371313 Chr.3: 130893999-130893999 on Build GRCh38
rs1018475466 Chr.3: 130893946-130893946 on Build GRCh38
rs907390961 Chr.3: 130894338-130894338 on Build GRCh38
rs974688990 Chr.3: 130894248-130894248 on Build GRCh38
rs536541340 Chr.3: 130894182-130894182 on Build GRCh38
rs1039023515 Chr.3: 130894029-130894029 on Build GRCh38
rs1024501816 Chr.3: 130893972-130893972 on Build GRCh38
rs558489901 Chr.3: 130894324-130894324 on Build GRCh38
rs962597241 Chr.3: 130894207-130894207 on Build GRCh38
rs929821101 Chr.3: 130894251-130894251 on Build GRCh38
rs916515123 Chr.3: 130894194-130894194 on Build GRCh38
rs942730057 Chr.3: 130894260-130894260 on Build GRCh38
rs852218 Chr.3: 130894222-130894222 on Build GRCh38
rs1028202974 Chr.3: 130893937-130893937 on Build GRCh38
rs535665200 Chr.3: 130894168-130894168 on Build GRCh38
rs930650583 Chr.3: 130894262-130894262 on Build GRCh38
rs377638434 Chr.3: 130894135-130894135 on Build GRCh38
rs553708590 Chr.3: 130894112-130894112 on Build GRCh38
rs1038582125 Chr.3: 130894064-130894064 on Build GRCh38
rs965917240 Chr.3: 130894240-130894240 on Build GRCh38
rs578027991 Chr.3: 130894329-130894329 on Build GRCh38
rs915019928 Chr.3: 130894014-130894014 on Build GRCh38
rs28362548 Chr.3: 130893987-130893987 on Build GRCh38
rs930013646 Chr.3: 130894087-130894087 on Build GRCh38
rs1049140055 Chr.3: 130894071-130894071 on Build GRCh38
rs567367641 Chr.3: 130894148-130894148 on Build GRCh38
rs757658239 Chr.3: 130894126-130894126 on Build GRCh38
rs190101631 Chr.3: 130894382-130894382 on Build GRCh38
rs2685194 Chr.3: 130894345-130894345 on Build GRCh38
rs971652123 Chr.3: 130893977-130893977 on Build GRCh38
rs941203718 Chr.3: 130894057-130894057 on Build GRCh38
rs959162123 Chr.3: 130894052-130894052 on Build GRCh38
rs1041897255 Chr.3: 130894315-130894315 on Build GRCh38
rs539960254 Chr.3: 130893942-130893942 on Build GRCh38
rs907443363 Chr.3: 130894099-130894099 on Build GRCh38
rs999648029 Chr.3: 130894178-130894178 on Build GRCh38
rs1004996479 Chr.3: 130894089-130894089 on Build GRCh38
rs374167562 Chr.3: 130894041-130894041 on Build GRCh38
rs373283492 Chr.3: 130894195-130894195 on Build GRCh38
rs1049555524 Chr.3: 130894305-130894305 on Build GRCh38
rs977953547 Chr.3: 130894023-130894023 on Build GRCh38
rs868581280 Chr.3: 130894241-130894241 on Build GRCh38
rs944623749 Chr.3: 130894028-130894028 on Build GRCh38
rs752748384 Chr.3: 130894062-130894062 on Build GRCh38
rs918384010 Chr.3: 130894220-130894220 on Build GRCh38
rs1039649426 Chr.3: 130894272-130894272 on Build GRCh38
rs1034796590 Chr.3: 130894336-130894336 on Build GRCh38
rs912626698 Chr.3: 130894256-130894256 on Build GRCh38
rs1011758704 Chr.3: 130894155-130894155 on Build GRCh38
rs959355632 Chr.3: 130894009-130894009 on Build GRCh38
rs777195269 Chr.3: 130894107-130894107 on Build GRCh38
rs1022038115 Chr.3: 130894139-130894139 on Build GRCh38
rs886554611 Chr.3: 130894078-130894078 on Build GRCh38
rs141589843 Chr.3: 130894003-130894003 on Build GRCh38
rs756107296 Chr.3: 130894105-130894105 on Build GRCh38
rs1020847043 Chr.3: 130894166-130894166 on Build GRCh38
rs543616852 Chr.3: 130894218-130894218 on Build GRCh38
rs886610955 Chr.3: 130893941-130893941 on Build GRCh38
rs989254721 Chr.3: 130894012-130894012 on Build GRCh38
rs560526260 Chr.3: 130894353-130894353 on Build GRCh38
rs1056048829 Chr.3: 130894355-130894355 on Build GRCh38
rs907936332 Chr.3: 130894314-130894314 on Build GRCh38
rs966473212 Chr.3: 130894156-130894156 on Build GRCh38
rs1001755712 Chr.3: 130894101-130894101 on Build GRCh38
rs547462138 Chr.3: 130893967-130893967 on Build GRCh38
rs370802021 Chr.3: 130894172-130894172 on Build GRCh38
rs1011938121 Chr.3: 130894384-130894384 on Build GRCh38
rs977895985 Chr.3: 130894206-130894206 on Build GRCh38
rs911793295 Chr.3: 130894026-130894026 on Build GRCh38
rs921962413 Chr.3: 130894037-130894037 on Build GRCh38

View Full Product Details