Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 67739600-67740136 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNRHR
Entrez Gene ID
2798
Gene Name
gonadotropin releasing hormone receptor
Gene Aliases
GNRHR1, GRHR, HH7, LHRHR, LRHR
UniGene
Hs.407587
Gene Chromosome Location
Chr.4: 67737375-67756086 on Build GRCh38
Transcripts
NM_000406.2 NM_001012763.1

SNP Detail

SNP ID Chromosome Location
rs558008136 Chr.4: 67739953-67739953 on Build GRCh38
rs553086935 Chr.4: 67740030-67740030 on Build GRCh38
rs1014935119 Chr.4: 67739845-67739845 on Build GRCh38
rs998382130 Chr.4: 67739836-67739836 on Build GRCh38
rs1018058880 Chr.4: 67739753-67739753 on Build GRCh38
rs35683646 Chr.4: 67739671-67739671 on Build GRCh38
rs1011463554 Chr.4: 67739778-67739778 on Build GRCh38
rs17635850 Chr.4: 67739651-67739651 on Build GRCh38
rs550275721 Chr.4: 67739660-67739660 on Build GRCh38
rs777619461 Chr.4: 67739846-67739847 on Build GRCh38
rs903790261 Chr.4: 67739628-67739628 on Build GRCh38
rs960736276 Chr.4: 67739883-67739883 on Build GRCh38
rs943737420 Chr.4: 67740049-67740049 on Build GRCh38
rs935289054 Chr.4: 67740065-67740065 on Build GRCh38
rs113838900 Chr.4: 67739674-67739674 on Build GRCh38
rs1045668221 Chr.4: 67740027-67740027 on Build GRCh38
rs1004852117 Chr.4: 67739824-67739824 on Build GRCh38
rs1052343063 Chr.4: 67740007-67740007 on Build GRCh38
rs1046456106 Chr.4: 67739684-67739684 on Build GRCh38
rs770663987 Chr.4: 67739871-67739872 on Build GRCh38
rs1029319746 Chr.4: 67739858-67739858 on Build GRCh38
rs181403929 Chr.4: 67739629-67739629 on Build GRCh38
rs996382264 Chr.4: 67739762-67739762 on Build GRCh38
rs982557486 Chr.4: 67739895-67739895 on Build GRCh38
rs925170891 Chr.4: 67739967-67739967 on Build GRCh38
rs886059563 Chr.4: 67739648-67739648 on Build GRCh38
rs111894416 Chr.4: 67740095-67740095 on Build GRCh38
rs916696718 Chr.4: 67739911-67739911 on Build GRCh38
rs35610027 Chr.4: 67739678-67739678 on Build GRCh38
rs775879571 Chr.4: 67740033-67740033 on Build GRCh38
rs562430297 Chr.4: 67740104-67740104 on Build GRCh38
rs573227804 Chr.4: 67740069-67740069 on Build GRCh38
rs538069466 Chr.4: 67739917-67739917 on Build GRCh38
rs540398065 Chr.4: 67739966-67739967 on Build GRCh38
rs778576238 Chr.4: 67740110-67740110 on Build GRCh38
rs1018501549 Chr.4: 67739816-67739816 on Build GRCh38
rs566465436 Chr.4: 67739750-67739750 on Build GRCh38
rs535382437 Chr.4: 67739818-67739818 on Build GRCh38
rs949976135 Chr.4: 67740009-67740009 on Build GRCh38
rs1039843363 Chr.4: 67740089-67740089 on Build GRCh38
rs991706140 Chr.4: 67739930-67739930 on Build GRCh38
rs546237977 Chr.4: 67739716-67739716 on Build GRCh38
rs941154657 Chr.4: 67739647-67739647 on Build GRCh38
rs59079898 Chr.4: 67740113-67740113 on Build GRCh38
rs1052946219 Chr.4: 67739756-67739756 on Build GRCh38
rs886059564 Chr.4: 67739661-67739661 on Build GRCh38
rs774545256 Chr.4: 67739801-67739801 on Build GRCh38
rs377227833 Chr.4: 67739998-67739998 on Build GRCh38
rs112891890 Chr.4: 67740108-67740108 on Build GRCh38
rs374645440 Chr.4: 67740029-67740029 on Build GRCh38
rs999650034 Chr.4: 67739755-67739755 on Build GRCh38
rs555675298 Chr.4: 67739871-67739871 on Build GRCh38
rs1028299654 Chr.4: 67739787-67739787 on Build GRCh38
rs893982192 Chr.4: 67739769-67739769 on Build GRCh38
rs115861964 Chr.4: 67739985-67739985 on Build GRCh38
rs1056991119 Chr.4: 67739901-67739901 on Build GRCh38
rs550237485 Chr.4: 67739670-67739670 on Build GRCh38
rs893860070 Chr.4: 67740078-67740078 on Build GRCh38
rs951051395 Chr.4: 67739879-67739879 on Build GRCh38
rs35845954 Chr.4: 67739897-67739897 on Build GRCh38

View Full Product Details