Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99072495-99073002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADH5
Entrez Gene ID
128
Gene Name
alcohol dehydrogenase 5 (class III), chi polypeptide
Gene Aliases
ADH-3, ADHX, FALDH, FDH, GSH-FDH, GSNOR, HEL-S-60p
UniGene
Hs.78989
Gene Chromosome Location
Chr.4: 99070978-99088788 on Build GRCh38
Transcripts
NM_000671.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4841917 c.1083G>A p.L361L Chr.4: 99072590-99072590 on Build GRCh38
242854 c.1035delT p.F345fs*2 Chr.4: 99072638-99072638 on Build GRCh38
588789 c.1011C>A p.S337S Chr.4: 99072662-99072662 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs780652018 Chr.4: 99072536-99072538 on Build GRCh38
rs973237141 Chr.4: 99072805-99072805 on Build GRCh38
rs951905010 Chr.4: 99072947-99072947 on Build GRCh38
rs757467339 Chr.4: 99072687-99072687 on Build GRCh38
rs917199211 Chr.4: 99072881-99072884 on Build GRCh38
rs745875396 Chr.4: 99072560-99072560 on Build GRCh38
rs760168218 Chr.4: 99072635-99072635 on Build GRCh38
rs902202850 Chr.4: 99072801-99072801 on Build GRCh38
rs770405704 Chr.4: 99072685-99072685 on Build GRCh38
rs764236768 Chr.4: 99072673-99072673 on Build GRCh38
rs1041459714 Chr.4: 99072555-99072555 on Build GRCh38
rs1020691770 Chr.4: 99072784-99072784 on Build GRCh38
rs774009418 Chr.4: 99072592-99072592 on Build GRCh38
rs28730641 Chr.4: 99072894-99072894 on Build GRCh38
rs893556801 Chr.4: 99072735-99072735 on Build GRCh38
rs756087470 Chr.4: 99072721-99072721 on Build GRCh38
rs28730643 Chr.4: 99072682-99072682 on Build GRCh38
rs866144976 Chr.4: 99072934-99072934 on Build GRCh38
rs771771363 Chr.4: 99072627-99072627 on Build GRCh38
rs62325243 Chr.4: 99072548-99072548 on Build GRCh38
rs768427929 Chr.4: 99072754-99072754 on Build GRCh38
rs1034948950 Chr.4: 99072882-99072882 on Build GRCh38
rs536388397 Chr.4: 99072902-99072902 on Build GRCh38
rs771690889 Chr.4: 99072543-99072543 on Build GRCh38
rs749080716 Chr.4: 99072585-99072585 on Build GRCh38
rs904280816 Chr.4: 99072605-99072605 on Build GRCh38
rs772832874 Chr.4: 99072633-99072633 on Build GRCh38
rs748162259 Chr.4: 99072707-99072707 on Build GRCh38
rs992702349 Chr.4: 99072858-99072858 on Build GRCh38
rs747454345 Chr.4: 99072554-99072554 on Build GRCh38
rs904250338 Chr.4: 99072895-99072895 on Build GRCh38
rs1003188032 Chr.4: 99072855-99072855 on Build GRCh38
rs781129997 Chr.4: 99072649-99072650 on Build GRCh38
rs559039777 Chr.4: 99072536-99072536 on Build GRCh38
rs749062410 Chr.4: 99072747-99072747 on Build GRCh38
rs937199605 Chr.4: 99072846-99072846 on Build GRCh38
rs779930720 Chr.4: 99072729-99072729 on Build GRCh38
rs199893694 Chr.4: 99072573-99072573 on Build GRCh38
rs753034992 Chr.4: 99072653-99072653 on Build GRCh38
rs927117843 Chr.4: 99072824-99072824 on Build GRCh38
rs900725113 Chr.4: 99072693-99072693 on Build GRCh38
rs17595424 Chr.4: 99072718-99072718 on Build GRCh38
rs758639521 Chr.4: 99072935-99072935 on Build GRCh38
rs28730640 Chr.4: 99072902-99072902 on Build GRCh38
rs374091868 Chr.4: 99072569-99072569 on Build GRCh38
rs761230948 Chr.4: 99072606-99072606 on Build GRCh38
rs62325244 Chr.4: 99072636-99072636 on Build GRCh38
rs16996593 Chr.4: 99072614-99072614 on Build GRCh38
rs781308361 Chr.4: 99072704-99072704 on Build GRCh38
rs28730644 Chr.4: 99072535-99072535 on Build GRCh38
rs755147692 Chr.4: 99072556-99072558 on Build GRCh38
rs959244018 Chr.4: 99072890-99072890 on Build GRCh38
rs141054145 Chr.4: 99072762-99072762 on Build GRCh38
rs763326856 Chr.4: 99072657-99072657 on Build GRCh38
rs180808768 Chr.4: 99072520-99072520 on Build GRCh38

View Full Product Details