Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 140524066-140524568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELMOD2
Entrez Gene ID
255520
Gene Name
ELMO domain containing 2
Gene Aliases
9830169G11Rik
UniGene
Hs.450105
Gene Chromosome Location
Chr.4: 140524146-140553770 on Build GRCh38
Transcripts
NM_153702.3 XM_005262885.3 XM_005262886.2 XM_011531818.2 XM_011531819.2

SNP Detail

SNP ID Chromosome Location
rs933697540 Chr.4: 140524344-140524344 on Build GRCh38
rs531627054 Chr.4: 140524460-140524460 on Build GRCh38
rs542221517 Chr.4: 140524361-140524361 on Build GRCh38
rs935763939 Chr.4: 140524165-140524165 on Build GRCh38
rs1026669500 Chr.4: 140524350-140524350 on Build GRCh38
rs973508129 Chr.4: 140524216-140524216 on Build GRCh38
rs891484479 Chr.4: 140524185-140524185 on Build GRCh38
rs1030035310 Chr.4: 140524189-140524189 on Build GRCh38
rs553489890 Chr.4: 140524160-140524160 on Build GRCh38
rs1014358355 Chr.4: 140524520-140524520 on Build GRCh38
rs1052828055 Chr.4: 140524278-140524278 on Build GRCh38
rs917505301 Chr.4: 140524091-140524091 on Build GRCh38
rs944887177 Chr.4: 140524476-140524495 on Build GRCh38
rs1051264660 Chr.4: 140524448-140524448 on Build GRCh38
rs576308779 Chr.4: 140524244-140524244 on Build GRCh38
rs571370695 Chr.4: 140524473-140524473 on Build GRCh38
rs945357258 Chr.4: 140524253-140524254 on Build GRCh38
rs910914573 Chr.4: 140524464-140524464 on Build GRCh38
rs866628039 Chr.4: 140524198-140524198 on Build GRCh38
rs945667073 Chr.4: 140524391-140524391 on Build GRCh38
rs898360868 Chr.4: 140524383-140524383 on Build GRCh38
rs926192838 Chr.4: 140524435-140524435 on Build GRCh38
rs370776186 Chr.4: 140524137-140524137 on Build GRCh38
rs1044579725 Chr.4: 140524154-140524154 on Build GRCh38
rs149998004 Chr.4: 140524356-140524356 on Build GRCh38
rs978471643 Chr.4: 140524413-140524413 on Build GRCh38
rs1024790554 Chr.4: 140524523-140524523 on Build GRCh38
rs561201384 Chr.4: 140524444-140524444 on Build GRCh38
rs888120935 Chr.4: 140524500-140524500 on Build GRCh38
rs1049255325 Chr.4: 140524136-140524136 on Build GRCh38
rs950779984 Chr.4: 140524396-140524396 on Build GRCh38
rs943642190 Chr.4: 140524150-140524150 on Build GRCh38
rs536562609 Chr.4: 140524222-140524222 on Build GRCh38
rs993994305 Chr.4: 140524384-140524384 on Build GRCh38
rs1038277018 Chr.4: 140524370-140524370 on Build GRCh38
rs897096677 Chr.4: 140524325-140524325 on Build GRCh38
rs549985486 Chr.4: 140524095-140524095 on Build GRCh38
rs912983380 Chr.4: 140524331-140524331 on Build GRCh38
rs970438728 Chr.4: 140524420-140524420 on Build GRCh38
rs200597474 Chr.4: 140524530-140524530 on Build GRCh38
rs577313181 Chr.4: 140524258-140524258 on Build GRCh38
rs910733257 Chr.4: 140524217-140524217 on Build GRCh38
rs957566025 Chr.4: 140524437-140524437 on Build GRCh38
rs1013463826 Chr.4: 140524261-140524261 on Build GRCh38
rs898413068 Chr.4: 140524480-140524480 on Build GRCh38
rs549756339 Chr.4: 140524471-140524471 on Build GRCh38
rs554815702 Chr.4: 140524230-140524230 on Build GRCh38
rs1016782612 Chr.4: 140524205-140524205 on Build GRCh38
rs1033533015 Chr.4: 140524173-140524173 on Build GRCh38
rs955580978 Chr.4: 140524399-140524399 on Build GRCh38
rs964842396 Chr.4: 140524376-140524376 on Build GRCh38
rs1024320977 Chr.4: 140524414-140524414 on Build GRCh38
rs559100397 Chr.4: 140524255-140524255 on Build GRCh38
rs1042859051 Chr.4: 140524164-140524164 on Build GRCh38
rs936151903 Chr.4: 140524273-140524273 on Build GRCh38
rs192346193 Chr.4: 140524186-140524186 on Build GRCh38
rs904664224 Chr.4: 140524163-140524163 on Build GRCh38
rs913370908 Chr.4: 140524442-140524442 on Build GRCh38
rs1026439995 Chr.4: 140524394-140524394 on Build GRCh38
rs895265094 Chr.4: 140524449-140524449 on Build GRCh38
rs144312000 Chr.4: 140524482-140524482 on Build GRCh38
rs372161527 Chr.4: 140524495-140524495 on Build GRCh38
rs184548646 Chr.4: 140524513-140524513 on Build GRCh38
rs541468500 Chr.4: 140524285-140524285 on Build GRCh38
rs745675726 Chr.4: 140524118-140524143 on Build GRCh38
rs1027031729 Chr.4: 140524499-140524499 on Build GRCh38
rs970110753 Chr.4: 140524531-140524531 on Build GRCh38
rs889715257 Chr.4: 140524191-140524191 on Build GRCh38
rs373635792 Chr.4: 140524161-140524161 on Build GRCh38
rs917198068 Chr.4: 140524248-140524248 on Build GRCh38
rs988944358 Chr.4: 140524440-140524440 on Build GRCh38
rs532004534 Chr.4: 140524254-140524254 on Build GRCh38
rs962515215 Chr.4: 140524214-140524214 on Build GRCh38
rs376850171 Chr.4: 140524170-140524170 on Build GRCh38
rs571295725 Chr.4: 140524104-140524104 on Build GRCh38
rs568834496 Chr.4: 140524423-140524423 on Build GRCh38
rs532114984 Chr.4: 140524481-140524481 on Build GRCh38
rs930797247 Chr.4: 140524120-140524120 on Build GRCh38
rs557520298 Chr.4: 140524357-140524357 on Build GRCh38
rs368707781 Chr.4: 140524329-140524329 on Build GRCh38
rs919653821 Chr.4: 140524206-140524206 on Build GRCh38
rs752102717 Chr.4: 140524493-140524493 on Build GRCh38
rs1007187809 Chr.4: 140524196-140524196 on Build GRCh38
rs1014719383 Chr.4: 140524400-140524400 on Build GRCh38
rs555488160 Chr.4: 140524512-140524512 on Build GRCh38
rs976251507 Chr.4: 140524380-140524380 on Build GRCh38
rs879362714 Chr.4: 140524257-140524257 on Build GRCh38
rs919724443 Chr.4: 140524094-140524094 on Build GRCh38
rs1040323452 Chr.4: 140524152-140524152 on Build GRCh38
rs1024887527 Chr.4: 140524306-140524306 on Build GRCh38

View Full Product Details