Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 140543349-140543868 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELMOD2
Entrez Gene ID
255520
Gene Name
ELMO domain containing 2
Gene Aliases
9830169G11Rik
UniGene
Hs.450105
Gene Chromosome Location
Chr.4: 140524146-140553770 on Build GRCh38
Transcripts
NM_153702.3 XM_005262885.3 XM_005262886.2 XM_011531818.2 XM_011531819.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4584806 c.698C>G p.P233R Chr.4: 140543548-140543548 on Build GRCh38
6423754 c.733T>C p.Y245H Chr.4: 140543583-140543583 on Build GRCh38
4680804 c.656T>C p.L219P Chr.4: 140543506-140543506 on Build GRCh38
6011646 c.706C>T p.P236S Chr.4: 140543556-140543556 on Build GRCh38
4403066 c.682C>T p.L228F Chr.4: 140543532-140543532 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs115172148 Chr.4: 140543667-140543667 on Build GRCh38
rs141412212 Chr.4: 140543553-140543553 on Build GRCh38
rs747521483 Chr.4: 140543761-140543761 on Build GRCh38
rs577667364 Chr.4: 140543692-140543692 on Build GRCh38
rs775551250 Chr.4: 140543610-140543610 on Build GRCh38
rs757655589 Chr.4: 140543564-140543564 on Build GRCh38
rs184171971 Chr.4: 140543650-140543650 on Build GRCh38
rs546897805 Chr.4: 140543741-140543741 on Build GRCh38
rs188722064 Chr.4: 140543714-140543714 on Build GRCh38
rs767196649 Chr.4: 140543547-140543547 on Build GRCh38
rs747242417 Chr.4: 140543611-140543611 on Build GRCh38
rs775674090 Chr.4: 140543416-140543416 on Build GRCh38
rs773028539 Chr.4: 140543625-140543625 on Build GRCh38
rs73856651 Chr.4: 140543426-140543426 on Build GRCh38
rs1040046222 Chr.4: 140543782-140543782 on Build GRCh38
rs562649947 Chr.4: 140543691-140543691 on Build GRCh38
rs146583936 Chr.4: 140543763-140543766 on Build GRCh38
rs755729412 Chr.4: 140543556-140543556 on Build GRCh38
rs745484892 Chr.4: 140543612-140543613 on Build GRCh38
rs754515295 Chr.4: 140543530-140543530 on Build GRCh38
rs561803357 Chr.4: 140543751-140543751 on Build GRCh38
rs1041858810 Chr.4: 140543397-140543397 on Build GRCh38
rs949434840 Chr.4: 140543387-140543387 on Build GRCh38
rs1045174236 Chr.4: 140543757-140543757 on Build GRCh38
rs150835853 Chr.4: 140543567-140543567 on Build GRCh38
rs772895784 Chr.4: 140543486-140543486 on Build GRCh38
rs545044901 Chr.4: 140543695-140543695 on Build GRCh38
rs796052155 Chr.4: 140543524-140543524 on Build GRCh38
rs200049901 Chr.4: 140543557-140543557 on Build GRCh38
rs985835981 Chr.4: 140543586-140543586 on Build GRCh38
rs182554775 Chr.4: 140543755-140543755 on Build GRCh38
rs796131869 Chr.4: 140543606-140543606 on Build GRCh38
rs191195969 Chr.4: 140543724-140543724 on Build GRCh38
rs768808970 Chr.4: 140543619-140543619 on Build GRCh38
rs772186748 Chr.4: 140543414-140543414 on Build GRCh38
rs767314304 Chr.4: 140543687-140543687 on Build GRCh38
rs746430787 Chr.4: 140543594-140543594 on Build GRCh38
rs749642183 Chr.4: 140543560-140543560 on Build GRCh38
rs903334956 Chr.4: 140543837-140543837 on Build GRCh38
rs772133657 Chr.4: 140543600-140543600 on Build GRCh38
rs892684534 Chr.4: 140543532-140543532 on Build GRCh38
rs200487590 Chr.4: 140543462-140543462 on Build GRCh38
rs762900314 Chr.4: 140543493-140543493 on Build GRCh38
rs1009758064 Chr.4: 140543543-140543543 on Build GRCh38
rs148286208 Chr.4: 140543512-140543512 on Build GRCh38
rs919198843 Chr.4: 140543674-140543674 on Build GRCh38
rs936700044 Chr.4: 140543775-140543775 on Build GRCh38
rs868285754 Chr.4: 140543785-140543785 on Build GRCh38
rs778689326 Chr.4: 140543609-140543610 on Build GRCh38
rs1054174512 Chr.4: 140543830-140543830 on Build GRCh38
rs751543156 Chr.4: 140543589-140543589 on Build GRCh38
rs945124015 Chr.4: 140543797-140543797 on Build GRCh38

View Full Product Details