Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 71483733-71484256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BDP1
Entrez Gene ID
55814
Gene Name
B double prime 1, subunit of RNA polymerase III transcription initiation factor IIIB
Gene Aliases
HSA238520, TAF3B1, TFC5, TFIIIB'', TFIIIB150, TFIIIB90, TFNR
UniGene
Hs.258272
Gene Chromosome Location
Chr.5: 71455615-71578288 on Build GRCh38
Transcripts
NM_018429.2 XM_011543511.2 XM_011543512.2 XM_017009630.1 XM_017009631.1 XM_017009632.1 XM_017009633.1 XM_017009634.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4802371 c.1030G>A p.E344K Chr.5: 71483857-71483857 on Build GRCh38
4952624 c.1058A>G p.D353G Chr.5: 71483885-71483885 on Build GRCh38
7294900 c.1047A>C p.G349G Chr.5: 71483874-71483874 on Build GRCh38
6380455 c.1051A>G p.R351G Chr.5: 71483878-71483878 on Build GRCh38
5356715 c.1019A>C p.K340T Chr.5: 71483846-71483846 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs796163735 Chr.5: 71484221-71484221 on Build GRCh38
rs759571437 Chr.5: 71483854-71483854 on Build GRCh38
rs11953420 Chr.5: 71483784-71483784 on Build GRCh38
rs760393818 Chr.5: 71483818-71483818 on Build GRCh38
rs765235493 Chr.5: 71483856-71483856 on Build GRCh38
rs975573066 Chr.5: 71483914-71483914 on Build GRCh38
rs756854042 Chr.5: 71483874-71483874 on Build GRCh38
rs757934216 Chr.5: 71483863-71483863 on Build GRCh38
rs187322416 Chr.5: 71484209-71484209 on Build GRCh38
rs937876315 Chr.5: 71484055-71484055 on Build GRCh38
rs769403915 Chr.5: 71484093-71484093 on Build GRCh38
rs992199057 Chr.5: 71484046-71484046 on Build GRCh38
rs1008192477 Chr.5: 71484134-71484134 on Build GRCh38
rs887123913 Chr.5: 71484133-71484133 on Build GRCh38
rs912626968 Chr.5: 71483872-71483872 on Build GRCh38
rs909099162 Chr.5: 71484088-71484088 on Build GRCh38
rs763757024 Chr.5: 71483829-71483831 on Build GRCh38
rs998372848 Chr.5: 71483836-71483836 on Build GRCh38
rs1014694413 Chr.5: 71483792-71483792 on Build GRCh38
rs916447735 Chr.5: 71484067-71484067 on Build GRCh38
rs781663061 Chr.5: 71483884-71483884 on Build GRCh38
rs545842046 Chr.5: 71484102-71484102 on Build GRCh38
rs368980687 Chr.5: 71484187-71484187 on Build GRCh38
rs978466831 Chr.5: 71484023-71484023 on Build GRCh38
rs780495313 Chr.5: 71483895-71483895 on Build GRCh38
rs80104574 Chr.5: 71484072-71484072 on Build GRCh38
rs774504353 Chr.5: 71483814-71483814 on Build GRCh38
rs542530993 Chr.5: 71484043-71484043 on Build GRCh38
rs1029798481 Chr.5: 71483873-71483873 on Build GRCh38
rs768662515 Chr.5: 71483902-71483902 on Build GRCh38
rs968101853 Chr.5: 71483982-71483982 on Build GRCh38
rs35279479 Chr.5: 71483841-71483842 on Build GRCh38
rs964151583 Chr.5: 71484163-71484163 on Build GRCh38
rs1019718682 Chr.5: 71484147-71484147 on Build GRCh38
rs375454512 Chr.5: 71484091-71484091 on Build GRCh38
rs763966295 Chr.5: 71483886-71483886 on Build GRCh38
rs913585668 Chr.5: 71484034-71484041 on Build GRCh38
rs761595562 Chr.5: 71483997-71483997 on Build GRCh38
rs751155184 Chr.5: 71483870-71483870 on Build GRCh38
rs756650377 Chr.5: 71483887-71483887 on Build GRCh38
rs753565272 Chr.5: 71483828-71483828 on Build GRCh38
rs777584097 Chr.5: 71483865-71483865 on Build GRCh38
rs1025633430 Chr.5: 71483785-71483785 on Build GRCh38
rs538041640 Chr.5: 71483987-71483987 on Build GRCh38
rs762985916 Chr.5: 71483791-71483791 on Build GRCh38
rs200094670 Chr.5: 71483819-71483819 on Build GRCh38
rs963317838 Chr.5: 71483783-71483784 on Build GRCh38
rs752288408 Chr.5: 71483857-71483857 on Build GRCh38
rs531473972 Chr.5: 71484228-71484228 on Build GRCh38
rs141064218 Chr.5: 71484071-71484071 on Build GRCh38
rs375768639 Chr.5: 71484185-71484185 on Build GRCh38
rs901597119 Chr.5: 71484108-71484108 on Build GRCh38
rs772812330 Chr.5: 71484126-71484126 on Build GRCh38
rs112266388 Chr.5: 71483920-71483920 on Build GRCh38
rs533967135 Chr.5: 71483809-71483809 on Build GRCh38
rs958558717 Chr.5: 71484231-71484231 on Build GRCh38
rs184806710 Chr.5: 71484036-71484036 on Build GRCh38
rs753385045 Chr.5: 71483897-71483898 on Build GRCh38
rs772165191 Chr.5: 71483925-71483925 on Build GRCh38
rs954192918 Chr.5: 71483896-71483896 on Build GRCh38
rs369727615 Chr.5: 71483921-71483921 on Build GRCh38
rs749389312 Chr.5: 71483897-71483897 on Build GRCh38
rs776139114 Chr.5: 71483879-71483879 on Build GRCh38
rs566543210 Chr.5: 71483795-71483795 on Build GRCh38
rs997715743 Chr.5: 71484158-71484158 on Build GRCh38
rs1016605397 Chr.5: 71483778-71483778 on Build GRCh38
rs147328721 Chr.5: 71484038-71484038 on Build GRCh38

View Full Product Details