Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 79291038-79291538 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JMY
Entrez Gene ID
133746
Gene Name
junction mediating and regulatory protein, p53 cofactor
Gene Aliases
WHAMM2, WHDC1L3
UniGene
Hs.482605
Gene Chromosome Location
Chr.5: 79236102-79327215 on Build GRCh38
Transcripts
NM_152405.4 XM_005248430.2 XM_011543155.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3618062 c.452C>T p.A151V Chr.5: 79291286-79291286 on Build GRCh38
4910429 c.1430T>C p.M477T Chr.5: 79291202-79291202 on Build GRCh38
3618061 c.1514C>T p.A505V Chr.5: 79291286-79291286 on Build GRCh38
6242764 c.1464_1465delTT p.L489fs*6 Chr.5: 79291236-79291237 on Build GRCh38
4910430 c.368T>C p.M123T Chr.5: 79291202-79291202 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs73769609 Chr.5: 79291106-79291106 on Build GRCh38
rs887453262 Chr.5: 79291505-79291505 on Build GRCh38
rs373326661 Chr.5: 79291265-79291265 on Build GRCh38
rs370651121 Chr.5: 79291370-79291370 on Build GRCh38
rs749993674 Chr.5: 79291169-79291169 on Build GRCh38
rs779369887 Chr.5: 79291198-79291198 on Build GRCh38
rs774331803 Chr.5: 79291114-79291114 on Build GRCh38
rs775231137 Chr.5: 79291221-79291221 on Build GRCh38
rs755251040 Chr.5: 79291195-79291195 on Build GRCh38
rs373926091 Chr.5: 79291188-79291188 on Build GRCh38
rs756965069 Chr.5: 79291141-79291141 on Build GRCh38
rs921500856 Chr.5: 79291470-79291470 on Build GRCh38
rs369214683 Chr.5: 79291264-79291264 on Build GRCh38
rs763961379 Chr.5: 79291125-79291125 on Build GRCh38
rs576453208 Chr.5: 79291453-79291453 on Build GRCh38
rs1023889094 Chr.5: 79291489-79291489 on Build GRCh38
rs1033038801 Chr.5: 79291064-79291064 on Build GRCh38
rs944214054 Chr.5: 79291454-79291454 on Build GRCh38
rs533530811 Chr.5: 79291382-79291382 on Build GRCh38
rs780473079 Chr.5: 79291187-79291187 on Build GRCh38
rs887493187 Chr.5: 79291248-79291248 on Build GRCh38
rs556050361 Chr.5: 79291475-79291475 on Build GRCh38
rs995823282 Chr.5: 79291189-79291189 on Build GRCh38
rs779129352 Chr.5: 79291317-79291317 on Build GRCh38
rs376098096 Chr.5: 79291235-79291235 on Build GRCh38
rs770814042 Chr.5: 79291110-79291110 on Build GRCh38
rs772475308 Chr.5: 79291212-79291212 on Build GRCh38
rs74709243 Chr.5: 79291205-79291205 on Build GRCh38
rs538435754 Chr.5: 79291220-79291220 on Build GRCh38
rs769966546 Chr.5: 79291079-79291080 on Build GRCh38
rs118124886 Chr.5: 79291277-79291277 on Build GRCh38
rs776955178 Chr.5: 79291237-79291237 on Build GRCh38
rs1032322232 Chr.5: 79291428-79291428 on Build GRCh38
rs377515393 Chr.5: 79291276-79291276 on Build GRCh38
rs758685826 Chr.5: 79291324-79291324 on Build GRCh38
rs749098248 Chr.5: 79291516-79291516 on Build GRCh38
rs759488908 Chr.5: 79291339-79291340 on Build GRCh38
rs16876619 Chr.5: 79291474-79291474 on Build GRCh38
rs1039988879 Chr.5: 79291146-79291146 on Build GRCh38
rs748599442 Chr.5: 79291321-79291321 on Build GRCh38
rs182152438 Chr.5: 79291100-79291100 on Build GRCh38
rs751122838 Chr.5: 79291139-79291139 on Build GRCh38
rs185590207 Chr.5: 79291432-79291432 on Build GRCh38
rs749367059 Chr.5: 79291098-79291098 on Build GRCh38
rs763755629 Chr.5: 79291238-79291238 on Build GRCh38
rs756786743 Chr.5: 79291283-79291283 on Build GRCh38
rs958288257 Chr.5: 79291082-79291082 on Build GRCh38
rs10043008 Chr.5: 79291091-79291091 on Build GRCh38
rs758905889 Chr.5: 79291123-79291123 on Build GRCh38
rs773469563 Chr.5: 79291088-79291088 on Build GRCh38
rs191410938 Chr.5: 79291498-79291498 on Build GRCh38
rs1039917999 Chr.5: 79291469-79291469 on Build GRCh38
rs766983260 Chr.5: 79291142-79291142 on Build GRCh38
rs17234734 Chr.5: 79291312-79291312 on Build GRCh38
rs773189435 Chr.5: 79291086-79291087 on Build GRCh38
rs925321199 Chr.5: 79291399-79291399 on Build GRCh38
rs1005005203 Chr.5: 79291517-79291517 on Build GRCh38
rs747149593 Chr.5: 79291218-79291218 on Build GRCh38
rs576789166 Chr.5: 79291217-79291217 on Build GRCh38
rs999950882 Chr.5: 79291396-79291396 on Build GRCh38
rs956984761 Chr.5: 79291419-79291419 on Build GRCh38
rs369155949 Chr.5: 79291172-79291172 on Build GRCh38
rs1048732911 Chr.5: 79291485-79291485 on Build GRCh38
rs774241777 Chr.5: 79291249-79291249 on Build GRCh38
rs754262682 Chr.5: 79291285-79291285 on Build GRCh38

View Full Product Details