Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 134306128-134306676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKL3
Entrez Gene ID
51265
Gene Name
cyclin dependent kinase like 3
Gene Aliases
NKIAMRE
UniGene
Hs.719926
Gene Chromosome Location
Chr.5: 134298424-134371159 on Build GRCh38
Transcripts
NM_001113575.1 NM_001300853.1 XM_011543429.2 XM_011543439.2 XM_011543444.2 XM_011543445.2 XM_011543447.2 XM_017009524.1 XM_017009525.1 XM_017009526.1 XM_017009527.1 XM_017009528.1 XM_017009529.1 XM_017009530.1 XM_017009531.1 XM_017009532.1 XM_017009533.1 XM_017009534.1 XM_017009535.1 XM_017009536.1 XM_017009537.1 XM_017009538.1 XM_017009539.1 XM_017009540.1 XM_017009541.1 XM_017009542.1 XM_017009543.1 XM_017009544.1 XM_017009545.1 XM_017009546.1 XM_017009547.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6529595 c.1451C>G p.P484R Chr.5: 134306616-134306616 on Build GRCh38
5011482 c.1438G>C p.E480Q Chr.5: 134306629-134306629 on Build GRCh38
4344409 c.1391G>A p.R464H Chr.5: 134306676-134306676 on Build GRCh38
5011484 c.553G>C p.E185Q Chr.5: 134306629-134306629 on Build GRCh38
6529596 c.1451C>G p.P484R Chr.5: 134306616-134306616 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767634380 Chr.5: 134306608-134306608 on Build GRCh38
rs984748879 Chr.5: 134306331-134306332 on Build GRCh38
rs142248452 Chr.5: 134306635-134306635 on Build GRCh38
rs369418961 Chr.5: 134306621-134306621 on Build GRCh38
rs1042148448 Chr.5: 134306340-134306340 on Build GRCh38
rs1017400538 Chr.5: 134306186-134306186 on Build GRCh38
rs775945416 Chr.5: 134306567-134306567 on Build GRCh38
rs372235805 Chr.5: 134306575-134306575 on Build GRCh38
rs972854247 Chr.5: 134306242-134306242 on Build GRCh38
rs902153767 Chr.5: 134306506-134306509 on Build GRCh38
rs751523195 Chr.5: 134306550-134306550 on Build GRCh38
rs370723160 Chr.5: 134306281-134306281 on Build GRCh38
rs17167441 Chr.5: 134306164-134306164 on Build GRCh38
rs532251891 Chr.5: 134306473-134306473 on Build GRCh38
rs945109516 Chr.5: 134306331-134306331 on Build GRCh38
rs190049871 Chr.5: 134306465-134306465 on Build GRCh38
rs1021034896 Chr.5: 134306175-134306175 on Build GRCh38
rs750146784 Chr.5: 134306429-134306429 on Build GRCh38
rs897185754 Chr.5: 134306476-134306476 on Build GRCh38
rs566329685 Chr.5: 134306274-134306274 on Build GRCh38
rs937638894 Chr.5: 134306318-134306318 on Build GRCh38
rs778319248 Chr.5: 134306374-134306374 on Build GRCh38
rs933845049 Chr.5: 134306381-134306381 on Build GRCh38
rs1042058813 Chr.5: 134306504-134306504 on Build GRCh38
rs1057319826 Chr.5: 134306355-134306355 on Build GRCh38
rs1006711172 Chr.5: 134306555-134306555 on Build GRCh38
rs551286369 Chr.5: 134306200-134306200 on Build GRCh38
rs750477467 Chr.5: 134306617-134306617 on Build GRCh38
rs961446562 Chr.5: 134306223-134306223 on Build GRCh38
rs1019371791 Chr.5: 134306562-134306562 on Build GRCh38
rs980734818 Chr.5: 134306279-134306279 on Build GRCh38
rs1001152005 Chr.5: 134306188-134306188 on Build GRCh38
rs561588023 Chr.5: 134306637-134306637 on Build GRCh38
rs989150853 Chr.5: 134306322-134306322 on Build GRCh38
rs1024462896 Chr.5: 134306273-134306273 on Build GRCh38
rs1001715976 Chr.5: 134306552-134306552 on Build GRCh38
rs763438786 Chr.5: 134306570-134306570 on Build GRCh38
rs17165391 Chr.5: 134306639-134306639 on Build GRCh38
rs765024560 Chr.5: 134306371-134306371 on Build GRCh38
rs770434972 Chr.5: 134306559-134306559 on Build GRCh38
rs994243000 Chr.5: 134306528-134306528 on Build GRCh38
rs147263328 Chr.5: 134306452-134306452 on Build GRCh38
rs918543920 Chr.5: 134306300-134306300 on Build GRCh38
rs1024410545 Chr.5: 134306541-134306541 on Build GRCh38
rs751779184 Chr.5: 134306581-134306581 on Build GRCh38
rs779991771 Chr.5: 134306629-134306629 on Build GRCh38
rs556488563 Chr.5: 134306478-134306478 on Build GRCh38
rs966663168 Chr.5: 134306180-134306180 on Build GRCh38
rs1005916065 Chr.5: 134306454-134306454 on Build GRCh38
rs182207397 Chr.5: 134306547-134306547 on Build GRCh38
rs889696842 Chr.5: 134306543-134306543 on Build GRCh38
rs140840904 Chr.5: 134306542-134306542 on Build GRCh38
rs914945749 Chr.5: 134306327-134306327 on Build GRCh38
rs1050069241 Chr.5: 134306451-134306451 on Build GRCh38
rs1032703599 Chr.5: 134306543-134306547 on Build GRCh38
rs545480997 Chr.5: 134306544-134306544 on Build GRCh38
rs533876631 Chr.5: 134306328-134306328 on Build GRCh38
rs1032194313 Chr.5: 134306213-134306213 on Build GRCh38
rs972716279 Chr.5: 134306286-134306286 on Build GRCh38
rs186119803 Chr.5: 134306596-134306596 on Build GRCh38
rs117590960 Chr.5: 134306475-134306475 on Build GRCh38
rs999709339 Chr.5: 134306405-134306405 on Build GRCh38
rs868124398 Chr.5: 134306622-134306622 on Build GRCh38
rs927568767 Chr.5: 134306283-134306283 on Build GRCh38

View Full Product Details