Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150276405-150276907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APH1A
Entrez Gene ID
51107
Gene Name
aph-1 homolog A, gamma-secretase subunit
Gene Aliases
6530402N02Rik, APH-1, APH-1A, CGI-78
UniGene
Hs.108408
Gene Chromosome Location
Chr.1: 150265399-150282291 on Build GRCh38
Transcripts
XM_017001417.1 NM_001301039.1 NM_001301040.1 NM_001301041.1 NM_001301042.1 NM_024579.3 XM_011509984.2

Gene Symbol
C1orf54
Entrez Gene ID
79630
Gene Name
chromosome 1 open reading frame 54
Gene Aliases
-
UniGene
Hs.91283
Gene Chromosome Location
Chr.1: 150272284-150280927 on Build GRCh38
Transcripts
XM_017001417.1 NM_001301039.1 NM_001301040.1 NM_001301041.1 NM_001301042.1 NM_024579.3 XM_011509984.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4818452 c.223G>A p.E75K Chr.1: 150276555-150276555 on Build GRCh38
7358306 c.253G>A p.A85T Chr.1: 150276585-150276585 on Build GRCh38
300576 c.294G>A p.T98T Chr.1: 150276626-150276626 on Build GRCh38
7358307 c.253G>A p.A85T Chr.1: 150276585-150276585 on Build GRCh38
5843536 c.283C>T p.P95S Chr.1: 150276615-150276615 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146541375 Chr.1: 150276837-150276837 on Build GRCh38
rs782075502 Chr.1: 150276678-150276678 on Build GRCh38
rs782329187 Chr.1: 150276655-150276655 on Build GRCh38
rs782800611 Chr.1: 150276682-150276682 on Build GRCh38
rs376420729 Chr.1: 150276863-150276863 on Build GRCh38
rs782295525 Chr.1: 150276660-150276660 on Build GRCh38
rs148384729 Chr.1: 150276595-150276595 on Build GRCh38
rs782438074 Chr.1: 150276586-150276586 on Build GRCh38
rs951962468 Chr.1: 150276754-150276754 on Build GRCh38
rs141645512 Chr.1: 150276626-150276626 on Build GRCh38
rs146852269 Chr.1: 150276452-150276452 on Build GRCh38
rs782611473 Chr.1: 150276650-150276650 on Build GRCh38
rs587652241 Chr.1: 150276773-150276773 on Build GRCh38
rs782785681 Chr.1: 150276625-150276625 on Build GRCh38
rs782699079 Chr.1: 150276551-150276551 on Build GRCh38
rs782317173 Chr.1: 150276582-150276582 on Build GRCh38
rs1022713665 Chr.1: 150276649-150276649 on Build GRCh38
rs782328140 Chr.1: 150276519-150276519 on Build GRCh38
rs375719934 Chr.1: 150276533-150276533 on Build GRCh38
rs782183092 Chr.1: 150276652-150276652 on Build GRCh38
rs587652473 Chr.1: 150276429-150276429 on Build GRCh38
rs781975228 Chr.1: 150276505-150276505 on Build GRCh38
rs782541055 Chr.1: 150276633-150276633 on Build GRCh38
rs376397420 Chr.1: 150276484-150276484 on Build GRCh38
rs372422740 Chr.1: 150276512-150276512 on Build GRCh38
rs782050212 Chr.1: 150276493-150276494 on Build GRCh38
rs781883482 Chr.1: 150276604-150276605 on Build GRCh38
rs74968977 Chr.1: 150276557-150276557 on Build GRCh38
rs781995219 Chr.1: 150276661-150276661 on Build GRCh38
rs781974344 Chr.1: 150276488-150276488 on Build GRCh38
rs782074662 Chr.1: 150276663-150276663 on Build GRCh38
rs781899332 Chr.1: 150276621-150276621 on Build GRCh38
rs782098840 Chr.1: 150276550-150276550 on Build GRCh38
rs782542523 Chr.1: 150276555-150276555 on Build GRCh38
rs782247549 Chr.1: 150276642-150276642 on Build GRCh38
rs782596838 Chr.1: 150276567-150276567 on Build GRCh38
rs587708459 Chr.1: 150276434-150276434 on Build GRCh38
rs781928993 Chr.1: 150276674-150276674 on Build GRCh38
rs782162901 Chr.1: 150276531-150276531 on Build GRCh38
rs782731510 Chr.1: 150276667-150276667 on Build GRCh38
rs782064288 Chr.1: 150276610-150276610 on Build GRCh38
rs1026194723 Chr.1: 150276717-150276717 on Build GRCh38
rs782813326 Chr.1: 150276659-150276659 on Build GRCh38
rs1049170024 Chr.1: 150276559-150276559 on Build GRCh38
rs144321018 Chr.1: 150276723-150276723 on Build GRCh38
rs587678315 Chr.1: 150276710-150276710 on Build GRCh38
rs782675937 Chr.1: 150276638-150276638 on Build GRCh38
rs781911800 Chr.1: 150276684-150276684 on Build GRCh38
rs915551786 Chr.1: 150276830-150276830 on Build GRCh38
rs781811900 Chr.1: 150276631-150276631 on Build GRCh38
rs965866895 Chr.1: 150276689-150276689 on Build GRCh38
rs11205354 Chr.1: 150276693-150276693 on Build GRCh38
rs892689820 Chr.1: 150276611-150276611 on Build GRCh38
rs782106866 Chr.1: 150276624-150276624 on Build GRCh38
rs782021460 Chr.1: 150276523-150276523 on Build GRCh38
rs782575298 Chr.1: 150276630-150276630 on Build GRCh38
rs1045267414 Chr.1: 150276585-150276585 on Build GRCh38
rs782784545 Chr.1: 150276614-150276614 on Build GRCh38
rs953577963 Chr.1: 150276440-150276440 on Build GRCh38
rs781850515 Chr.1: 150276564-150276564 on Build GRCh38
rs142605908 Chr.1: 150276544-150276544 on Build GRCh38
rs1018955749 Chr.1: 150276685-150276685 on Build GRCh38
rs781984719 Chr.1: 150276594-150276594 on Build GRCh38
rs928330878 Chr.1: 150276874-150276874 on Build GRCh38
rs782237284 Chr.1: 150276578-150276578 on Build GRCh38
rs782258005 Chr.1: 150276491-150276491 on Build GRCh38
rs139591160 Chr.1: 150276596-150276596 on Build GRCh38
rs781919681 Chr.1: 150276608-150276608 on Build GRCh38
rs185521424 Chr.1: 150276565-150276565 on Build GRCh38
rs781802591 Chr.1: 150276553-150276553 on Build GRCh38
rs782351170 Chr.1: 150276497-150276497 on Build GRCh38
rs782385586 Chr.1: 150276646-150276646 on Build GRCh38
rs984499059 Chr.1: 150276814-150276814 on Build GRCh38

View Full Product Details