Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 24806321-24806834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM65B
Entrez Gene ID
9750
Gene Name
family with sequence similarity 65 member B
Gene Aliases
C6orf32, DFNB104, DIFF40, DIFF48, MYONAP, PL48
UniGene
Hs.559459
Gene Chromosome Location
Chr.6: 24804281-25042288 on Build GRCh38
Transcripts
NM_001286445.1 NM_014722.3 XM_006715275.2 XM_006715277.1 XM_006715279.2 XM_011515007.1 XM_011515008.2 XM_011515009.1 XM_011515010.2 XM_011515012.1 XM_017011523.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3784477 c.3175C>T p.R1059C Chr.6: 24806405-24806405 on Build GRCh38
1442495 c.3119G>A p.R1040Q Chr.6: 24806461-24806461 on Build GRCh38
4487191 c.3088C>T p.R1030* Chr.6: 24806429-24806429 on Build GRCh38
3784478 c.3112C>T p.R1038C Chr.6: 24806405-24806405 on Build GRCh38
1442494 c.3168C>T p.V1056V Chr.6: 24806412-24806412 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773105127 Chr.6: 24806515-24806515 on Build GRCh38
rs549011780 Chr.6: 24806504-24806504 on Build GRCh38
rs761711347 Chr.6: 24806365-24806365 on Build GRCh38
rs756843533 Chr.6: 24806423-24806423 on Build GRCh38
rs928066818 Chr.6: 24806788-24806788 on Build GRCh38
rs557513530 Chr.6: 24806731-24806731 on Build GRCh38
rs779358474 Chr.6: 24806617-24806617 on Build GRCh38
rs956988427 Chr.6: 24806645-24806645 on Build GRCh38
rs960550621 Chr.6: 24806808-24806808 on Build GRCh38
rs796869794 Chr.6: 24806514-24806514 on Build GRCh38
rs368088673 Chr.6: 24806425-24806425 on Build GRCh38
rs543575710 Chr.6: 24806479-24806479 on Build GRCh38
rs767133936 Chr.6: 24806428-24806428 on Build GRCh38
rs762906231 Chr.6: 24806349-24806349 on Build GRCh38
rs4074420 Chr.6: 24806524-24806524 on Build GRCh38
rs915320500 Chr.6: 24806411-24806411 on Build GRCh38
rs115680381 Chr.6: 24806485-24806485 on Build GRCh38
rs72544164 Chr.6: 24806510-24806511 on Build GRCh38
rs750061582 Chr.6: 24806462-24806462 on Build GRCh38
rs764496027 Chr.6: 24806444-24806444 on Build GRCh38
rs369466238 Chr.6: 24806592-24806592 on Build GRCh38
rs1011119163 Chr.6: 24806671-24806671 on Build GRCh38
rs145459697 Chr.6: 24806456-24806456 on Build GRCh38
rs373913240 Chr.6: 24806384-24806384 on Build GRCh38
rs968251971 Chr.6: 24806412-24806412 on Build GRCh38
rs34752048 Chr.6: 24806512-24806513 on Build GRCh38
rs779486427 Chr.6: 24806488-24806488 on Build GRCh38
rs753906236 Chr.6: 24806404-24806404 on Build GRCh38
rs955064179 Chr.6: 24806361-24806361 on Build GRCh38
rs969811050 Chr.6: 24806748-24806748 on Build GRCh38
rs186715720 Chr.6: 24806762-24806762 on Build GRCh38
rs571683346 Chr.6: 24806672-24806672 on Build GRCh38
rs759677545 Chr.6: 24806414-24806414 on Build GRCh38
rs9358799 Chr.6: 24806366-24806366 on Build GRCh38
rs529137682 Chr.6: 24806491-24806491 on Build GRCh38
rs1031213007 Chr.6: 24806608-24806608 on Build GRCh38
rs930099262 Chr.6: 24806619-24806619 on Build GRCh38
rs997009136 Chr.6: 24806348-24806348 on Build GRCh38
rs896527683 Chr.6: 24806683-24806683 on Build GRCh38
rs201521593 Chr.6: 24806446-24806446 on Build GRCh38
rs9379690 Chr.6: 24806695-24806695 on Build GRCh38
rs746807950 Chr.6: 24806518-24806518 on Build GRCh38
rs747474841 Chr.6: 24806742-24806742 on Build GRCh38
rs568002107 Chr.6: 24806732-24806732 on Build GRCh38
rs758868767 Chr.6: 24806443-24806443 on Build GRCh38
rs5875006 Chr.6: 24806511-24806512 on Build GRCh38
rs551193791 Chr.6: 24806540-24806541 on Build GRCh38
rs1056548618 Chr.6: 24806657-24806657 on Build GRCh38
rs750363302 Chr.6: 24806387-24806387 on Build GRCh38
rs941239848 Chr.6: 24806542-24806542 on Build GRCh38
rs9379689 Chr.6: 24806549-24806549 on Build GRCh38
rs1038009564 Chr.6: 24806466-24806466 on Build GRCh38
rs896470249 Chr.6: 24806368-24806368 on Build GRCh38
rs765293470 Chr.6: 24806420-24806420 on Build GRCh38
rs759024282 Chr.6: 24806613-24806613 on Build GRCh38
rs1023607612 Chr.6: 24806710-24806710 on Build GRCh38
rs904119487 Chr.6: 24806718-24806718 on Build GRCh38
rs941002030 Chr.6: 24806459-24806459 on Build GRCh38
rs369994154 Chr.6: 24806454-24806454 on Build GRCh38
rs988950296 Chr.6: 24806460-24806460 on Build GRCh38

View Full Product Details