Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 42962949-42963466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNPY3-GNMT
Entrez Gene ID
107080644
Gene Name
CNPY3-GNMT readthrough
Gene Aliases
-
UniGene
Hs.144914
Gene Chromosome Location
Chr.6: 42929130-42963880 on Build GRCh38
Transcripts
NM_001318856.1 NM_001318857.1 NM_001318858.1 NR_134890.1 NR_134891.1 NR_134892.1 NM_001318865.1 NM_018960.5 NR_134899.1

Gene Symbol
GNMT
Entrez Gene ID
27232
Gene Name
glycine N-methyltransferase
Gene Aliases
HEL-S-182mP
UniGene
Hs.144914
Gene Chromosome Location
Chr.6: 42960754-42963880 on Build GRCh38
Transcripts
NM_001318856.1 NM_001318857.1 NM_001318858.1 NR_134890.1 NR_134891.1 NR_134892.1 NM_001318865.1 NM_018960.5 NR_134899.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6513967 c.478C>A p.L160M Chr.6: 42963098-42963098 on Build GRCh38
4687498 c.554G>A p.S185N Chr.6: 42963174-42963174 on Build GRCh38
3874152 c.491C>T p.A164V Chr.6: 42963111-42963111 on Build GRCh38
1621669 c.541G>A p.D181N Chr.6: 42963161-42963161 on Build GRCh38
3874151 c.466C>T p.H156Y Chr.6: 42963086-42963086 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146275920 Chr.6: 42963074-42963074 on Build GRCh38
rs121907889 Chr.6: 42963149-42963149 on Build GRCh38
rs1031947375 Chr.6: 42962989-42962989 on Build GRCh38
rs200015105 Chr.6: 42963057-42963057 on Build GRCh38
rs577108844 Chr.6: 42962976-42962976 on Build GRCh38
rs144716335 Chr.6: 42963123-42963123 on Build GRCh38
rs752057158 Chr.6: 42963142-42963142 on Build GRCh38
rs759792510 Chr.6: 42963399-42963399 on Build GRCh38
rs113625532 Chr.6: 42963281-42963281 on Build GRCh38
rs369431719 Chr.6: 42963298-42963298 on Build GRCh38
rs762369159 Chr.6: 42963144-42963144 on Build GRCh38
rs770519701 Chr.6: 42963128-42963128 on Build GRCh38
rs566244116 Chr.6: 42963153-42963153 on Build GRCh38
rs771883382 Chr.6: 42963121-42963121 on Build GRCh38
rs202053748 Chr.6: 42963079-42963079 on Build GRCh38
rs752763722 Chr.6: 42963404-42963404 on Build GRCh38
rs1000528575 Chr.6: 42963027-42963027 on Build GRCh38
rs142613217 Chr.6: 42963089-42963089 on Build GRCh38
rs774844861 Chr.6: 42963227-42963244 on Build GRCh38
rs748895567 Chr.6: 42963232-42963233 on Build GRCh38
rs144865362 Chr.6: 42963365-42963365 on Build GRCh38
rs755296389 Chr.6: 42963318-42963318 on Build GRCh38
rs563826316 Chr.6: 42963090-42963090 on Build GRCh38
rs772929804 Chr.6: 42963122-42963122 on Build GRCh38
rs756343530 Chr.6: 42963102-42963102 on Build GRCh38
rs919773362 Chr.6: 42963260-42963260 on Build GRCh38
rs768087069 Chr.6: 42963088-42963088 on Build GRCh38
rs776271853 Chr.6: 42963220-42963220 on Build GRCh38
rs772636605 Chr.6: 42963417-42963417 on Build GRCh38
rs759061456 Chr.6: 42963132-42963132 on Build GRCh38
rs769917180 Chr.6: 42963025-42963025 on Build GRCh38
rs779697649 Chr.6: 42963443-42963443 on Build GRCh38
rs138085632 Chr.6: 42963087-42963087 on Build GRCh38
rs758306749 Chr.6: 42963174-42963174 on Build GRCh38
rs769281678 Chr.6: 42963230-42963230 on Build GRCh38
rs149792195 Chr.6: 42963112-42963112 on Build GRCh38
rs147291064 Chr.6: 42963139-42963139 on Build GRCh38
rs371794899 Chr.6: 42963130-42963130 on Build GRCh38
rs780155247 Chr.6: 42963107-42963107 on Build GRCh38
rs147746130 Chr.6: 42963160-42963160 on Build GRCh38
rs1014621237 Chr.6: 42963046-42963046 on Build GRCh38
rs772909903 Chr.6: 42963058-42963058 on Build GRCh38
rs1012093959 Chr.6: 42963039-42963039 on Build GRCh38
rs766824673 Chr.6: 42963386-42963386 on Build GRCh38
rs145997766 Chr.6: 42963108-42963108 on Build GRCh38
rs758305053 Chr.6: 42963346-42963346 on Build GRCh38
rs746753351 Chr.6: 42963352-42963352 on Build GRCh38
rs904285290 Chr.6: 42962981-42962981 on Build GRCh38
rs749218665 Chr.6: 42963029-42963029 on Build GRCh38
rs908521275 Chr.6: 42963243-42963243 on Build GRCh38
rs765577156 Chr.6: 42963402-42963402 on Build GRCh38
rs768654043 Chr.6: 42963111-42963111 on Build GRCh38
rs1042140749 Chr.6: 42962986-42962986 on Build GRCh38
rs755100354 Chr.6: 42963161-42963161 on Build GRCh38
rs961349985 Chr.6: 42963433-42963433 on Build GRCh38
rs762948139 Chr.6: 42963405-42963405 on Build GRCh38
rs201556777 Chr.6: 42963030-42963030 on Build GRCh38
rs772561384 Chr.6: 42963379-42963379 on Build GRCh38
rs200542100 Chr.6: 42963367-42963367 on Build GRCh38
rs775033910 Chr.6: 42963237-42963237 on Build GRCh38
rs767944079 Chr.6: 42963299-42963299 on Build GRCh38
rs777583453 Chr.6: 42963207-42963207 on Build GRCh38
rs750649643 Chr.6: 42963097-42963097 on Build GRCh38
rs764303705 Chr.6: 42963416-42963416 on Build GRCh38
rs778228488 Chr.6: 42963126-42963136 on Build GRCh38
rs770560995 Chr.6: 42963214-42963214 on Build GRCh38
rs998590857 Chr.6: 42962984-42962984 on Build GRCh38
rs756998914 Chr.6: 42963360-42963360 on Build GRCh38
rs754095466 Chr.6: 42963313-42963313 on Build GRCh38
rs201256313 Chr.6: 42963150-42963150 on Build GRCh38
rs1042612730 Chr.6: 42963221-42963221 on Build GRCh38
rs750253852 Chr.6: 42963438-42963438 on Build GRCh38
rs745443121 Chr.6: 42963227-42963238 on Build GRCh38
rs769497018 Chr.6: 42963227-42963232 on Build GRCh38
rs765376257 Chr.6: 42963320-42963320 on Build GRCh38
rs1038661129 Chr.6: 42963255-42963255 on Build GRCh38
rs760950411 Chr.6: 42963307-42963307 on Build GRCh38
rs781476590 Chr.6: 42963096-42963096 on Build GRCh38
rs746799680 Chr.6: 42963211-42963211 on Build GRCh38
rs142634786 Chr.6: 42963348-42963348 on Build GRCh38
rs777175305 Chr.6: 42963390-42963390 on Build GRCh38
rs780937019 Chr.6: 42963434-42963434 on Build GRCh38
rs768682610 Chr.6: 42963254-42963255 on Build GRCh38
rs200543916 Chr.6: 42963165-42963165 on Build GRCh38
rs200406766 Chr.6: 42963056-42963056 on Build GRCh38
rs939966773 Chr.6: 42963249-42963249 on Build GRCh38
rs774477434 Chr.6: 42963349-42963351 on Build GRCh38
rs745587549 Chr.6: 42963225-42963225 on Build GRCh38
rs779187760 Chr.6: 42963163-42963163 on Build GRCh38
rs201222489 Chr.6: 42963345-42963345 on Build GRCh38
rs773579103 Chr.6: 42963300-42963300 on Build GRCh38
rs762354296 Chr.6: 42963244-42963244 on Build GRCh38
rs199634079 Chr.6: 42963308-42963308 on Build GRCh38
rs773586353 Chr.6: 42963384-42963384 on Build GRCh38
rs902687624 Chr.6: 42963257-42963257 on Build GRCh38
rs753327439 Chr.6: 42963080-42963080 on Build GRCh38
rs755716132 Chr.6: 42963442-42963442 on Build GRCh38
rs777583238 Chr.6: 42963350-42963350 on Build GRCh38
rs781024528 Chr.6: 42963361-42963361 on Build GRCh38
rs757268426 Chr.6: 42963419-42963419 on Build GRCh38
rs1022889192 Chr.6: 42963052-42963052 on Build GRCh38
rs774236296 Chr.6: 42963047-42963047 on Build GRCh38

View Full Product Details