Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43187162-43187660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUL9
Entrez Gene ID
23113
Gene Name
cullin 9
Gene Aliases
H7AP1, PARC
UniGene
Hs.485434
Gene Chromosome Location
Chr.6: 43182170-43224587 on Build GRCh38
Transcripts
NM_015089.3 XM_011514422.1 XM_011514423.2 XM_011514424.1 XM_011514425.1 XM_011514426.1 XM_011514427.1 XM_011514428.1 XM_011514430.1 XM_017010589.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4929717 c.1411C>T p.P471S Chr.6: 43187269-43187269 on Build GRCh38
3874233 c.1449C>T p.P483P Chr.6: 43187307-43187307 on Build GRCh38
6056300 c.1440_1441CC>TT p.L481F Chr.6: 43187298-43187299 on Build GRCh38
1697254 c.1444C>T p.Q482* Chr.6: 43187302-43187302 on Build GRCh38
3874234 c.1546C>T p.P516S Chr.6: 43187404-43187404 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs895892981 Chr.6: 43187605-43187605 on Build GRCh38
rs749016351 Chr.6: 43187456-43187456 on Build GRCh38
rs374801171 Chr.6: 43187475-43187475 on Build GRCh38
rs1022964261 Chr.6: 43187626-43187626 on Build GRCh38
rs904455106 Chr.6: 43187548-43187548 on Build GRCh38
rs765556577 Chr.6: 43187311-43187311 on Build GRCh38
rs918991124 Chr.6: 43187434-43187434 on Build GRCh38
rs754608063 Chr.6: 43187465-43187465 on Build GRCh38
rs762923359 Chr.6: 43187329-43187329 on Build GRCh38
rs936022397 Chr.6: 43187528-43187528 on Build GRCh38
rs778443257 Chr.6: 43187466-43187466 on Build GRCh38
rs988354356 Chr.6: 43187199-43187199 on Build GRCh38
rs1046362652 Chr.6: 43187595-43187595 on Build GRCh38
rs535299393 Chr.6: 43187353-43187353 on Build GRCh38
rs554681943 Chr.6: 43187515-43187515 on Build GRCh38
rs908705460 Chr.6: 43187597-43187597 on Build GRCh38
rs755969849 Chr.6: 43187419-43187419 on Build GRCh38
rs180792663 Chr.6: 43187330-43187330 on Build GRCh38
rs369921760 Chr.6: 43187226-43187226 on Build GRCh38
rs961504021 Chr.6: 43187388-43187388 on Build GRCh38
rs374275067 Chr.6: 43187285-43187285 on Build GRCh38
rs542638811 Chr.6: 43187270-43187270 on Build GRCh38
rs748743327 Chr.6: 43187293-43187293 on Build GRCh38
rs564437299 Chr.6: 43187247-43187247 on Build GRCh38
rs767457457 Chr.6: 43187383-43187383 on Build GRCh38
rs745669104 Chr.6: 43187280-43187280 on Build GRCh38
rs752762877 Chr.6: 43187252-43187252 on Build GRCh38
rs560171801 Chr.6: 43187189-43187189 on Build GRCh38
rs144518453 Chr.6: 43187294-43187294 on Build GRCh38
rs998624383 Chr.6: 43187575-43187575 on Build GRCh38
rs199817299 Chr.6: 43187407-43187407 on Build GRCh38
rs1043329627 Chr.6: 43187530-43187530 on Build GRCh38
rs758575726 Chr.6: 43187253-43187253 on Build GRCh38
rs760915690 Chr.6: 43187202-43187202 on Build GRCh38
rs150947009 Chr.6: 43187222-43187222 on Build GRCh38
rs368631463 Chr.6: 43187273-43187273 on Build GRCh38
rs771369812 Chr.6: 43187308-43187308 on Build GRCh38
rs759788784 Chr.6: 43187227-43187227 on Build GRCh38
rs772121136 Chr.6: 43187493-43187493 on Build GRCh38
rs760800925 Chr.6: 43187393-43187393 on Build GRCh38
rs943050664 Chr.6: 43187482-43187482 on Build GRCh38
rs777065100 Chr.6: 43187310-43187310 on Build GRCh38
rs779740471 Chr.6: 43187286-43187286 on Build GRCh38
rs760174596 Chr.6: 43187361-43187361 on Build GRCh38
rs765556937 Chr.6: 43187242-43187242 on Build GRCh38
rs539923389 Chr.6: 43187187-43187187 on Build GRCh38
rs916179942 Chr.6: 43187587-43187587 on Build GRCh38
rs772444496 Chr.6: 43187200-43187200 on Build GRCh38
rs757153646 Chr.6: 43187365-43187365 on Build GRCh38
rs762463281 Chr.6: 43187195-43187195 on Build GRCh38
rs61739541 Chr.6: 43187307-43187307 on Build GRCh38
rs1014326739 Chr.6: 43187617-43187617 on Build GRCh38
rs577684428 Chr.6: 43187184-43187184 on Build GRCh38
rs777544352 Chr.6: 43187256-43187256 on Build GRCh38
rs201152324 Chr.6: 43187309-43187309 on Build GRCh38
rs1031689977 Chr.6: 43187215-43187215 on Build GRCh38
rs148427416 Chr.6: 43187295-43187295 on Build GRCh38
rs940129085 Chr.6: 43187620-43187620 on Build GRCh38
rs568427391 Chr.6: 43187523-43187523 on Build GRCh38
rs933084190 Chr.6: 43187460-43187460 on Build GRCh38
rs1052909135 Chr.6: 43187604-43187604 on Build GRCh38
rs150504259 Chr.6: 43187433-43187433 on Build GRCh38
rs769380090 Chr.6: 43187283-43187283 on Build GRCh38
rs878860628 Chr.6: 43187300-43187300 on Build GRCh38
rs775898360 Chr.6: 43187315-43187315 on Build GRCh38
rs367963137 Chr.6: 43187488-43187488 on Build GRCh38
rs950313384 Chr.6: 43187593-43187593 on Build GRCh38
rs201148900 Chr.6: 43187476-43187476 on Build GRCh38
rs537572417 Chr.6: 43187553-43187553 on Build GRCh38
rs766459450 Chr.6: 43187487-43187488 on Build GRCh38
rs757166762 Chr.6: 43187272-43187272 on Build GRCh38
rs1003964190 Chr.6: 43187230-43187230 on Build GRCh38

View Full Product Details