Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43451860-43452298 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC10
Entrez Gene ID
89845
Gene Name
ATP binding cassette subfamily C member 10
Gene Aliases
EST182763, MRP7, SIMRP7
UniGene
Hs.55879
Gene Chromosome Location
Chr.6: 43427538-43451994 on Build GRCh38
Transcripts
XM_017011449.1 NM_001286655.1 NM_001286656.1 NM_023932.3 NM_206539.2 XM_005249308.4 XM_011514823.2 XM_017011204.1

Gene Symbol
DLK2
Entrez Gene ID
65989
Gene Name
delta like non-canonical Notch ligand 2
Gene Aliases
DLK-2, EGFL9
UniGene
Hs.337251
Gene Chromosome Location
Chr.6: 43450352-43456632 on Build GRCh38
Transcripts
XM_017011449.1 NM_001286655.1 NM_001286656.1 NM_023932.3 NM_206539.2 XM_005249308.4 XM_011514823.2 XM_017011204.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
310596 c.376C>T p.R126C Chr.6: 43451980-43451980 on Build GRCh38
4938681 c.386A>G p.E129G Chr.6: 43451970-43451970 on Build GRCh38
1250199 c.361C>A p.P121T Chr.6: 43451995-43451995 on Build GRCh38
1079589 c.357C>T p.C119C Chr.6: 43451999-43451999 on Build GRCh38
7296909 c.330C>T p.D110D Chr.6: 43452026-43452026 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369156573 Chr.6: 43452007-43452007 on Build GRCh38
rs371287743 Chr.6: 43451888-43451888 on Build GRCh38
rs372062583 Chr.6: 43451984-43451984 on Build GRCh38
rs914313379 Chr.6: 43451885-43451885 on Build GRCh38
rs746387048 Chr.6: 43451940-43451940 on Build GRCh38
rs759269315 Chr.6: 43451987-43451987 on Build GRCh38
rs189224804 Chr.6: 43452127-43452127 on Build GRCh38
rs770952333 Chr.6: 43452059-43452059 on Build GRCh38
rs888511039 Chr.6: 43451967-43451967 on Build GRCh38
rs766988405 Chr.6: 43452104-43452104 on Build GRCh38
rs766294294 Chr.6: 43451902-43451902 on Build GRCh38
rs773342263 Chr.6: 43451979-43451979 on Build GRCh38
rs777996120 Chr.6: 43452132-43452132 on Build GRCh38
rs139581243 Chr.6: 43451887-43451887 on Build GRCh38
rs754417791 Chr.6: 43452230-43452232 on Build GRCh38
rs147808380 Chr.6: 43451908-43451908 on Build GRCh38
rs775409039 Chr.6: 43452078-43452078 on Build GRCh38
rs899569499 Chr.6: 43451925-43451925 on Build GRCh38
rs1014676927 Chr.6: 43451977-43451977 on Build GRCh38
rs754315604 Chr.6: 43452118-43452118 on Build GRCh38
rs748556732 Chr.6: 43452122-43452122 on Build GRCh38
rs749764675 Chr.6: 43452020-43452020 on Build GRCh38
rs768701190 Chr.6: 43452122-43452133 on Build GRCh38
rs998307900 Chr.6: 43452239-43452239 on Build GRCh38
rs770481699 Chr.6: 43451952-43451952 on Build GRCh38
rs776461451 Chr.6: 43452065-43452065 on Build GRCh38
rs949001112 Chr.6: 43452247-43452247 on Build GRCh38
rs201377693 Chr.6: 43452120-43452120 on Build GRCh38
rs985783463 Chr.6: 43452152-43452152 on Build GRCh38
rs141570361 Chr.6: 43452079-43452079 on Build GRCh38
rs561584025 Chr.6: 43452197-43452197 on Build GRCh38
rs780702112 Chr.6: 43451962-43451962 on Build GRCh38
rs747143741 Chr.6: 43452033-43452033 on Build GRCh38
rs375089342 Chr.6: 43452006-43452006 on Build GRCh38
rs749652048 Chr.6: 43451963-43451963 on Build GRCh38
rs755304127 Chr.6: 43452021-43452021 on Build GRCh38
rs368152078 Chr.6: 43451971-43451971 on Build GRCh38
rs748499645 Chr.6: 43452025-43452025 on Build GRCh38
rs753645907 Chr.6: 43451906-43451906 on Build GRCh38
rs948732437 Chr.6: 43451929-43451929 on Build GRCh38
rs530331479 Chr.6: 43452209-43452209 on Build GRCh38
rs2275667 Chr.6: 43451896-43451896 on Build GRCh38
rs147898599 Chr.6: 43452066-43452066 on Build GRCh38
rs992688536 Chr.6: 43452134-43452134 on Build GRCh38
rs780473767 Chr.6: 43452019-43452019 on Build GRCh38
rs763878348 Chr.6: 43452092-43452092 on Build GRCh38
rs751232064 Chr.6: 43452093-43452093 on Build GRCh38
rs954991184 Chr.6: 43452017-43452017 on Build GRCh38
rs779279995 Chr.6: 43452022-43452022 on Build GRCh38
rs749547318 Chr.6: 43452076-43452077 on Build GRCh38
rs776612619 Chr.6: 43451985-43451985 on Build GRCh38
rs751180998 Chr.6: 43452016-43452016 on Build GRCh38
rs541360445 Chr.6: 43452166-43452166 on Build GRCh38
rs924392127 Chr.6: 43452201-43452201 on Build GRCh38
rs938830573 Chr.6: 43452203-43452203 on Build GRCh38
rs763581344 Chr.6: 43451892-43451892 on Build GRCh38
rs772154206 Chr.6: 43451972-43451972 on Build GRCh38
rs777349286 Chr.6: 43451934-43451934 on Build GRCh38
rs776517816 Chr.6: 43452168-43452168 on Build GRCh38
rs144772479 Chr.6: 43452018-43452018 on Build GRCh38
rs576502207 Chr.6: 43451923-43451923 on Build GRCh38
rs373026615 Chr.6: 43452026-43452026 on Build GRCh38
rs917037669 Chr.6: 43452165-43452165 on Build GRCh38
rs375111558 Chr.6: 43451920-43451920 on Build GRCh38
rs769018546 Chr.6: 43451968-43451968 on Build GRCh38
rs916153380 Chr.6: 43452212-43452212 on Build GRCh38
rs111958097 Chr.6: 43452125-43452125 on Build GRCh38
rs763824814 Chr.6: 43452008-43452008 on Build GRCh38
rs756859737 Chr.6: 43452097-43452097 on Build GRCh38
rs371976722 Chr.6: 43451910-43451910 on Build GRCh38
rs140806227 Chr.6: 43452002-43452002 on Build GRCh38
rs1030228843 Chr.6: 43452115-43452115 on Build GRCh38

View Full Product Details