Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 53070895-53071374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO9
Entrez Gene ID
26268
Gene Name
F-box protein 9
Gene Aliases
FBX9, NY-REN-57, VCIA1, dJ341E18.2
UniGene
Hs.216653
Gene Chromosome Location
Chr.6: 53057807-53100873 on Build GRCh38
Transcripts
NM_012347.4 NM_033480.2 NM_033481.3 XM_005248995.4 XM_005248996.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1445192 c.33G>T p.Q11H Chr.6: 53071056-53071056 on Build GRCh38
4831804 c.82G>A p.D28N Chr.6: 53071105-53071105 on Build GRCh38
6251328 c.90+1G>T p.? Chr.6: 53071144-53071144 on Build GRCh38
6346932 c.98G>T p.S33I Chr.6: 53071121-53071121 on Build GRCh38
6251329 c.120+1G>T p.? Chr.6: 53071144-53071144 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771344873 Chr.6: 53071158-53071158 on Build GRCh38
rs760399016 Chr.6: 53071033-53071033 on Build GRCh38
rs373570237 Chr.6: 53071026-53071026 on Build GRCh38
rs375245317 Chr.6: 53071155-53071155 on Build GRCh38
rs1045863167 Chr.6: 53071243-53071243 on Build GRCh38
rs201193210 Chr.6: 53071288-53071288 on Build GRCh38
rs549684821 Chr.6: 53071219-53071219 on Build GRCh38
rs753300894 Chr.6: 53071028-53071028 on Build GRCh38
rs966124045 Chr.6: 53071047-53071047 on Build GRCh38
rs757029491 Chr.6: 53071099-53071101 on Build GRCh38
rs927503105 Chr.6: 53071256-53071256 on Build GRCh38
rs867919519 Chr.6: 53071051-53071051 on Build GRCh38
rs893418586 Chr.6: 53071330-53071330 on Build GRCh38
rs377399172 Chr.6: 53071076-53071076 on Build GRCh38
rs994547416 Chr.6: 53070953-53070953 on Build GRCh38
rs778011310 Chr.6: 53071137-53071137 on Build GRCh38
rs370453561 Chr.6: 53071140-53071140 on Build GRCh38
rs957263050 Chr.6: 53071316-53071316 on Build GRCh38
rs774700673 Chr.6: 53071179-53071179 on Build GRCh38
rs1038516128 Chr.6: 53071193-53071193 on Build GRCh38
rs756205334 Chr.6: 53071315-53071315 on Build GRCh38
rs961450230 Chr.6: 53070937-53070937 on Build GRCh38
rs971703635 Chr.6: 53071008-53071008 on Build GRCh38
rs1024739145 Chr.6: 53071239-53071239 on Build GRCh38
rs751616684 Chr.6: 53071098-53071098 on Build GRCh38
rs762278689 Chr.6: 53071347-53071347 on Build GRCh38
rs868352826 Chr.6: 53071105-53071105 on Build GRCh38
rs989988566 Chr.6: 53071054-53071054 on Build GRCh38
rs985395450 Chr.6: 53071114-53071114 on Build GRCh38
rs766936072 Chr.6: 53071216-53071216 on Build GRCh38
rs760030223 Chr.6: 53071014-53071014 on Build GRCh38
rs918372230 Chr.6: 53070964-53070964 on Build GRCh38
rs994239281 Chr.6: 53071235-53071235 on Build GRCh38
rs754879745 Chr.6: 53071099-53071099 on Build GRCh38
rs759835994 Chr.6: 53071186-53071186 on Build GRCh38
rs940964238 Chr.6: 53070925-53070925 on Build GRCh38
rs1055077886 Chr.6: 53071013-53071013 on Build GRCh38
rs781130396 Chr.6: 53071100-53071100 on Build GRCh38
rs909754677 Chr.6: 53071126-53071126 on Build GRCh38
rs748203633 Chr.6: 53071125-53071125 on Build GRCh38
rs906012761 Chr.6: 53071006-53071006 on Build GRCh38
rs756742490 Chr.6: 53071052-53071052 on Build GRCh38
rs371539794 Chr.6: 53070990-53070990 on Build GRCh38
rs41273864 Chr.6: 53071293-53071293 on Build GRCh38
rs779980105 Chr.6: 53071088-53071088 on Build GRCh38
rs1020240754 Chr.6: 53071036-53071036 on Build GRCh38
rs1001807430 Chr.6: 53071007-53071007 on Build GRCh38
rs529643861 Chr.6: 53071085-53071085 on Build GRCh38
rs533315640 Chr.6: 53071111-53071111 on Build GRCh38
rs149719732 Chr.6: 53071079-53071079 on Build GRCh38
rs543094199 Chr.6: 53071012-53071012 on Build GRCh38
rs566346429 Chr.6: 53071226-53071226 on Build GRCh38
rs1036788427 Chr.6: 53070927-53070927 on Build GRCh38
rs1016935851 Chr.6: 53070924-53070924 on Build GRCh38
rs897253766 Chr.6: 53071205-53071205 on Build GRCh38
rs971387195 Chr.6: 53071257-53071257 on Build GRCh38
rs1054622530 Chr.6: 53071320-53071320 on Build GRCh38
rs912955760 Chr.6: 53071096-53071096 on Build GRCh38
rs763340354 Chr.6: 53070976-53070976 on Build GRCh38
rs769878268 Chr.6: 53071130-53071130 on Build GRCh38
rs990008231 Chr.6: 53071329-53071329 on Build GRCh38

View Full Product Details