Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 55513840-55514345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMGCLL1
Entrez Gene ID
54511
Gene Name
3-hydroxymethyl-3-methylglutaryl-CoA lyase like 1
Gene Aliases
bA418P12.1, er-cHL
UniGene
Hs.147054
Gene Chromosome Location
Chr.6: 55434373-55579214 on Build GRCh38
Transcripts
NM_001042406.1 NM_001287741.1 NM_001287746.1 NM_001287753.1 NM_019036.2 NR_109867.1 NR_109869.1 XM_011514700.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3874874 c.632G>A p.G211E Chr.6: 55514048-55514048 on Build GRCh38
6892055 c.577G>A p.G193R Chr.6: 55514103-55514103 on Build GRCh38
3874875 c.594T>C p.V198V Chr.6: 55514086-55514086 on Build GRCh38
4896090 c.625G>A p.A209T Chr.6: 55514055-55514055 on Build GRCh38
1445250 c.579A>G p.G193G Chr.6: 55514101-55514101 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113328041 Chr.6: 55513885-55513885 on Build GRCh38
rs764634937 Chr.6: 55514097-55514097 on Build GRCh38
rs753138656 Chr.6: 55513993-55513993 on Build GRCh38
rs769587328 Chr.6: 55514145-55514151 on Build GRCh38
rs368385220 Chr.6: 55514129-55514129 on Build GRCh38
rs751051634 Chr.6: 55514196-55514196 on Build GRCh38
rs746718401 Chr.6: 55514034-55514034 on Build GRCh38
rs766132955 Chr.6: 55514065-55514065 on Build GRCh38
rs772315880 Chr.6: 55514145-55514145 on Build GRCh38
rs746593770 Chr.6: 55514118-55514118 on Build GRCh38
rs759419234 Chr.6: 55514083-55514085 on Build GRCh38
rs754637204 Chr.6: 55514219-55514219 on Build GRCh38
rs1010617705 Chr.6: 55514036-55514036 on Build GRCh38
rs756662188 Chr.6: 55514001-55514001 on Build GRCh38
rs1027719619 Chr.6: 55513868-55513868 on Build GRCh38
rs1009139400 Chr.6: 55514274-55514274 on Build GRCh38
rs765637698 Chr.6: 55514207-55514207 on Build GRCh38
rs757603663 Chr.6: 55514011-55514011 on Build GRCh38
rs1052523047 Chr.6: 55513982-55513982 on Build GRCh38
rs773062410 Chr.6: 55513940-55513940 on Build GRCh38
rs973374103 Chr.6: 55513867-55513867 on Build GRCh38
rs756543479 Chr.6: 55514084-55514084 on Build GRCh38
rs762775214 Chr.6: 55514058-55514058 on Build GRCh38
rs116026773 Chr.6: 55514323-55514323 on Build GRCh38
rs748038716 Chr.6: 55514048-55514048 on Build GRCh38
rs79694232 Chr.6: 55513950-55513950 on Build GRCh38
rs999632846 Chr.6: 55513968-55513968 on Build GRCh38
rs770705139 Chr.6: 55514134-55514134 on Build GRCh38
rs754352526 Chr.6: 55514106-55514106 on Build GRCh38
rs925721886 Chr.6: 55514181-55514181 on Build GRCh38
rs900410162 Chr.6: 55514307-55514307 on Build GRCh38
rs982625995 Chr.6: 55514138-55514138 on Build GRCh38
rs774149079 Chr.6: 55514136-55514136 on Build GRCh38
rs973505488 Chr.6: 55514228-55514228 on Build GRCh38
rs759709123 Chr.6: 55514141-55514141 on Build GRCh38
rs983915766 Chr.6: 55513911-55513911 on Build GRCh38
rs948337775 Chr.6: 55513932-55513932 on Build GRCh38
rs890304234 Chr.6: 55514264-55514264 on Build GRCh38
rs759817782 Chr.6: 55514072-55514072 on Build GRCh38
rs769792498 Chr.6: 55514051-55514051 on Build GRCh38
rs934439092 Chr.6: 55514212-55514212 on Build GRCh38
rs767531076 Chr.6: 55514074-55514074 on Build GRCh38
rs749697273 Chr.6: 55514007-55514007 on Build GRCh38
rs780960016 Chr.6: 55514239-55514239 on Build GRCh38
rs760787951 Chr.6: 55514173-55514173 on Build GRCh38
rs772842454 Chr.6: 55514052-55514052 on Build GRCh38
rs764138659 Chr.6: 55514180-55514180 on Build GRCh38
rs370153394 Chr.6: 55513968-55513970 on Build GRCh38
rs762698764 Chr.6: 55514040-55514040 on Build GRCh38
rs749239890 Chr.6: 55514132-55514132 on Build GRCh38
rs754368185 Chr.6: 55514184-55514184 on Build GRCh38
rs779832908 Chr.6: 55514022-55514022 on Build GRCh38
rs773175695 Chr.6: 55514130-55514130 on Build GRCh38
rs747724023 Chr.6: 55514126-55514126 on Build GRCh38
rs961122662 Chr.6: 55513939-55513939 on Build GRCh38
rs780798314 Chr.6: 55514120-55514120 on Build GRCh38
rs951224321 Chr.6: 55514109-55514109 on Build GRCh38
rs757628484 Chr.6: 55514115-55514115 on Build GRCh38
rs928174730 Chr.6: 55513926-55513926 on Build GRCh38
rs752799058 Chr.6: 55514078-55514078 on Build GRCh38
rs762232730 Chr.6: 55514192-55514192 on Build GRCh38
rs765998435 Chr.6: 55514117-55514117 on Build GRCh38
rs774540992 Chr.6: 55514071-55514071 on Build GRCh38
rs374617575 Chr.6: 55514309-55514309 on Build GRCh38
rs1046656802 Chr.6: 55514314-55514314 on Build GRCh38
rs963740838 Chr.6: 55514049-55514049 on Build GRCh38
rs1038967504 Chr.6: 55514282-55514282 on Build GRCh38
rs755632714 Chr.6: 55514243-55514243 on Build GRCh38
rs776284961 Chr.6: 55514047-55514047 on Build GRCh38
rs776463771 Chr.6: 55514067-55514068 on Build GRCh38
rs573572685 Chr.6: 55514242-55514242 on Build GRCh38
rs775733260 Chr.6: 55514163-55514163 on Build GRCh38
rs777953528 Chr.6: 55514004-55514004 on Build GRCh38
rs775195272 Chr.6: 55514241-55514242 on Build GRCh38
rs560113946 Chr.6: 55514081-55514081 on Build GRCh38
rs1038109150 Chr.6: 55513908-55513908 on Build GRCh38
rs777211864 Chr.6: 55514246-55514246 on Build GRCh38
rs750838794 Chr.6: 55514208-55514208 on Build GRCh38
rs948405578 Chr.6: 55514080-55514080 on Build GRCh38

View Full Product Details