Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 73723159-73723558 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD109
Entrez Gene ID
135228
Gene Name
CD109 molecule
Gene Aliases
CPAMD7, p180, r150
UniGene
Hs.399891
Gene Chromosome Location
Chr.6: 73679192-73828317 on Build GRCh38
Transcripts
NM_001159587.2 NM_001159588.2 NM_133493.4 XM_005248659.3 XM_011535473.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1200225 c.271C>T p.P91S Chr.6: 73723274-73723274 on Build GRCh38
1445861 c.265A>G p.T89A Chr.6: 73723268-73723268 on Build GRCh38
1191034 c.255_256insT p.K86fs*1 Chr.6: 73723258-73723259 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs944346272 Chr.6: 73723279-73723279 on Build GRCh38
rs557740716 Chr.6: 73723498-73723498 on Build GRCh38
rs993916889 Chr.6: 73723394-73723394 on Build GRCh38
rs774481360 Chr.6: 73723330-73723330 on Build GRCh38
rs371150975 Chr.6: 73723273-73723273 on Build GRCh38
rs1034319394 Chr.6: 73723432-73723432 on Build GRCh38
rs760280946 Chr.6: 73723240-73723240 on Build GRCh38
rs372909469 Chr.6: 73723406-73723406 on Build GRCh38
rs4706552 Chr.6: 73723489-73723489 on Build GRCh38
rs56093139 Chr.6: 73723200-73723200 on Build GRCh38
rs562881739 Chr.6: 73723348-73723348 on Build GRCh38
rs754554057 Chr.6: 73723303-73723304 on Build GRCh38
rs747212067 Chr.6: 73723295-73723295 on Build GRCh38
rs538274650 Chr.6: 73723522-73723522 on Build GRCh38
rs757712494 Chr.6: 73723265-73723265 on Build GRCh38
rs200266982 Chr.6: 73723229-73723229 on Build GRCh38
rs778260950 Chr.6: 73723305-73723306 on Build GRCh38
rs767257686 Chr.6: 73723207-73723207 on Build GRCh38
rs764473267 Chr.6: 73723244-73723244 on Build GRCh38
rs763024363 Chr.6: 73723402-73723406 on Build GRCh38
rs996103742 Chr.6: 73723343-73723343 on Build GRCh38
rs900380006 Chr.6: 73723322-73723322 on Build GRCh38
rs548327264 Chr.6: 73723388-73723388 on Build GRCh38
rs560321949 Chr.6: 73723413-73723413 on Build GRCh38
rs750484349 Chr.6: 73723274-73723274 on Build GRCh38
rs376196947 Chr.6: 73723211-73723211 on Build GRCh38
rs745341964 Chr.6: 73723331-73723331 on Build GRCh38
rs1055337446 Chr.6: 73723264-73723264 on Build GRCh38
rs1048859622 Chr.6: 73723381-73723381 on Build GRCh38
rs570737609 Chr.6: 73723501-73723501 on Build GRCh38
rs766110938 Chr.6: 73723294-73723294 on Build GRCh38
rs368832044 Chr.6: 73723284-73723284 on Build GRCh38
rs990378364 Chr.6: 73723483-73723483 on Build GRCh38
rs749105922 Chr.6: 73723312-73723312 on Build GRCh38
rs958917557 Chr.6: 73723480-73723480 on Build GRCh38
rs75468728 Chr.6: 73723385-73723385 on Build GRCh38
rs990242286 Chr.6: 73723197-73723197 on Build GRCh38
rs780217749 Chr.6: 73723285-73723285 on Build GRCh38
rs751858604 Chr.6: 73723224-73723225 on Build GRCh38
rs147928290 Chr.6: 73723269-73723269 on Build GRCh38
rs777565353 Chr.6: 73723310-73723310 on Build GRCh38
rs941239287 Chr.6: 73723382-73723382 on Build GRCh38
rs201088800 Chr.6: 73723246-73723246 on Build GRCh38
rs753423775 Chr.6: 73723300-73723303 on Build GRCh38
rs760337012 Chr.6: 73723267-73723268 on Build GRCh38
rs939185357 Chr.6: 73723263-73723263 on Build GRCh38
rs770978690 Chr.6: 73723327-73723327 on Build GRCh38
rs763057949 Chr.6: 73723252-73723252 on Build GRCh38

View Full Product Details