Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 73756416-73756916 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD109
Entrez Gene ID
135228
Gene Name
CD109 molecule
Gene Aliases
CPAMD7, p180, r150
UniGene
Hs.399891
Gene Chromosome Location
Chr.6: 73679192-73828317 on Build GRCh38
Transcripts
NM_001159587.2 NM_001159588.2 NM_133493.4 XM_005248659.3 XM_011535473.1 XM_017010283.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4586917 c.671A>G p.Y224C Chr.6: 73756680-73756680 on Build GRCh38
7121636 c.645C>T p.Y215Y Chr.6: 73756654-73756654 on Build GRCh38
1487920 c.673G>A p.V225I Chr.6: 73756682-73756682 on Build GRCh38
6623389 c.639G>T p.Q213H Chr.6: 73756648-73756648 on Build GRCh38
743158 c.649C>G p.Q217E Chr.6: 73756658-73756658 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199818913 Chr.6: 73756600-73756600 on Build GRCh38
rs138812608 Chr.6: 73756866-73756867 on Build GRCh38
rs547903162 Chr.6: 73756627-73756627 on Build GRCh38
rs757114505 Chr.6: 73756664-73756670 on Build GRCh38
rs372509668 Chr.6: 73756719-73756719 on Build GRCh38
rs559203174 Chr.6: 73756826-73756826 on Build GRCh38
rs111593985 Chr.6: 73756756-73756756 on Build GRCh38
rs763116036 Chr.6: 73756658-73756658 on Build GRCh38
rs373400039 Chr.6: 73756582-73756582 on Build GRCh38
rs781516504 Chr.6: 73756683-73756684 on Build GRCh38
rs6453696 Chr.6: 73756654-73756654 on Build GRCh38
rs139297534 Chr.6: 73756872-73756873 on Build GRCh38
rs189808550 Chr.6: 73756642-73756642 on Build GRCh38
rs750048495 Chr.6: 73756705-73756705 on Build GRCh38
rs948425834 Chr.6: 73756888-73756888 on Build GRCh38
rs777715032 Chr.6: 73756618-73756619 on Build GRCh38
rs117457886 Chr.6: 73756890-73756890 on Build GRCh38
rs948372653 Chr.6: 73756613-73756613 on Build GRCh38
rs938619228 Chr.6: 73756491-73756491 on Build GRCh38
rs780593778 Chr.6: 73756625-73756625 on Build GRCh38
rs376620938 Chr.6: 73756876-73756877 on Build GRCh38
rs534355335 Chr.6: 73756689-73756689 on Build GRCh38
rs991119969 Chr.6: 73756803-73756803 on Build GRCh38
rs375658036 Chr.6: 73756639-73756639 on Build GRCh38
rs766789009 Chr.6: 73756710-73756710 on Build GRCh38
rs546960946 Chr.6: 73756455-73756455 on Build GRCh38
rs181408445 Chr.6: 73756663-73756663 on Build GRCh38
rs978558068 Chr.6: 73756795-73756795 on Build GRCh38
rs534964255 Chr.6: 73756458-73756458 on Build GRCh38
rs76076115 Chr.6: 73756576-73756576 on Build GRCh38
rs758833392 Chr.6: 73756604-73756604 on Build GRCh38
rs557144233 Chr.6: 73756852-73756852 on Build GRCh38
rs764087650 Chr.6: 73756685-73756685 on Build GRCh38
rs769893136 Chr.6: 73756640-73756640 on Build GRCh38
rs947241886 Chr.6: 73756444-73756444 on Build GRCh38
rs966821676 Chr.6: 73756776-73756776 on Build GRCh38
rs538914684 Chr.6: 73756815-73756815 on Build GRCh38
rs901048035 Chr.6: 73756542-73756542 on Build GRCh38
rs559419270 Chr.6: 73756539-73756539 on Build GRCh38
rs141855592 Chr.6: 73756660-73756660 on Build GRCh38
rs938506417 Chr.6: 73756825-73756825 on Build GRCh38
rs752703708 Chr.6: 73756610-73756610 on Build GRCh38
rs934347966 Chr.6: 73756439-73756439 on Build GRCh38
rs915827570 Chr.6: 73756837-73756837 on Build GRCh38
rs761439206 Chr.6: 73756692-73756692 on Build GRCh38
rs147241519 Chr.6: 73756675-73756675 on Build GRCh38
rs552914327 Chr.6: 73756707-73756707 on Build GRCh38
rs992729566 Chr.6: 73756833-73756833 on Build GRCh38
rs924461751 Chr.6: 73756810-73756810 on Build GRCh38
rs376755460 Chr.6: 73756696-73756696 on Build GRCh38
rs778688498 Chr.6: 73756595-73756595 on Build GRCh38
rs780861262 Chr.6: 73756781-73756781 on Build GRCh38
rs369291804 Chr.6: 73756632-73756632 on Build GRCh38
rs370813601 Chr.6: 73756680-73756680 on Build GRCh38
rs76369582 Chr.6: 73756767-73756767 on Build GRCh38
rs989487678 Chr.6: 73756724-73756724 on Build GRCh38
rs148527025 Chr.6: 73756657-73756657 on Build GRCh38
rs924408104 Chr.6: 73756463-73756463 on Build GRCh38
rs746057493 Chr.6: 73756718-73756718 on Build GRCh38
rs890597267 Chr.6: 73756677-73756677 on Build GRCh38

View Full Product Details