Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 79915406-79915910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELOVL4
Entrez Gene ID
6785
Gene Name
ELOVL fatty acid elongase 4
Gene Aliases
ADMD, CT118, ISQMR, SCA34, STGD2, STGD3
UniGene
Hs.101915
Gene Chromosome Location
Chr.6: 79914812-79947598 on Build GRCh38
Transcripts
NM_022726.3

SNP Detail

SNP ID Chromosome Location
rs114429396 Chr.6: 79915558-79915558 on Build GRCh38
rs188242906 Chr.6: 79915533-79915533 on Build GRCh38
rs878876427 Chr.6: 79915782-79915782 on Build GRCh38
rs45594338 Chr.6: 79915542-79915542 on Build GRCh38
rs931101042 Chr.6: 79915748-79915748 on Build GRCh38
rs1037762210 Chr.6: 79915703-79915703 on Build GRCh38
rs759008361 Chr.6: 79915723-79915723 on Build GRCh38
rs559809180 Chr.6: 79915872-79915872 on Build GRCh38
rs948732991 Chr.6: 79915498-79915498 on Build GRCh38
rs889463099 Chr.6: 79915492-79915492 on Build GRCh38
rs1044862575 Chr.6: 79915776-79915776 on Build GRCh38
rs563759730 Chr.6: 79915588-79915588 on Build GRCh38
rs1022225350 Chr.6: 79915543-79915543 on Build GRCh38
rs1044442997 Chr.6: 79915511-79915512 on Build GRCh38
rs898315711 Chr.6: 79915431-79915431 on Build GRCh38
rs1043267906 Chr.6: 79915863-79915863 on Build GRCh38
rs530070127 Chr.6: 79915646-79915646 on Build GRCh38
rs975992005 Chr.6: 79915667-79915667 on Build GRCh38
rs1019275812 Chr.6: 79915531-79915531 on Build GRCh38
rs929120193 Chr.6: 79915429-79915429 on Build GRCh38
rs889748338 Chr.6: 79915780-79915780 on Build GRCh38
rs578261043 Chr.6: 79915570-79915570 on Build GRCh38
rs770421537 Chr.6: 79915668-79915668 on Build GRCh38
rs548709796 Chr.6: 79915773-79915773 on Build GRCh38
rs140197858 Chr.6: 79915813-79915814 on Build GRCh38
rs775385606 Chr.6: 79915867-79915867 on Build GRCh38
rs909266474 Chr.6: 79915497-79915497 on Build GRCh38
rs773873948 Chr.6: 79915688-79915688 on Build GRCh38
rs967782167 Chr.6: 79915576-79915577 on Build GRCh38
rs1008131780 Chr.6: 79915789-79915789 on Build GRCh38
rs907195814 Chr.6: 79915521-79915521 on Build GRCh38
rs909038344 Chr.6: 79915772-79915772 on Build GRCh38
rs1028205304 Chr.6: 79915451-79915451 on Build GRCh38
rs886061799 Chr.6: 79915703-79915703 on Build GRCh38
rs1053378333 Chr.6: 79915794-79915794 on Build GRCh38
rs1012160462 Chr.6: 79915837-79915837 on Build GRCh38
rs950562787 Chr.6: 79915680-79915680 on Build GRCh38
rs753760882 Chr.6: 79915821-79915821 on Build GRCh38
rs1019729337 Chr.6: 79915806-79915806 on Build GRCh38
rs185625060 Chr.6: 79915581-79915581 on Build GRCh38
rs569179100 Chr.6: 79915560-79915560 on Build GRCh38
rs574200602 Chr.6: 79915679-79915679 on Build GRCh38
rs995436000 Chr.6: 79915440-79915440 on Build GRCh38
rs542722575 Chr.6: 79915805-79915805 on Build GRCh38
rs1007336363 Chr.6: 79915869-79915869 on Build GRCh38
rs535224309 Chr.6: 79915484-79915484 on Build GRCh38
rs1007932330 Chr.6: 79915514-79915514 on Build GRCh38
rs767482354 Chr.6: 79915825-79915825 on Build GRCh38

View Full Product Details