Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 87407882-87408383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP206
Entrez Gene ID
154313
Gene Name
cilia and flagella associated protein 206
Gene Aliases
C6orf165, dJ382I10.1
UniGene
Hs.82921
Gene Chromosome Location
Chr.6: 87407972-87464473 on Build GRCh38
Transcripts
NM_001031743.2

SNP Detail

SNP ID Chromosome Location
rs900372162 Chr.6: 87408164-87408181 on Build GRCh38
rs928211191 Chr.6: 87408304-87408304 on Build GRCh38
rs1040686982 Chr.6: 87408200-87408200 on Build GRCh38
rs768082844 Chr.6: 87408017-87408017 on Build GRCh38
rs112145549 Chr.6: 87408238-87408238 on Build GRCh38
rs538728543 Chr.6: 87408005-87408005 on Build GRCh38
rs1003334351 Chr.6: 87408003-87408003 on Build GRCh38
rs953793120 Chr.6: 87408240-87408240 on Build GRCh38
rs976430386 Chr.6: 87408236-87408236 on Build GRCh38
rs1006462600 Chr.6: 87407945-87407945 on Build GRCh38
rs888016086 Chr.6: 87407914-87407914 on Build GRCh38
rs571691234 Chr.6: 87407982-87407982 on Build GRCh38
rs904365008 Chr.6: 87408199-87408199 on Build GRCh38
rs1055273915 Chr.6: 87407937-87407937 on Build GRCh38
rs942245383 Chr.6: 87408166-87408166 on Build GRCh38
rs576896255 Chr.6: 87408146-87408146 on Build GRCh38
rs543988781 Chr.6: 87408123-87408123 on Build GRCh38
rs989452347 Chr.6: 87408096-87408096 on Build GRCh38
rs959629202 Chr.6: 87408330-87408330 on Build GRCh38
rs972700450 Chr.6: 87408301-87408301 on Build GRCh38
rs756631037 Chr.6: 87408029-87408029 on Build GRCh38
rs898883082 Chr.6: 87407938-87407938 on Build GRCh38
rs994510125 Chr.6: 87407964-87407964 on Build GRCh38
rs1031719809 Chr.6: 87408082-87408082 on Build GRCh38
rs930353080 Chr.6: 87408152-87408152 on Build GRCh38
rs932430620 Chr.6: 87408129-87408129 on Build GRCh38
rs1021504545 Chr.6: 87408057-87408057 on Build GRCh38
rs976885503 Chr.6: 87408102-87408102 on Build GRCh38
rs1020949100 Chr.6: 87408272-87408272 on Build GRCh38
rs190188870 Chr.6: 87408170-87408170 on Build GRCh38
rs1057449367 Chr.6: 87408172-87408172 on Build GRCh38
rs1013047629 Chr.6: 87408189-87408189 on Build GRCh38
rs554840206 Chr.6: 87408061-87408061 on Build GRCh38
rs115922456 Chr.6: 87408021-87408021 on Build GRCh38
rs909349195 Chr.6: 87408267-87408267 on Build GRCh38
rs894553166 Chr.6: 87408187-87408187 on Build GRCh38
rs11964198 Chr.6: 87408262-87408262 on Build GRCh38
rs764488935 Chr.6: 87408030-87408030 on Build GRCh38
rs11964212 Chr.6: 87408363-87408363 on Build GRCh38
rs998744304 Chr.6: 87408202-87408202 on Build GRCh38
rs1021431005 Chr.6: 87408196-87408196 on Build GRCh38
rs559600212 Chr.6: 87408229-87408229 on Build GRCh38
rs369736660 Chr.6: 87408288-87408288 on Build GRCh38
rs938183652 Chr.6: 87408145-87408145 on Build GRCh38
rs986254771 Chr.6: 87408133-87408133 on Build GRCh38
rs577796985 Chr.6: 87408244-87408244 on Build GRCh38
rs200000217 Chr.6: 87408060-87408060 on Build GRCh38
rs938967804 Chr.6: 87408338-87408338 on Build GRCh38
rs938712756 Chr.6: 87408171-87408171 on Build GRCh38
rs1031607591 Chr.6: 87408212-87408212 on Build GRCh38
rs542183132 Chr.6: 87408239-87408239 on Build GRCh38
rs1010925993 Chr.6: 87408268-87408268 on Build GRCh38
rs575672749 Chr.6: 87408100-87408100 on Build GRCh38
rs11966859 Chr.6: 87408346-87408346 on Build GRCh38
rs937812183 Chr.6: 87407908-87407908 on Build GRCh38
rs7759848 Chr.6: 87408025-87408025 on Build GRCh38
rs919506051 Chr.6: 87408318-87408318 on Build GRCh38
rs543392173 Chr.6: 87408109-87408109 on Build GRCh38
rs1036578921 Chr.6: 87408167-87408167 on Build GRCh38
rs1036316110 Chr.6: 87407963-87407963 on Build GRCh38
rs757497962 Chr.6: 87408362-87408362 on Build GRCh38

View Full Product Details