Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 122416121-122416654 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSF2
Entrez Gene ID
3298
Gene Name
heat shock transcription factor 2
Gene Aliases
HSF 2, HSTF 2
UniGene
Hs.158195
Gene Chromosome Location
Chr.6: 122399551-122433119 on Build GRCh38
Transcripts
NM_001135564.1 NM_001243094.1 NM_004506.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5442946 c.506C>A p.A169E Chr.6: 122416271-122416271 on Build GRCh38
739390 c.468C>G p.S156S Chr.6: 122416233-122416233 on Build GRCh38
6682439 c.527_528insA p.I178fs*12 Chr.6: 122416292-122416293 on Build GRCh38
3171266 c.513G>A p.Q171Q Chr.6: 122416278-122416278 on Build GRCh38
4927182 c.531+2T>C p.? Chr.6: 122416298-122416298 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753417407 Chr.6: 122416259-122416259 on Build GRCh38
rs753391592 Chr.6: 122416181-122416181 on Build GRCh38
rs763820147 Chr.6: 122416248-122416248 on Build GRCh38
rs945369516 Chr.6: 122416244-122416244 on Build GRCh38
rs978049103 Chr.6: 122416554-122416554 on Build GRCh38
rs952468303 Chr.6: 122416448-122416448 on Build GRCh38
rs778537452 Chr.6: 122416292-122416292 on Build GRCh38
rs759180444 Chr.6: 122416478-122416478 on Build GRCh38
rs941551984 Chr.6: 122416511-122416511 on Build GRCh38
rs1024623212 Chr.6: 122416156-122416156 on Build GRCh38
rs1033223713 Chr.6: 122416367-122416367 on Build GRCh38
rs750436191 Chr.6: 122416313-122416313 on Build GRCh38
rs998477734 Chr.6: 122416544-122416544 on Build GRCh38
rs1055704541 Chr.6: 122416575-122416580 on Build GRCh38
rs372841188 Chr.6: 122416224-122416224 on Build GRCh38
rs534560243 Chr.6: 122416159-122416159 on Build GRCh38
rs773244630 Chr.6: 122416212-122416212 on Build GRCh38
rs957228548 Chr.6: 122416373-122416373 on Build GRCh38
rs770090619 Chr.6: 122416201-122416201 on Build GRCh38
rs9375134 Chr.6: 122416514-122416514 on Build GRCh38
rs753091494 Chr.6: 122416227-122416227 on Build GRCh38
rs961072009 Chr.6: 122416422-122416422 on Build GRCh38
rs897393079 Chr.6: 122416517-122416517 on Build GRCh38
rs749437321 Chr.6: 122416329-122416329 on Build GRCh38
rs1027180719 Chr.6: 122416435-122416435 on Build GRCh38
rs903916820 Chr.6: 122416258-122416258 on Build GRCh38
rs770909741 Chr.6: 122416216-122416216 on Build GRCh38
rs756860985 Chr.6: 122416291-122416291 on Build GRCh38
rs779973951 Chr.6: 122416192-122416192 on Build GRCh38
rs750455064 Chr.6: 122416204-122416207 on Build GRCh38
rs756663561 Chr.6: 122416182-122416182 on Build GRCh38
rs1006179167 Chr.6: 122416211-122416211 on Build GRCh38
rs1042488220 Chr.6: 122416247-122416247 on Build GRCh38
rs1013806199 Chr.6: 122416319-122416319 on Build GRCh38
rs746736313 Chr.6: 122416197-122416197 on Build GRCh38
rs754443723 Chr.6: 122416426-122416426 on Build GRCh38
rs774876294 Chr.6: 122416217-122416217 on Build GRCh38
rs778194879 Chr.6: 122416308-122416309 on Build GRCh38
rs779054570 Chr.6: 122416183-122416183 on Build GRCh38
rs993816166 Chr.6: 122416433-122416433 on Build GRCh38
rs757603837 Chr.6: 122416324-122416325 on Build GRCh38
rs754049382 Chr.6: 122416181-122416181 on Build GRCh38
rs1055270153 Chr.6: 122416288-122416288 on Build GRCh38
rs1006677866 Chr.6: 122416324-122416324 on Build GRCh38
rs945400859 Chr.6: 122416533-122416533 on Build GRCh38
rs971313421 Chr.6: 122416421-122416421 on Build GRCh38
rs923237886 Chr.6: 122416242-122416242 on Build GRCh38
rs1013270317 Chr.6: 122416149-122416149 on Build GRCh38
rs888630939 Chr.6: 122416178-122416178 on Build GRCh38
rs895351244 Chr.6: 122416603-122416603 on Build GRCh38
rs114361685 Chr.6: 122416604-122416604 on Build GRCh38
rs961124732 Chr.6: 122416157-122416157 on Build GRCh38
rs769364967 Chr.6: 122416494-122416494 on Build GRCh38
rs1037304765 Chr.6: 122416233-122416233 on Build GRCh38
rs74866696 Chr.6: 122416490-122416490 on Build GRCh38
rs149946798 Chr.6: 122416412-122416412 on Build GRCh38
rs1050741724 Chr.6: 122416563-122416563 on Build GRCh38
rs919673422 Chr.6: 122416161-122416161 on Build GRCh38
rs774433319 Chr.6: 122416333-122416333 on Build GRCh38
rs368820590 Chr.6: 122416213-122416213 on Build GRCh38
rs779596668 Chr.6: 122416327-122416329 on Build GRCh38
rs772572349 Chr.6: 122416336-122416336 on Build GRCh38
rs745797459 Chr.6: 122416187-122416187 on Build GRCh38
rs2243361 Chr.6: 122416182-122416182 on Build GRCh38
rs760010056 Chr.6: 122416218-122416218 on Build GRCh38
rs150012876 Chr.6: 122416182-122416182 on Build GRCh38
rs769632276 Chr.6: 122416520-122416520 on Build GRCh38
rs775140972 Chr.6: 122416552-122416552 on Build GRCh38
rs139201701 Chr.6: 122416315-122416315 on Build GRCh38
rs45461293 Chr.6: 122416171-122416171 on Build GRCh38
rs759568319 Chr.6: 122416335-122416335 on Build GRCh38
rs758897747 Chr.6: 122416307-122416307 on Build GRCh38
rs997838797 Chr.6: 122416312-122416312 on Build GRCh38
rs748946612 Chr.6: 122416331-122416331 on Build GRCh38
rs896785800 Chr.6: 122416272-122416272 on Build GRCh38
rs768281330 Chr.6: 122416322-122416322 on Build GRCh38
rs775801915 Chr.6: 122416338-122416338 on Build GRCh38
rs771044520 Chr.6: 122416332-122416332 on Build GRCh38
rs1001468193 Chr.6: 122416275-122416275 on Build GRCh38
rs776126922 Chr.6: 122416200-122416200 on Build GRCh38
rs772033360 Chr.6: 122416191-122416191 on Build GRCh38
rs201316705 Chr.6: 122416236-122416236 on Build GRCh38
rs200754759 Chr.6: 122416314-122416314 on Build GRCh38
rs374128477 Chr.6: 122416449-122416449 on Build GRCh38
rs910018830 Chr.6: 122416510-122416510 on Build GRCh38
rs925372210 Chr.6: 122416557-122416557 on Build GRCh38
rs765272626 Chr.6: 122416181-122416182 on Build GRCh38
rs747339246 Chr.6: 122416312-122416312 on Build GRCh38
rs989108880 Chr.6: 122416393-122416393 on Build GRCh38
rs185193584 Chr.6: 122416318-122416318 on Build GRCh38
rs1007011043 Chr.6: 122416620-122416620 on Build GRCh38

View Full Product Details