Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 139166973-139167482 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECA
Entrez Gene ID
51696
Gene Name
hdc homolog, cell cycle regulator
Gene Aliases
HDC, HDCL, HHDC, dJ225E12.1
UniGene
Hs.197644
Gene Chromosome Location
Chr.6: 139135112-139180809 on Build GRCh38
Transcripts
NM_016217.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5908627 c.1265C>T p.P422L Chr.6: 139167277-139167277 on Build GRCh38
3781986 c.999G>T p.R333S Chr.6: 139167011-139167011 on Build GRCh38
7403611 c.1097delT p.V366fs*31 Chr.6: 139167109-139167109 on Build GRCh38
3725768 c.1110C>G p.D370E Chr.6: 139167122-139167122 on Build GRCh38
4689890 c.1268C>T p.S423L Chr.6: 139167280-139167280 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367751153 Chr.6: 139167224-139167224 on Build GRCh38
rs147766051 Chr.6: 139167012-139167012 on Build GRCh38
rs1042616058 Chr.6: 139167277-139167277 on Build GRCh38
rs760777393 Chr.6: 139167007-139167007 on Build GRCh38
rs950626351 Chr.6: 139167194-139167194 on Build GRCh38
rs747284214 Chr.6: 139167173-139167173 on Build GRCh38
rs747226950 Chr.6: 139167063-139167063 on Build GRCh38
rs990452240 Chr.6: 139167343-139167343 on Build GRCh38
rs150947459 Chr.6: 139167032-139167032 on Build GRCh38
rs983283988 Chr.6: 139167197-139167197 on Build GRCh38
rs749296857 Chr.6: 139167317-139167317 on Build GRCh38
rs945602715 Chr.6: 139167249-139167249 on Build GRCh38
rs781452221 Chr.6: 139167185-139167185 on Build GRCh38
rs367927843 Chr.6: 139167062-139167062 on Build GRCh38
rs994543335 Chr.6: 139167389-139167389 on Build GRCh38
rs374585437 Chr.6: 139167296-139167296 on Build GRCh38
rs749178753 Chr.6: 139167217-139167217 on Build GRCh38
rs751085471 Chr.6: 139167026-139167026 on Build GRCh38
rs766148085 Chr.6: 139167171-139167171 on Build GRCh38
rs771149308 Chr.6: 139167374-139167375 on Build GRCh38
rs201702729 Chr.6: 139167248-139167248 on Build GRCh38
rs756285622 Chr.6: 139167302-139167302 on Build GRCh38
rs34059634 Chr.6: 139167236-139167236 on Build GRCh38
rs774114464 Chr.6: 139167220-139167220 on Build GRCh38
rs772308570 Chr.6: 139166995-139166995 on Build GRCh38
rs774057363 Chr.6: 139167107-139167107 on Build GRCh38
rs1038758816 Chr.6: 139167337-139167337 on Build GRCh38
rs750992864 Chr.6: 139167237-139167237 on Build GRCh38
rs751931133 Chr.6: 139167278-139167278 on Build GRCh38
rs887309277 Chr.6: 139167445-139167445 on Build GRCh38
rs373143043 Chr.6: 139167376-139167376 on Build GRCh38
rs541841684 Chr.6: 139167198-139167198 on Build GRCh38
rs551402071 Chr.6: 139167242-139167242 on Build GRCh38
rs777701910 Chr.6: 139167306-139167306 on Build GRCh38
rs775732645 Chr.6: 139167001-139167001 on Build GRCh38
rs1027728271 Chr.6: 139167435-139167435 on Build GRCh38
rs773136893 Chr.6: 139167215-139167215 on Build GRCh38
rs138490244 Chr.6: 139167321-139167321 on Build GRCh38
rs868064784 Chr.6: 139167309-139167309 on Build GRCh38
rs757761408 Chr.6: 139167045-139167045 on Build GRCh38
rs762752981 Chr.6: 139167094-139167094 on Build GRCh38
rs540500580 Chr.6: 139167100-139167100 on Build GRCh38
rs866245653 Chr.6: 139167030-139167030 on Build GRCh38
rs765715627 Chr.6: 139167122-139167122 on Build GRCh38
rs778714047 Chr.6: 139167341-139167341 on Build GRCh38
rs542204936 Chr.6: 139167431-139167431 on Build GRCh38
rs200417003 Chr.6: 139167174-139167174 on Build GRCh38
rs373649622 Chr.6: 139167165-139167165 on Build GRCh38
rs752224117 Chr.6: 139167115-139167115 on Build GRCh38
rs140780413 Chr.6: 139167048-139167048 on Build GRCh38
rs771714756 Chr.6: 139167357-139167357 on Build GRCh38
rs138982846 Chr.6: 139167188-139167188 on Build GRCh38
rs142228127 Chr.6: 139167223-139167223 on Build GRCh38
rs774887565 Chr.6: 139167227-139167227 on Build GRCh38
rs776829499 Chr.6: 139167085-139167085 on Build GRCh38
rs768911290 Chr.6: 139167078-139167078 on Build GRCh38
rs532956161 Chr.6: 139167158-139167158 on Build GRCh38
rs575237379 Chr.6: 139167013-139167013 on Build GRCh38
rs540390271 Chr.6: 139167439-139167440 on Build GRCh38
rs192382133 Chr.6: 139167027-139167027 on Build GRCh38
rs145909139 Chr.6: 139167230-139167230 on Build GRCh38
rs917078350 Chr.6: 139167209-139167209 on Build GRCh38
rs748075019 Chr.6: 139167086-139167086 on Build GRCh38
rs760404644 Chr.6: 139167356-139167359 on Build GRCh38
rs145108878 Chr.6: 139167170-139167170 on Build GRCh38
rs772705673 Chr.6: 139167351-139167354 on Build GRCh38
rs375410347 Chr.6: 139167281-139167281 on Build GRCh38
rs200541769 Chr.6: 139167014-139167014 on Build GRCh38
rs754422841 Chr.6: 139167028-139167028 on Build GRCh38
rs562355741 Chr.6: 139167172-139167172 on Build GRCh38
rs769729277 Chr.6: 139167088-139167088 on Build GRCh38
rs759180001 Chr.6: 139167108-139167108 on Build GRCh38
rs754400708 Chr.6: 139167033-139167033 on Build GRCh38
rs745616016 Chr.6: 139167344-139167344 on Build GRCh38
rs1004902380 Chr.6: 139167114-139167114 on Build GRCh38
rs771658394 Chr.6: 139167009-139167010 on Build GRCh38
rs950676209 Chr.6: 139167457-139167457 on Build GRCh38
rs900233574 Chr.6: 139167352-139167352 on Build GRCh38
rs370753473 Chr.6: 139167137-139167137 on Build GRCh38
rs746205849 Chr.6: 139167059-139167059 on Build GRCh38
rs770770683 Chr.6: 139167219-139167219 on Build GRCh38
rs757738486 Chr.6: 139167117-139167117 on Build GRCh38
rs189354892 Chr.6: 139167011-139167011 on Build GRCh38
rs35480523 Chr.6: 139167310-139167311 on Build GRCh38
rs937669137 Chr.6: 139167041-139167041 on Build GRCh38
rs767067217 Chr.6: 139167111-139167111 on Build GRCh38
rs760161738 Chr.6: 139167229-139167229 on Build GRCh38
rs148816859 Chr.6: 139167017-139167017 on Build GRCh38
rs927899185 Chr.6: 139167270-139167270 on Build GRCh38
rs1047462161 Chr.6: 139167433-139167433 on Build GRCh38
rs34476011 Chr.6: 139167023-139167023 on Build GRCh38

View Full Product Details