Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 144008311-144008771 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HYMAI
Entrez Gene ID
57061
Gene Name
hydatidiform mole associated and imprinted (non-protein coding)
Gene Aliases
NCRNA00020
UniGene
Hs.657760
Gene Chromosome Location
Chr.6: 144004916-144008730 on Build GRCh38
Transcripts
NR_002768.2 NM_001080951.2 NM_001080952.2 NM_001080953.2 NM_001080954.2 NM_001080955.2 NM_001080956.2 NM_001289037.1 NM_001289038.1 NM_001289039.1 NM_001289040.1 NM_001289041.1 NM_001289042.1 NM_001289043.1 NM_001289044.1 NM_001289045.1 NM_001289046.1 NM_001289047.1 NM_001289048.1 NM_001289049.1 NM_001317159.1 NM_001317160.1 NM_001317161.1 NM_001317162.1 NM_006718.4

Gene Symbol
PLAGL1
Entrez Gene ID
5325
Gene Name
PLAG1 like zinc finger 1
Gene Aliases
LOT1, ZAC, ZAC1
UniGene
Hs.743225
Gene Chromosome Location
Chr.6: 143940300-144064599 on Build GRCh38
Transcripts
NR_002768.2 NM_001080951.2 NM_001080952.2 NM_001080953.2 NM_001080954.2 NM_001080955.2 NM_001080956.2 NM_001289037.1 NM_001289038.1 NM_001289039.1 NM_001289040.1 NM_001289041.1 NM_001289042.1 NM_001289043.1 NM_001289044.1 NM_001289045.1 NM_001289046.1 NM_001289047.1 NM_001289048.1 NM_001289049.1 NM_001317159.1 NM_001317160.1 NM_001317161.1 NM_001317162.1 NM_006718.4

SNP Detail

SNP ID Chromosome Location
rs1036426810 Chr.6: 144008664-144008664 on Build GRCh38
rs895302452 Chr.6: 144008383-144008383 on Build GRCh38
rs1017538089 Chr.6: 144008483-144008483 on Build GRCh38
rs1004963428 Chr.6: 144008525-144008525 on Build GRCh38
rs967307734 Chr.6: 144008537-144008537 on Build GRCh38
rs1002414276 Chr.6: 144008629-144008629 on Build GRCh38
rs866114612 Chr.6: 144008449-144008449 on Build GRCh38
rs772600408 Chr.6: 144008484-144008484 on Build GRCh38
rs1042043503 Chr.6: 144008348-144008348 on Build GRCh38
rs769073336 Chr.6: 144008737-144008737 on Build GRCh38
rs1013990945 Chr.6: 144008646-144008646 on Build GRCh38
rs761344553 Chr.6: 144008702-144008702 on Build GRCh38
rs77574073 Chr.6: 144008535-144008535 on Build GRCh38
rs905432545 Chr.6: 144008620-144008620 on Build GRCh38
rs1025039055 Chr.6: 144008391-144008391 on Build GRCh38
rs186166234 Chr.6: 144008732-144008732 on Build GRCh38
rs1035557358 Chr.6: 144008634-144008634 on Build GRCh38
rs1026875994 Chr.6: 144008546-144008546 on Build GRCh38
rs963743420 Chr.6: 144008427-144008427 on Build GRCh38
rs941397701 Chr.6: 144008743-144008743 on Build GRCh38
rs948303387 Chr.6: 144008619-144008619 on Build GRCh38
rs899429146 Chr.6: 144008387-144008418 on Build GRCh38
rs954105133 Chr.6: 144008548-144008548 on Build GRCh38
rs568512232 Chr.6: 144008425-144008425 on Build GRCh38
rs1039941240 Chr.6: 144008559-144008559 on Build GRCh38
rs895405252 Chr.6: 144008512-144008512 on Build GRCh38
rs922715024 Chr.6: 144008568-144008568 on Build GRCh38
rs942583257 Chr.6: 144008556-144008556 on Build GRCh38
rs975282091 Chr.6: 144008709-144008709 on Build GRCh38
rs923709096 Chr.6: 144008595-144008595 on Build GRCh38
rs1001119952 Chr.6: 144008354-144008354 on Build GRCh38
rs567164800 Chr.6: 144008511-144008511 on Build GRCh38
rs1020030391 Chr.6: 144008653-144008653 on Build GRCh38
rs908620091 Chr.6: 144008739-144008739 on Build GRCh38
rs566539077 Chr.6: 144008582-144008582 on Build GRCh38
rs79396063 Chr.6: 144008589-144008589 on Build GRCh38
rs887367655 Chr.6: 144008470-144008470 on Build GRCh38
rs780471576 Chr.6: 144008666-144008666 on Build GRCh38
rs934043018 Chr.6: 144008574-144008574 on Build GRCh38
rs574088654 Chr.6: 144008672-144008672 on Build GRCh38
rs111938944 Chr.6: 144008475-144008475 on Build GRCh38
rs936362687 Chr.6: 144008505-144008505 on Build GRCh38
rs756166957 Chr.6: 144008655-144008655 on Build GRCh38
rs28364590 Chr.6: 144008677-144008677 on Build GRCh38
rs902731491 Chr.6: 144008353-144008353 on Build GRCh38
rs906739635 Chr.6: 144008524-144008524 on Build GRCh38
rs978826355 Chr.6: 144008657-144008657 on Build GRCh38
rs180731629 Chr.6: 144008555-144008555 on Build GRCh38
rs959773683 Chr.6: 144008510-144008510 on Build GRCh38
rs1046715771 Chr.6: 144008632-144008632 on Build GRCh38
rs775783341 Chr.6: 144008499-144008499 on Build GRCh38
rs970811325 Chr.6: 144008397-144008397 on Build GRCh38
rs576363135 Chr.6: 144008644-144008644 on Build GRCh38
rs1047062798 Chr.6: 144008577-144008577 on Build GRCh38
rs139952428 Chr.6: 144008751-144008751 on Build GRCh38
rs1053875080 Chr.6: 144008611-144008611 on Build GRCh38
rs922410726 Chr.6: 144008725-144008725 on Build GRCh38
rs529481417 Chr.6: 144008454-144008454 on Build GRCh38
rs555439966 Chr.6: 144008669-144008669 on Build GRCh38
rs1033042003 Chr.6: 144008378-144008378 on Build GRCh38
rs1025151871 Chr.6: 144008514-144008514 on Build GRCh38
rs1034134854 Chr.6: 144008501-144008501 on Build GRCh38
rs901245802 Chr.6: 144008456-144008456 on Build GRCh38
rs992495356 Chr.6: 144008515-144008515 on Build GRCh38
rs1054021866 Chr.6: 144008511-144008511 on Build GRCh38
rs1038642208 Chr.6: 144008610-144008610 on Build GRCh38
rs938234169 Chr.6: 144008517-144008517 on Build GRCh38
rs919863179 Chr.6: 144008466-144008466 on Build GRCh38
rs916800882 Chr.6: 144008617-144008617 on Build GRCh38
rs998180982 Chr.6: 144008458-144008458 on Build GRCh38
rs978701041 Chr.6: 144008551-144008551 on Build GRCh38
rs556110894 Chr.6: 144008730-144008730 on Build GRCh38
rs890896082 Chr.6: 144008339-144008339 on Build GRCh38
rs983412090 Chr.6: 144008398-144008398 on Build GRCh38
rs569512852 Chr.6: 144008495-144008495 on Build GRCh38
rs746410932 Chr.6: 144008463-144008463 on Build GRCh38
rs973832040 Chr.6: 144008543-144008543 on Build GRCh38
rs941346985 Chr.6: 144008590-144008590 on Build GRCh38

View Full Product Details