Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 169766538-169767082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERMARD
Entrez Gene ID
55780
Gene Name
ER membrane associated RNA degradation
Gene Aliases
C6orf70, PVNH6, dJ266L20.3
UniGene
Hs.47546
Gene Chromosome Location
Chr.6: 169751622-169781584 on Build GRCh38
Transcripts
NM_001278531.1 NM_001278532.1 NM_001278533.1 NM_018341.2 XM_011535938.2 XM_011535939.2 XM_011535940.2 XM_017011027.1 XM_017011028.1 XM_017011029.1 XM_017011030.1 XM_017011031.1 XM_017011032.1 XM_017011033.1

SNP Detail

SNP ID Chromosome Location
rs1035308163 Chr.6: 169766772-169766772 on Build GRCh38
rs373666092 Chr.6: 169766658-169766658 on Build GRCh38
rs911038460 Chr.6: 169766876-169766876 on Build GRCh38
rs535630504 Chr.6: 169767041-169767041 on Build GRCh38
rs764964589 Chr.6: 169766787-169766787 on Build GRCh38
rs112511356 Chr.6: 169767042-169767042 on Build GRCh38
rs368704279 Chr.6: 169766709-169766709 on Build GRCh38
rs182558816 Chr.6: 169766774-169766774 on Build GRCh38
rs745667163 Chr.6: 169766684-169766688 on Build GRCh38
rs1054187481 Chr.6: 169766979-169766979 on Build GRCh38
rs769710382 Chr.6: 169766697-169766700 on Build GRCh38
rs774926473 Chr.6: 169766618-169766618 on Build GRCh38
rs13199463 Chr.6: 169766576-169766576 on Build GRCh38
rs759562560 Chr.6: 169766604-169766604 on Build GRCh38
rs575001349 Chr.6: 169766992-169766992 on Build GRCh38
rs1049521259 Chr.6: 169766577-169766577 on Build GRCh38
rs536966443 Chr.6: 169766693-169766693 on Build GRCh38
rs1034504373 Chr.6: 169767043-169767043 on Build GRCh38
rs763756965 Chr.6: 169766846-169766846 on Build GRCh38
rs544085672 Chr.6: 169766988-169766988 on Build GRCh38
rs969819313 Chr.6: 169766764-169766764 on Build GRCh38
rs960180656 Chr.6: 169766789-169766789 on Build GRCh38
rs1051015699 Chr.6: 169766867-169766867 on Build GRCh38
rs762527819 Chr.6: 169766842-169766842 on Build GRCh38
rs913272576 Chr.6: 169766825-169766825 on Build GRCh38
rs573240983 Chr.6: 169766856-169766856 on Build GRCh38
rs889509489 Chr.6: 169766585-169766585 on Build GRCh38
rs563232602 Chr.6: 169766860-169766860 on Build GRCh38
rs780599068 Chr.6: 169766710-169766710 on Build GRCh38
rs952884913 Chr.6: 169766745-169766745 on Build GRCh38
rs746841069 Chr.6: 169766594-169766594 on Build GRCh38
rs768098325 Chr.6: 169766989-169766989 on Build GRCh38
rs200063598 Chr.6: 169766654-169766654 on Build GRCh38
rs370250565 Chr.6: 169766613-169766613 on Build GRCh38
rs756379986 Chr.6: 169767008-169767008 on Build GRCh38
rs896810326 Chr.6: 169766633-169766633 on Build GRCh38
rs1008378115 Chr.6: 169766598-169766598 on Build GRCh38
rs962427168 Chr.6: 169766840-169766840 on Build GRCh38
rs759208010 Chr.6: 169766641-169766641 on Build GRCh38
rs544867797 Chr.6: 169767048-169767048 on Build GRCh38
rs185014362 Chr.6: 169766905-169766905 on Build GRCh38
rs1040739207 Chr.6: 169766612-169766612 on Build GRCh38
rs1041183147 Chr.6: 169766904-169766904 on Build GRCh38
rs763698481 Chr.6: 169766648-169766648 on Build GRCh38
rs1003787799 Chr.6: 169766769-169766769 on Build GRCh38
rs75863442 Chr.6: 169766602-169766602 on Build GRCh38
rs905187639 Chr.6: 169766999-169766999 on Build GRCh38
rs761413776 Chr.6: 169766661-169766661 on Build GRCh38
rs540094127 Chr.6: 169766805-169766805 on Build GRCh38
rs1027159587 Chr.6: 169766655-169766655 on Build GRCh38
rs943839723 Chr.6: 169766896-169766896 on Build GRCh38
rs750726960 Chr.6: 169766699-169766699 on Build GRCh38
rs140056998 Chr.6: 169766950-169766950 on Build GRCh38
rs558206920 Chr.6: 169766828-169766828 on Build GRCh38
rs770884551 Chr.6: 169766599-169766599 on Build GRCh38
rs925154139 Chr.6: 169766853-169766853 on Build GRCh38
rs201484006 Chr.6: 169766622-169766622 on Build GRCh38
rs779174022 Chr.6: 169766587-169766587 on Build GRCh38
rs776352078 Chr.6: 169766602-169766605 on Build GRCh38

View Full Product Details