Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30924120-30924629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQP1
Entrez Gene ID
358
Gene Name
aquaporin 1 (Colton blood group)
Gene Aliases
AQP-CHIP, CHIP28, CO
UniGene
Hs.76152
Gene Chromosome Location
Chr.7: 30911800-30925516 on Build GRCh38
Transcripts
NM_001185060.1 NM_001185061.1 NM_001185062.1 NM_198098.2

SNP Detail

SNP ID Chromosome Location
rs916614938 Chr.7: 30924415-30924415 on Build GRCh38
rs556752153 Chr.7: 30924333-30924333 on Build GRCh38
rs146357004 Chr.7: 30924519-30924519 on Build GRCh38
rs888842508 Chr.7: 30924311-30924311 on Build GRCh38
rs1049305 Chr.7: 30924207-30924207 on Build GRCh38
rs993040652 Chr.7: 30924557-30924557 on Build GRCh38
rs754606739 Chr.7: 30924380-30924380 on Build GRCh38
rs1008586512 Chr.7: 30924547-30924547 on Build GRCh38
rs994450381 Chr.7: 30924495-30924495 on Build GRCh38
rs948186687 Chr.7: 30924271-30924271 on Build GRCh38
rs1041410605 Chr.7: 30924173-30924173 on Build GRCh38
rs1028696634 Chr.7: 30924310-30924310 on Build GRCh38
rs965683280 Chr.7: 30924246-30924246 on Build GRCh38
rs11537661 Chr.7: 30924530-30924530 on Build GRCh38
rs1018719121 Chr.7: 30924340-30924340 on Build GRCh38
rs1007044228 Chr.7: 30924425-30924425 on Build GRCh38
rs1019810181 Chr.7: 30924452-30924452 on Build GRCh38
rs916651172 Chr.7: 30924238-30924238 on Build GRCh38
rs917139385 Chr.7: 30924352-30924352 on Build GRCh38
rs904241462 Chr.7: 30924195-30924195 on Build GRCh38
rs1020282346 Chr.7: 30924242-30924242 on Build GRCh38
rs1036521655 Chr.7: 30924485-30924485 on Build GRCh38
rs533939937 Chr.7: 30924237-30924237 on Build GRCh38
rs542594185 Chr.7: 30924475-30924475 on Build GRCh38
rs952723631 Chr.7: 30924518-30924518 on Build GRCh38
rs1033335617 Chr.7: 30924366-30924366 on Build GRCh38
rs11537660 Chr.7: 30924154-30924154 on Build GRCh38
rs747652286 Chr.7: 30924508-30924508 on Build GRCh38
rs1018665261 Chr.7: 30924572-30924572 on Build GRCh38
rs958806907 Chr.7: 30924531-30924531 on Build GRCh38
rs954199500 Chr.7: 30924218-30924218 on Build GRCh38
rs536285394 Chr.7: 30924384-30924384 on Build GRCh38
rs542396091 Chr.7: 30924367-30924367 on Build GRCh38
rs575012514 Chr.7: 30924361-30924361 on Build GRCh38
rs572268335 Chr.7: 30924416-30924416 on Build GRCh38
rs940830145 Chr.7: 30924482-30924482 on Build GRCh38
rs1050499662 Chr.7: 30924525-30924525 on Build GRCh38
rs982140060 Chr.7: 30924459-30924459 on Build GRCh38
rs773740640 Chr.7: 30924596-30924596 on Build GRCh38
rs999910274 Chr.7: 30924368-30924368 on Build GRCh38
rs948072119 Chr.7: 30924445-30924445 on Build GRCh38
rs779662246 Chr.7: 30924290-30924290 on Build GRCh38
rs577290927 Chr.7: 30924297-30924297 on Build GRCh38
rs11537659 Chr.7: 30924274-30924274 on Build GRCh38
rs142345160 Chr.7: 30924328-30924328 on Build GRCh38
rs928056649 Chr.7: 30924477-30924477 on Build GRCh38
rs1010316295 Chr.7: 30924159-30924159 on Build GRCh38
rs189828375 Chr.7: 30924476-30924476 on Build GRCh38
rs899423536 Chr.7: 30924301-30924301 on Build GRCh38
rs994901260 Chr.7: 30924302-30924302 on Build GRCh38
rs925910143 Chr.7: 30924204-30924204 on Build GRCh38
rs999963907 Chr.7: 30924197-30924197 on Build GRCh38
rs989531322 Chr.7: 30924230-30924230 on Build GRCh38
rs778905104 Chr.7: 30924189-30924189 on Build GRCh38
rs1018811332 Chr.7: 30924150-30924150 on Build GRCh38
rs940883990 Chr.7: 30924294-30924294 on Build GRCh38
rs967179777 Chr.7: 30924360-30924360 on Build GRCh38
rs973132957 Chr.7: 30924269-30924269 on Build GRCh38
rs917107286 Chr.7: 30924569-30924569 on Build GRCh38

View Full Product Details