Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 36296856-36297352 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEPD1
Entrez Gene ID
80820
Gene Name
endonuclease/exonuclease/phosphatase family domain containing 1
Gene Aliases
HSPC107
UniGene
Hs.487994
Gene Chromosome Location
Chr.7: 36153149-36301543 on Build GRCh38
Transcripts
NM_030636.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7296653 c.1381C>A p.L461M Chr.7: 36297058-36297058 on Build GRCh38
5026739 c.1501G>T p.V501F Chr.7: 36297178-36297178 on Build GRCh38
6177539 c.1479G>T p.W493C Chr.7: 36297156-36297156 on Build GRCh38
4916393 c.1333A>G p.I445V Chr.7: 36297010-36297010 on Build GRCh38
4922474 c.1334T>C p.I445T Chr.7: 36297011-36297011 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372494407 Chr.7: 36297050-36297050 on Build GRCh38
rs781141453 Chr.7: 36297172-36297174 on Build GRCh38
rs757158300 Chr.7: 36297161-36297161 on Build GRCh38
rs373260241 Chr.7: 36297203-36297203 on Build GRCh38
rs773807961 Chr.7: 36297098-36297098 on Build GRCh38
rs766479139 Chr.7: 36296961-36296961 on Build GRCh38
rs577655317 Chr.7: 36296911-36296911 on Build GRCh38
rs765503854 Chr.7: 36297134-36297134 on Build GRCh38
rs953950974 Chr.7: 36297015-36297015 on Build GRCh38
rs916113523 Chr.7: 36297199-36297199 on Build GRCh38
rs781064146 Chr.7: 36297170-36297170 on Build GRCh38
rs940018002 Chr.7: 36297244-36297244 on Build GRCh38
rs1028456419 Chr.7: 36296887-36296887 on Build GRCh38
rs901430743 Chr.7: 36297269-36297269 on Build GRCh38
rs376545349 Chr.7: 36297235-36297235 on Build GRCh38
rs757041039 Chr.7: 36297056-36297056 on Build GRCh38
rs779055419 Chr.7: 36296993-36296993 on Build GRCh38
rs752697832 Chr.7: 36297135-36297135 on Build GRCh38
rs182014927 Chr.7: 36296916-36296916 on Build GRCh38
rs907169885 Chr.7: 36297241-36297241 on Build GRCh38
rs1045752847 Chr.7: 36296883-36296883 on Build GRCh38
rs898564354 Chr.7: 36297282-36297282 on Build GRCh38
rs140394885 Chr.7: 36297091-36297091 on Build GRCh38
rs185730374 Chr.7: 36296957-36296957 on Build GRCh38
rs570300492 Chr.7: 36296910-36296910 on Build GRCh38
rs765235830 Chr.7: 36297019-36297019 on Build GRCh38
rs778307507 Chr.7: 36297236-36297236 on Build GRCh38
rs986714523 Chr.7: 36297029-36297029 on Build GRCh38
rs996025488 Chr.7: 36297312-36297312 on Build GRCh38
rs932890119 Chr.7: 36297290-36297290 on Build GRCh38
rs1024893793 Chr.7: 36297088-36297088 on Build GRCh38
rs146771076 Chr.7: 36297003-36297003 on Build GRCh38
rs759752037 Chr.7: 36297114-36297114 on Build GRCh38
rs369979437 Chr.7: 36296958-36296958 on Build GRCh38
rs755811338 Chr.7: 36297196-36297196 on Build GRCh38
rs748714888 Chr.7: 36297215-36297215 on Build GRCh38
rs771234485 Chr.7: 36297102-36297102 on Build GRCh38
rs752843855 Chr.7: 36297036-36297036 on Build GRCh38
rs776992343 Chr.7: 36297113-36297113 on Build GRCh38
rs199807446 Chr.7: 36297145-36297145 on Build GRCh38
rs370007109 Chr.7: 36297093-36297093 on Build GRCh38
rs867175742 Chr.7: 36297205-36297205 on Build GRCh38
rs772596995 Chr.7: 36297178-36297178 on Build GRCh38
rs758255571 Chr.7: 36297038-36297038 on Build GRCh38
rs745462745 Chr.7: 36297090-36297090 on Build GRCh38
rs1049574368 Chr.7: 36297320-36297320 on Build GRCh38
rs375762204 Chr.7: 36297096-36297096 on Build GRCh38
rs755213697 Chr.7: 36297007-36297007 on Build GRCh38
rs1015688416 Chr.7: 36296930-36296930 on Build GRCh38
rs201649550 Chr.7: 36297034-36297034 on Build GRCh38
rs79767813 Chr.7: 36296915-36296915 on Build GRCh38
rs761116760 Chr.7: 36297100-36297100 on Build GRCh38
rs866565277 Chr.7: 36296931-36296931 on Build GRCh38
rs745771307 Chr.7: 36297195-36297195 on Build GRCh38
rs751284587 Chr.7: 36297051-36297051 on Build GRCh38
rs747814725 Chr.7: 36296938-36296938 on Build GRCh38
rs149881014 Chr.7: 36297092-36297092 on Build GRCh38
rs904617726 Chr.7: 36296903-36296903 on Build GRCh38
rs7778561 Chr.7: 36297265-36297265 on Build GRCh38
rs763134888 Chr.7: 36297143-36297143 on Build GRCh38
rs751469851 Chr.7: 36297151-36297151 on Build GRCh38
rs781159525 Chr.7: 36297072-36297072 on Build GRCh38

View Full Product Details