Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 47658823-47659326 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf65
Entrez Gene ID
401335
Gene Name
chromosome 7 open reading frame 65
Gene Aliases
-
UniGene
Hs.520816
Gene Chromosome Location
Chr.7: 47655244-47661648 on Build GRCh38
Transcripts
NM_001123065.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5617413 c.351A>G p.L117L Chr.7: 47659123-47659123 on Build GRCh38
319129 c.185G>C p.R62T Chr.7: 47658957-47658957 on Build GRCh38
5700580 c.295G>A p.G99S Chr.7: 47659067-47659067 on Build GRCh38
3263623 c.211A>T p.S71C Chr.7: 47658983-47658983 on Build GRCh38
6177744 c.416A>G p.K139R Chr.7: 47659188-47659188 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746173827 Chr.7: 47659257-47659257 on Build GRCh38
rs764645662 Chr.7: 47659166-47659166 on Build GRCh38
rs952103712 Chr.7: 47659292-47659292 on Build GRCh38
rs370571842 Chr.7: 47658860-47658860 on Build GRCh38
rs779809697 Chr.7: 47658946-47658947 on Build GRCh38
rs1001251210 Chr.7: 47659043-47659043 on Build GRCh38
rs201413304 Chr.7: 47658909-47658909 on Build GRCh38
rs749604619 Chr.7: 47659058-47659058 on Build GRCh38
rs752573497 Chr.7: 47659225-47659225 on Build GRCh38
rs905332945 Chr.7: 47659248-47659248 on Build GRCh38
rs765570303 Chr.7: 47658995-47658995 on Build GRCh38
rs984107350 Chr.7: 47659293-47659293 on Build GRCh38
rs746942023 Chr.7: 47659040-47659040 on Build GRCh38
rs888200446 Chr.7: 47659035-47659035 on Build GRCh38
rs775594736 Chr.7: 47658888-47658888 on Build GRCh38
rs527897958 Chr.7: 47659160-47659160 on Build GRCh38
rs535576401 Chr.7: 47659041-47659041 on Build GRCh38
rs372501553 Chr.7: 47659109-47659109 on Build GRCh38
rs200327521 Chr.7: 47659249-47659249 on Build GRCh38
rs368460508 Chr.7: 47659235-47659235 on Build GRCh38
rs773552161 Chr.7: 47659132-47659132 on Build GRCh38
rs201809988 Chr.7: 47659208-47659208 on Build GRCh38
rs779791666 Chr.7: 47659098-47659098 on Build GRCh38
rs751316003 Chr.7: 47659189-47659189 on Build GRCh38
rs115611995 Chr.7: 47659009-47659009 on Build GRCh38
rs769992699 Chr.7: 47659049-47659049 on Build GRCh38
rs571868304 Chr.7: 47658933-47658933 on Build GRCh38
rs769532649 Chr.7: 47658869-47658869 on Build GRCh38
rs35314142 Chr.7: 47659150-47659151 on Build GRCh38
rs771242118 Chr.7: 47659246-47659246 on Build GRCh38
rs768074058 Chr.7: 47659158-47659158 on Build GRCh38
rs760155784 Chr.7: 47659150-47659150 on Build GRCh38
rs768705655 Chr.7: 47658992-47658992 on Build GRCh38
rs775445551 Chr.7: 47659209-47659219 on Build GRCh38
rs772097438 Chr.7: 47658879-47658879 on Build GRCh38
rs974722972 Chr.7: 47659057-47659057 on Build GRCh38
rs772002000 Chr.7: 47658979-47658979 on Build GRCh38
rs114109572 Chr.7: 47659139-47659139 on Build GRCh38
rs540115520 Chr.7: 47659117-47659117 on Build GRCh38
rs944971853 Chr.7: 47658960-47658960 on Build GRCh38
rs774223449 Chr.7: 47658874-47658874 on Build GRCh38
rs202217764 Chr.7: 47659172-47659172 on Build GRCh38
rs778751610 Chr.7: 47658971-47658971 on Build GRCh38
rs372106308 Chr.7: 47659081-47659081 on Build GRCh38
rs759489502 Chr.7: 47658876-47658876 on Build GRCh38
rs560148679 Chr.7: 47659121-47659121 on Build GRCh38
rs377259928 Chr.7: 47658890-47658890 on Build GRCh38
rs534484572 Chr.7: 47658953-47658953 on Build GRCh38
rs755754059 Chr.7: 47659021-47659021 on Build GRCh38
rs12531737 Chr.7: 47659087-47659087 on Build GRCh38
rs772233330 Chr.7: 47659176-47659176 on Build GRCh38
rs375012975 Chr.7: 47658957-47658957 on Build GRCh38
rs1001905395 Chr.7: 47658908-47658908 on Build GRCh38
rs765627902 Chr.7: 47658943-47658943 on Build GRCh38
rs746996669 Chr.7: 47659106-47659106 on Build GRCh38
rs745818820 Chr.7: 47658972-47658972 on Build GRCh38
rs766650573 Chr.7: 47659064-47659064 on Build GRCh38
rs769393027 Chr.7: 47659280-47659280 on Build GRCh38
rs372865348 Chr.7: 47658873-47658873 on Build GRCh38
rs772548647 Chr.7: 47659259-47659259 on Build GRCh38
rs781280787 Chr.7: 47659116-47659116 on Build GRCh38
rs868112972 Chr.7: 47659010-47659010 on Build GRCh38
rs944710627 Chr.7: 47659212-47659212 on Build GRCh38
rs556116108 Chr.7: 47658927-47658927 on Build GRCh38
rs756259297 Chr.7: 47659228-47659228 on Build GRCh38
rs764230955 Chr.7: 47658892-47658892 on Build GRCh38
rs370939216 Chr.7: 47658998-47658998 on Build GRCh38
rs756889678 Chr.7: 47659026-47659026 on Build GRCh38
rs1049266401 Chr.7: 47659247-47659247 on Build GRCh38
rs368003065 Chr.7: 47659135-47659135 on Build GRCh38
rs375333624 Chr.7: 47658984-47658984 on Build GRCh38
rs763435054 Chr.7: 47659060-47659060 on Build GRCh38
rs754923191 Chr.7: 47659192-47659192 on Build GRCh38
rs760100931 Chr.7: 47659070-47659070 on Build GRCh38
rs758277977 Chr.7: 47659038-47659038 on Build GRCh38
rs1037054959 Chr.7: 47659219-47659219 on Build GRCh38
rs371871055 Chr.7: 47658963-47658963 on Build GRCh38
rs750027131 Chr.7: 47659096-47659096 on Build GRCh38
rs766181828 Chr.7: 47659184-47659184 on Build GRCh38
rs761440956 Chr.7: 47659164-47659164 on Build GRCh38
rs201291615 Chr.7: 47659197-47659197 on Build GRCh38
rs769651161 Chr.7: 47659198-47659198 on Build GRCh38
rs777715066 Chr.7: 47658871-47658871 on Build GRCh38
rs113647031 Chr.7: 47659148-47659148 on Build GRCh38
rs994997270 Chr.7: 47658991-47658991 on Build GRCh38
rs749441446 Chr.7: 47659133-47659133 on Build GRCh38
rs776128366 Chr.7: 47659272-47659272 on Build GRCh38
rs370209990 Chr.7: 47658952-47658952 on Build GRCh38
rs964767596 Chr.7: 47659238-47659238 on Build GRCh38
rs769750074 Chr.7: 47659124-47659124 on Build GRCh38
rs375422953 Chr.7: 47658911-47658911 on Build GRCh38
rs749550998 Chr.7: 47659240-47659240 on Build GRCh38
rs770674705 Chr.7: 47658970-47658970 on Build GRCh38
rs776904379 Chr.7: 47658993-47658993 on Build GRCh38
rs866953623 Chr.7: 47659126-47659126 on Build GRCh38
rs745606738 Chr.7: 47659037-47659037 on Build GRCh38
rs575354469 Chr.7: 47659085-47659085 on Build GRCh38
rs577917764 Chr.7: 47659097-47659097 on Build GRCh38
rs763669231 Chr.7: 47659023-47659023 on Build GRCh38
rs565332971 Chr.7: 47659253-47659253 on Build GRCh38
rs779536067 Chr.7: 47659102-47659102 on Build GRCh38
rs370044672 Chr.7: 47659094-47659094 on Build GRCh38
rs760688238 Chr.7: 47658987-47658987 on Build GRCh38
rs745591002 Chr.7: 47658869-47658869 on Build GRCh38
rs773462076 Chr.7: 47659051-47659051 on Build GRCh38
rs368781043 Chr.7: 47658936-47658936 on Build GRCh38
rs76203945 Chr.7: 47658962-47658962 on Build GRCh38
rs902340361 Chr.7: 47659193-47659193 on Build GRCh38
rs551789387 Chr.7: 47658872-47658872 on Build GRCh38
rs766851688 Chr.7: 47659003-47659003 on Build GRCh38
rs528016461 Chr.7: 47659291-47659291 on Build GRCh38
rs774810478 Chr.7: 47659145-47659145 on Build GRCh38
rs750750036 Chr.7: 47658997-47658997 on Build GRCh38
rs753596399 Chr.7: 47659024-47659024 on Build GRCh38
rs538889219 Chr.7: 47659127-47659127 on Build GRCh38
rs763374504 Chr.7: 47659002-47659002 on Build GRCh38
rs778608178 Chr.7: 47659031-47659031 on Build GRCh38
rs776669132 Chr.7: 47659042-47659042 on Build GRCh38

View Full Product Details