Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100125042-100125542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNPY4
Entrez Gene ID
245812
Gene Name
canopy FGF signaling regulator 4
Gene Aliases
PRAT4B
UniGene
Hs.632293
Gene Chromosome Location
Chr.7: 100119642-100125508 on Build GRCh38
Transcripts
NM_152755.1

SNP Detail

SNP ID Chromosome Location
rs1052134362 Chr.7: 100125343-100125343 on Build GRCh38
rs1011130501 Chr.7: 100125383-100125383 on Build GRCh38
rs1014939598 Chr.7: 100125459-100125459 on Build GRCh38
rs932529605 Chr.7: 100125330-100125330 on Build GRCh38
rs1002288845 Chr.7: 100125423-100125423 on Build GRCh38
rs1040858172 Chr.7: 100125311-100125311 on Build GRCh38
rs879894068 Chr.7: 100125261-100125261 on Build GRCh38
rs928036076 Chr.7: 100125130-100125130 on Build GRCh38
rs561769183 Chr.7: 100125426-100125426 on Build GRCh38
rs931771484 Chr.7: 100125291-100125291 on Build GRCh38
rs926090096 Chr.7: 100125273-100125273 on Build GRCh38
rs890253461 Chr.7: 100125312-100125312 on Build GRCh38
rs528403959 Chr.7: 100125321-100125321 on Build GRCh38
rs888236230 Chr.7: 100125403-100125403 on Build GRCh38
rs969535034 Chr.7: 100125409-100125410 on Build GRCh38
rs908525291 Chr.7: 100125234-100125235 on Build GRCh38
rs1024896796 Chr.7: 100125135-100125135 on Build GRCh38
rs528835283 Chr.7: 100125441-100125441 on Build GRCh38
rs961657612 Chr.7: 100125126-100125126 on Build GRCh38
rs1005215226 Chr.7: 100125422-100125422 on Build GRCh38
rs970145006 Chr.7: 100125204-100125204 on Build GRCh38
rs931886052 Chr.7: 100125508-100125508 on Build GRCh38
rs945295935 Chr.7: 100125309-100125309 on Build GRCh38
rs183981798 Chr.7: 100125274-100125274 on Build GRCh38
rs747278381 Chr.7: 100125308-100125308 on Build GRCh38
rs1049924278 Chr.7: 100125354-100125354 on Build GRCh38
rs556342233 Chr.7: 100125078-100125078 on Build GRCh38
rs771149508 Chr.7: 100125490-100125490 on Build GRCh38
rs575412081 Chr.7: 100125107-100125107 on Build GRCh38
rs74946837 Chr.7: 100125379-100125379 on Build GRCh38
rs982841891 Chr.7: 100125229-100125229 on Build GRCh38
rs962674879 Chr.7: 100125475-100125475 on Build GRCh38
rs960993117 Chr.7: 100125195-100125195 on Build GRCh38
rs11763611 Chr.7: 100125276-100125276 on Build GRCh38
rs988804308 Chr.7: 100125297-100125297 on Build GRCh38
rs1024450152 Chr.7: 100125470-100125470 on Build GRCh38
rs545565953 Chr.7: 100125258-100125258 on Build GRCh38
rs369281742 Chr.7: 100125325-100125325 on Build GRCh38
rs540125560 Chr.7: 100125425-100125425 on Build GRCh38
rs920472648 Chr.7: 100125494-100125494 on Build GRCh38
rs980833759 Chr.7: 100125122-100125122 on Build GRCh38
rs372603498 Chr.7: 100125456-100125456 on Build GRCh38
rs992963330 Chr.7: 100125134-100125134 on Build GRCh38
rs892112713 Chr.7: 100125348-100125348 on Build GRCh38
rs1022474197 Chr.7: 100125389-100125389 on Build GRCh38

View Full Product Details