Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 107203523-107204035 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COG5
Entrez Gene ID
10466
Gene Name
component of oligomeric golgi complex 5
Gene Aliases
CDG2I, GOLTC1, GTC90
UniGene
Hs.239631
Gene Chromosome Location
Chr.7: 107201743-107564514 on Build GRCh38
Transcripts
NM_006348.3 NM_181733.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4155364 c.2470G>T p.G824* Chr.7: 107203629-107203629 on Build GRCh38
5434193 c.2513G>A p.G838D Chr.7: 107203586-107203586 on Build GRCh38
1568708 c.2489T>G p.V830G Chr.7: 107203610-107203610 on Build GRCh38
1083840 c.2518G>T p.E840* Chr.7: 107203581-107203581 on Build GRCh38
6536353 c.2526A>C p.A842A Chr.7: 107203573-107203573 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778275731 Chr.7: 107203714-107203714 on Build GRCh38
rs551961576 Chr.7: 107203692-107203692 on Build GRCh38
rs977132495 Chr.7: 107203859-107203859 on Build GRCh38
rs777292941 Chr.7: 107203680-107203680 on Build GRCh38
rs775853693 Chr.7: 107203573-107203573 on Build GRCh38
rs567077223 Chr.7: 107203992-107203992 on Build GRCh38
rs369522129 Chr.7: 107203666-107203666 on Build GRCh38
rs201706597 Chr.7: 107203644-107203644 on Build GRCh38
rs1049500887 Chr.7: 107203799-107203799 on Build GRCh38
rs944860654 Chr.7: 107203907-107203907 on Build GRCh38
rs889454863 Chr.7: 107203556-107203556 on Build GRCh38
rs769417529 Chr.7: 107203667-107203667 on Build GRCh38
rs983903809 Chr.7: 107203766-107203766 on Build GRCh38
rs1025955056 Chr.7: 107203723-107203723 on Build GRCh38
rs762082725 Chr.7: 107203607-107203607 on Build GRCh38
rs754773616 Chr.7: 107203660-107203660 on Build GRCh38
rs760038268 Chr.7: 107203555-107203555 on Build GRCh38
rs896826430 Chr.7: 107203842-107203842 on Build GRCh38
rs567947457 Chr.7: 107203886-107203886 on Build GRCh38
rs970023291 Chr.7: 107203661-107203661 on Build GRCh38
rs1012069291 Chr.7: 107203933-107203933 on Build GRCh38
rs12537730 Chr.7: 107203836-107203836 on Build GRCh38
rs780927785 Chr.7: 107203810-107203810 on Build GRCh38
rs897091614 Chr.7: 107203579-107203579 on Build GRCh38
rs557342098 Chr.7: 107204008-107204008 on Build GRCh38
rs542733135 Chr.7: 107203887-107203887 on Build GRCh38
rs956288651 Chr.7: 107203713-107203713 on Build GRCh38
rs1014252892 Chr.7: 107203640-107203640 on Build GRCh38
rs758443491 Chr.7: 107203638-107203638 on Build GRCh38
rs781040627 Chr.7: 107203663-107203663 on Build GRCh38
rs941004561 Chr.7: 107203823-107203823 on Build GRCh38
rs878866018 Chr.7: 107203989-107203989 on Build GRCh38
rs1050606884 Chr.7: 107203574-107203574 on Build GRCh38
rs565386860 Chr.7: 107203820-107203820 on Build GRCh38
rs757235456 Chr.7: 107203773-107203773 on Build GRCh38
rs760015202 Chr.7: 107203603-107203606 on Build GRCh38
rs888353477 Chr.7: 107204006-107204006 on Build GRCh38
rs911005744 Chr.7: 107203811-107203811 on Build GRCh38
rs757371565 Chr.7: 107203894-107203894 on Build GRCh38
rs367597633 Chr.7: 107203834-107203834 on Build GRCh38
rs993521985 Chr.7: 107203706-107203706 on Build GRCh38
rs201334786 Chr.7: 107203586-107203586 on Build GRCh38
rs1000701645 Chr.7: 107203958-107203958 on Build GRCh38
rs889191758 Chr.7: 107203584-107203584 on Build GRCh38
rs993885403 Chr.7: 107203976-107203976 on Build GRCh38
rs556891535 Chr.7: 107203660-107203661 on Build GRCh38
rs945674155 Chr.7: 107203851-107203851 on Build GRCh38
rs926179361 Chr.7: 107203868-107203868 on Build GRCh38
rs922728833 Chr.7: 107203736-107203736 on Build GRCh38
rs750462856 Chr.7: 107203622-107203622 on Build GRCh38
rs141942249 Chr.7: 107203605-107203605 on Build GRCh38
rs958444049 Chr.7: 107203803-107203803 on Build GRCh38
rs868650147 Chr.7: 107203583-107203583 on Build GRCh38
rs1018955164 Chr.7: 107203629-107203629 on Build GRCh38
rs34100214 Chr.7: 107203550-107203550 on Build GRCh38
rs1040828486 Chr.7: 107203946-107203946 on Build GRCh38
rs1032981706 Chr.7: 107203959-107203959 on Build GRCh38
rs200090407 Chr.7: 107203602-107203602 on Build GRCh38
rs780093185 Chr.7: 107203651-107203651 on Build GRCh38
rs1005257968 Chr.7: 107203557-107203557 on Build GRCh38
rs548306418 Chr.7: 107203844-107203844 on Build GRCh38
rs970092559 Chr.7: 107203952-107203952 on Build GRCh38
rs952681190 Chr.7: 107203746-107203746 on Build GRCh38
rs1017248688 Chr.7: 107203560-107203560 on Build GRCh38
rs989075929 Chr.7: 107203993-107203995 on Build GRCh38
rs1022992387 Chr.7: 107203943-107203943 on Build GRCh38
rs1027681694 Chr.7: 107203988-107203988 on Build GRCh38
rs184346859 Chr.7: 107203972-107203972 on Build GRCh38
rs188853071 Chr.7: 107204007-107204007 on Build GRCh38
rs934187968 Chr.7: 107203753-107203753 on Build GRCh38
rs751523967 Chr.7: 107203658-107203658 on Build GRCh38
rs989476612 Chr.7: 107203722-107203722 on Build GRCh38
rs772525300 Chr.7: 107203565-107203565 on Build GRCh38

View Full Product Details