Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 90051575-90052124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DECR1
Entrez Gene ID
1666
Gene Name
2,4-dienoyl-CoA reductase 1, mitochondrial
Gene Aliases
DECR, NADPH, SDR18C1
UniGene
Hs.492212
Gene Chromosome Location
Chr.8: 90001337-90052004 on Build GRCh38
Transcripts
NM_001359.1 XM_005250808.4 XM_011516880.2 XM_017013147.1 XM_017013148.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3901985 c.900C>T p.D300D Chr.8: 90051691-90051691 on Build GRCh38
6114138 c.948+1G>T p.? Chr.8: 90051740-90051740 on Build GRCh38
6430154 c.940C>A p.L314M Chr.8: 90051731-90051731 on Build GRCh38
5063368 c.894A>G p.K298K Chr.8: 90051685-90051685 on Build GRCh38
6429623 c.907G>T p.E303* Chr.8: 90051698-90051698 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776442947 Chr.8: 90051662-90051662 on Build GRCh38
rs754221305 Chr.8: 90051652-90051652 on Build GRCh38
rs778132448 Chr.8: 90051705-90051705 on Build GRCh38
rs759151725 Chr.8: 90051934-90051934 on Build GRCh38
rs758008347 Chr.8: 90051912-90051913 on Build GRCh38
rs766014372 Chr.8: 90051691-90051691 on Build GRCh38
rs747193294 Chr.8: 90051735-90051735 on Build GRCh38
rs998274646 Chr.8: 90051797-90051797 on Build GRCh38
rs772160149 Chr.8: 90051882-90051882 on Build GRCh38
rs1476590 Chr.8: 90051808-90051808 on Build GRCh38
rs775628189 Chr.8: 90051731-90051731 on Build GRCh38
rs774123923 Chr.8: 90051921-90051921 on Build GRCh38
rs780110714 Chr.8: 90051799-90051799 on Build GRCh38
rs750967110 Chr.8: 90051870-90051870 on Build GRCh38
rs770378450 Chr.8: 90051838-90051838 on Build GRCh38
rs749987681 Chr.8: 90051674-90051674 on Build GRCh38
rs752390775 Chr.8: 90051862-90051862 on Build GRCh38
rs779627906 Chr.8: 90051792-90051792 on Build GRCh38
rs764330022 Chr.8: 90051684-90051685 on Build GRCh38
rs763932796 Chr.8: 90051660-90051660 on Build GRCh38
rs755011769 Chr.8: 90051889-90051889 on Build GRCh38
rs111307450 Chr.8: 90051887-90051887 on Build GRCh38
rs765635535 Chr.8: 90051864-90051864 on Build GRCh38
rs757788865 Chr.8: 90051722-90051722 on Build GRCh38
rs758945732 Chr.8: 90051848-90051848 on Build GRCh38
rs3180374 Chr.8: 90052003-90052003 on Build GRCh38
rs747222335 Chr.8: 90051807-90051807 on Build GRCh38
rs751176360 Chr.8: 90051770-90051770 on Build GRCh38
rs761950491 Chr.8: 90051747-90051750 on Build GRCh38
rs759186825 Chr.8: 90051777-90051777 on Build GRCh38
rs768371218 Chr.8: 90051740-90051740 on Build GRCh38
rs375403201 Chr.8: 90051960-90051960 on Build GRCh38
rs769565252 Chr.8: 90051833-90051833 on Build GRCh38
rs750950640 Chr.8: 90051796-90051796 on Build GRCh38
rs774532339 Chr.8: 90051629-90051629 on Build GRCh38
rs554605005 Chr.8: 90051640-90051640 on Build GRCh38
rs112034915 Chr.8: 90051917-90051917 on Build GRCh38
rs749843604 Chr.8: 90051717-90051717 on Build GRCh38
rs15094 Chr.8: 90051888-90051888 on Build GRCh38
rs766846448 Chr.8: 90051901-90051904 on Build GRCh38
rs767238001 Chr.8: 90051854-90051854 on Build GRCh38
rs149321434 Chr.8: 90051918-90051918 on Build GRCh38
rs745981525 Chr.8: 90051728-90051728 on Build GRCh38
rs754398418 Chr.8: 90051910-90051911 on Build GRCh38
rs200122846 Chr.8: 90051890-90051890 on Build GRCh38
rs928546745 Chr.8: 90052093-90052093 on Build GRCh38
rs777017998 Chr.8: 90052093-90052093 on Build GRCh38
rs779204338 Chr.8: 90051692-90051692 on Build GRCh38
rs750779359 Chr.8: 90051854-90051854 on Build GRCh38
rs776442151 Chr.8: 90051743-90051743 on Build GRCh38
rs759387645 Chr.8: 90052080-90052080 on Build GRCh38
rs777504937 Chr.8: 90051903-90051903 on Build GRCh38
rs773147598 Chr.8: 90051836-90051836 on Build GRCh38
rs746750395 Chr.8: 90051913-90051914 on Build GRCh38
rs772470843 Chr.8: 90051646-90051646 on Build GRCh38
rs774712484 Chr.8: 90051742-90051745 on Build GRCh38
rs757874448 Chr.8: 90051679-90051679 on Build GRCh38
rs1016027407 Chr.8: 90051988-90051988 on Build GRCh38
rs758417462 Chr.8: 90051798-90051798 on Build GRCh38
rs763103653 Chr.8: 90051679-90051684 on Build GRCh38
rs7162 Chr.8: 90051898-90051898 on Build GRCh38
rs962951669 Chr.8: 90051883-90051883 on Build GRCh38
rs761575792 Chr.8: 90051755-90051755 on Build GRCh38
rs761704057 Chr.8: 90051668-90051668 on Build GRCh38
rs748006825 Chr.8: 90051824-90051824 on Build GRCh38
rs113652590 Chr.8: 90051742-90051742 on Build GRCh38
rs773862868 Chr.8: 90051844-90051844 on Build GRCh38
rs767869784 Chr.8: 90051808-90051809 on Build GRCh38
rs377574337 Chr.8: 90051727-90051727 on Build GRCh38
rs954034169 Chr.8: 90051813-90051813 on Build GRCh38
rs776751707 Chr.8: 90051630-90051630 on Build GRCh38
rs985434184 Chr.8: 90051845-90051845 on Build GRCh38
rs892829075 Chr.8: 90051979-90051979 on Build GRCh38
rs757664185 Chr.8: 90051783-90051783 on Build GRCh38
rs577455044 Chr.8: 90051871-90051871 on Build GRCh38
rs754301897 Chr.8: 90051782-90051782 on Build GRCh38
rs755244555 Chr.8: 90051671-90051671 on Build GRCh38
rs936959453 Chr.8: 90051949-90051949 on Build GRCh38
rs952873871 Chr.8: 90051958-90051958 on Build GRCh38
rs372463312 Chr.8: 90051650-90051650 on Build GRCh38
rs772185951 Chr.8: 90051730-90051730 on Build GRCh38
rs762323323 Chr.8: 90051863-90051863 on Build GRCh38
rs756447890 Chr.8: 90051699-90051699 on Build GRCh38
rs773131144 Chr.8: 90051757-90051757 on Build GRCh38
rs945931673 Chr.8: 90052049-90052049 on Build GRCh38
rs762510011 Chr.8: 90051758-90051758 on Build GRCh38
rs969846578 Chr.8: 90052011-90052011 on Build GRCh38
rs771691905 Chr.8: 90051935-90051935 on Build GRCh38
rs760450928 Chr.8: 90051658-90051658 on Build GRCh38
rs376370571 Chr.8: 90052092-90052093 on Build GRCh38
rs764388131 Chr.8: 90051780-90051780 on Build GRCh38
rs756538937 Chr.8: 90051864-90051867 on Build GRCh38
rs559137210 Chr.8: 90051816-90051816 on Build GRCh38
rs776347130 Chr.8: 90052076-90052076 on Build GRCh38
rs541854163 Chr.8: 90051985-90051985 on Build GRCh38
rs765856938 Chr.8: 90051769-90051769 on Build GRCh38
rs762811591 Chr.8: 90051837-90051837 on Build GRCh38
rs745887799 Chr.8: 90051768-90051768 on Build GRCh38
rs780408237 Chr.8: 90051881-90051881 on Build GRCh38

View Full Product Details