Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 131944664-131945179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFR3A
Entrez Gene ID
23167
Gene Name
EFR3 homolog A
Gene Aliases
-
UniGene
Hs.204564
Gene Chromosome Location
Chr.8: 131904088-132013642 on Build GRCh38
Transcripts
NM_001323553.1 NM_001323554.1 NM_001323555.1 NM_001323556.1 NM_001323557.1 NM_001323558.1 NM_015137.5 NR_136615.1 NR_136616.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
274944 c.158G>A p.R53Q Chr.8: 131944815-131944815 on Build GRCh38
486091 c.95T>G p.L32R Chr.8: 131944752-131944752 on Build GRCh38
396789 c.215G>A p.G72E Chr.8: 131944872-131944872 on Build GRCh38
6179972 c.190A>C p.R64R Chr.8: 131944847-131944847 on Build GRCh38
7182567 c.215+2T>C p.? Chr.8: 131944874-131944874 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774734048 Chr.8: 131944958-131944958 on Build GRCh38
rs545230669 Chr.8: 131944701-131944701 on Build GRCh38
rs770205932 Chr.8: 131944703-131944703 on Build GRCh38
rs755336530 Chr.8: 131945121-131945122 on Build GRCh38
rs768002043 Chr.8: 131944743-131944743 on Build GRCh38
rs375503062 Chr.8: 131944763-131944763 on Build GRCh38
rs1041250615 Chr.8: 131944923-131944923 on Build GRCh38
rs536766867 Chr.8: 131945032-131945032 on Build GRCh38
rs1000908508 Chr.8: 131944724-131944724 on Build GRCh38
rs781271015 Chr.8: 131944789-131944789 on Build GRCh38
rs757875404 Chr.8: 131944767-131944767 on Build GRCh38
rs555827880 Chr.8: 131944932-131944932 on Build GRCh38
rs373337601 Chr.8: 131944859-131944859 on Build GRCh38
rs761046290 Chr.8: 131944881-131944881 on Build GRCh38
rs573646376 Chr.8: 131945141-131945141 on Build GRCh38
rs756796163 Chr.8: 131944908-131944908 on Build GRCh38
rs936951171 Chr.8: 131944966-131944966 on Build GRCh38
rs768632978 Chr.8: 131944823-131944823 on Build GRCh38
rs550221945 Chr.8: 131944710-131944710 on Build GRCh38
rs751180528 Chr.8: 131944900-131944900 on Build GRCh38
rs539697739 Chr.8: 131944888-131944888 on Build GRCh38
rs148616251 Chr.8: 131945082-131945082 on Build GRCh38
rs766296116 Chr.8: 131944910-131944910 on Build GRCh38
rs774551723 Chr.8: 131944827-131944827 on Build GRCh38
rs562018150 Chr.8: 131944775-131944775 on Build GRCh38
rs530931394 Chr.8: 131944931-131944931 on Build GRCh38
rs1000856260 Chr.8: 131945119-131945119 on Build GRCh38
rs908676427 Chr.8: 131945079-131945079 on Build GRCh38
rs1019671910 Chr.8: 131944869-131944869 on Build GRCh38
rs143933422 Chr.8: 131944815-131944815 on Build GRCh38
rs959471829 Chr.8: 131944770-131944770 on Build GRCh38
rs374918162 Chr.8: 131944722-131944722 on Build GRCh38
rs750346436 Chr.8: 131944914-131944914 on Build GRCh38
rs773690040 Chr.8: 131944875-131944875 on Build GRCh38
rs563434419 Chr.8: 131944702-131944702 on Build GRCh38
rs996807666 Chr.8: 131944894-131944894 on Build GRCh38
rs1013683683 Chr.8: 131945143-131945143 on Build GRCh38
rs769584711 Chr.8: 131944814-131944814 on Build GRCh38
rs202067577 Chr.8: 131944780-131944780 on Build GRCh38
rs768744623 Chr.8: 131944712-131944712 on Build GRCh38
rs760999169 Chr.8: 131944764-131944764 on Build GRCh38
rs146485355 Chr.8: 131944861-131944861 on Build GRCh38
rs771752586 Chr.8: 131944873-131944873 on Build GRCh38
rs749463078 Chr.8: 131944820-131944820 on Build GRCh38
rs943809822 Chr.8: 131944918-131944918 on Build GRCh38
rs755866169 Chr.8: 131944922-131944922 on Build GRCh38
rs376461585 Chr.8: 131944911-131944911 on Build GRCh38
rs766722660 Chr.8: 131944886-131944886 on Build GRCh38
rs775748311 Chr.8: 131944709-131944709 on Build GRCh38
rs919635507 Chr.8: 131945017-131945017 on Build GRCh38
rs762250685 Chr.8: 131944742-131944742 on Build GRCh38
rs752342746 Chr.8: 131944765-131944765 on Build GRCh38
rs981157831 Chr.8: 131944947-131944947 on Build GRCh38
rs551567753 Chr.8: 131944898-131944898 on Build GRCh38
rs1032384200 Chr.8: 131944897-131944897 on Build GRCh38
rs369573495 Chr.8: 131944855-131944855 on Build GRCh38
rs1041069164 Chr.8: 131945125-131945125 on Build GRCh38
rs967220833 Chr.8: 131944942-131944942 on Build GRCh38
rs913076017 Chr.8: 131944925-131944926 on Build GRCh38
rs192357671 Chr.8: 131944776-131944776 on Build GRCh38
rs991560286 Chr.8: 131944993-131944993 on Build GRCh38
rs1055152274 Chr.8: 131945121-131945121 on Build GRCh38
rs931543403 Chr.8: 131945031-131945031 on Build GRCh38
rs928375045 Chr.8: 131944954-131944954 on Build GRCh38
rs75973656 Chr.8: 131944706-131944706 on Build GRCh38
rs766500421 Chr.8: 131944796-131944796 on Build GRCh38

View Full Product Details