Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 124451059-124451567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRD2
Entrez Gene ID
347088
Gene Name
adhesion G protein-coupled receptor D2
Gene Aliases
GPR144, PGR24
UniGene
Hs.454099
Gene Chromosome Location
Chr.9: 124451087-124477795 on Build GRCh38
Transcripts
XM_006717348.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5531853 c.25G>A p.E9K Chr.9: 124451168-124451168 on Build GRCh38
5665419 c.40G>C p.G14R Chr.9: 124451183-124451183 on Build GRCh38
6502627 c.52G>C p.D18H Chr.9: 124451195-124451195 on Build GRCh38
6018994 c.33G>T p.W11C Chr.9: 124451176-124451176 on Build GRCh38
4638058 c.19G>A p.A7T Chr.9: 124451162-124451162 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs929013597 Chr.9: 124451302-124451302 on Build GRCh38
rs954496350 Chr.9: 124451451-124451451 on Build GRCh38
rs761501662 Chr.9: 124451220-124451220 on Build GRCh38
rs543313198 Chr.9: 124451217-124451217 on Build GRCh38
rs535019271 Chr.9: 124451510-124451510 on Build GRCh38
rs751717443 Chr.9: 124451409-124451409 on Build GRCh38
rs888531877 Chr.9: 124451132-124451132 on Build GRCh38
rs541374306 Chr.9: 124451324-124451324 on Build GRCh38
rs145769710 Chr.9: 124451444-124451444 on Build GRCh38
rs760414006 Chr.9: 124451162-124451162 on Build GRCh38
rs757013569 Chr.9: 124451168-124451169 on Build GRCh38
rs941529105 Chr.9: 124451376-124451376 on Build GRCh38
rs749898254 Chr.9: 124451239-124451239 on Build GRCh38
rs991799882 Chr.9: 124451297-124451297 on Build GRCh38
rs766517300 Chr.9: 124451386-124451386 on Build GRCh38
rs142771751 Chr.9: 124451452-124451452 on Build GRCh38
rs779271191 Chr.9: 124451096-124451096 on Build GRCh38
rs113796531 Chr.9: 124451398-124451398 on Build GRCh38
rs374828078 Chr.9: 124451293-124451293 on Build GRCh38
rs138607585 Chr.9: 124451445-124451445 on Build GRCh38
rs191758920 Chr.9: 124451485-124451485 on Build GRCh38
rs375471631 Chr.9: 124451224-124451224 on Build GRCh38
rs372188859 Chr.9: 124451177-124451177 on Build GRCh38
rs988317670 Chr.9: 124451194-124451194 on Build GRCh38
rs1035869286 Chr.9: 124451368-124451368 on Build GRCh38
rs1022501312 Chr.9: 124451216-124451216 on Build GRCh38
rs114739506 Chr.9: 124451095-124451095 on Build GRCh38
rs746594149 Chr.9: 124451323-124451323 on Build GRCh38
rs76928670 Chr.9: 124451166-124451166 on Build GRCh38
rs987306557 Chr.9: 124451464-124451464 on Build GRCh38
rs750976249 Chr.9: 124451335-124451335 on Build GRCh38
rs780918503 Chr.9: 124451466-124451466 on Build GRCh38
rs991513393 Chr.9: 124451112-124451112 on Build GRCh38
rs556332630 Chr.9: 124451180-124451180 on Build GRCh38
rs187641798 Chr.9: 124451379-124451379 on Build GRCh38
rs775408680 Chr.9: 124451149-124451149 on Build GRCh38
rs1033531239 Chr.9: 124451524-124451524 on Build GRCh38
rs914599892 Chr.9: 124451135-124451135 on Build GRCh38
rs191713904 Chr.9: 124451115-124451115 on Build GRCh38
rs926515373 Chr.9: 124451247-124451247 on Build GRCh38
rs866735666 Chr.9: 124451167-124451168 on Build GRCh38
rs1047490082 Chr.9: 124451277-124451277 on Build GRCh38
rs1047619282 Chr.9: 124451121-124451121 on Build GRCh38
rs182946994 Chr.9: 124451246-124451246 on Build GRCh38
rs958987840 Chr.9: 124451378-124451378 on Build GRCh38
rs548909829 Chr.9: 124451535-124451535 on Build GRCh38
rs897680462 Chr.9: 124451105-124451105 on Build GRCh38
rs908994178 Chr.9: 124451341-124451341 on Build GRCh38
rs1051676261 Chr.9: 124451183-124451183 on Build GRCh38
rs980481835 Chr.9: 124451232-124451232 on Build GRCh38
rs147377334 Chr.9: 124451494-124451494 on Build GRCh38
rs557215129 Chr.9: 124451270-124451270 on Build GRCh38
rs920253689 Chr.9: 124451511-124451511 on Build GRCh38
rs73666890 Chr.9: 124451298-124451298 on Build GRCh38
rs942992045 Chr.9: 124451226-124451226 on Build GRCh38
rs776424959 Chr.9: 124451191-124451191 on Build GRCh38
rs960585017 Chr.9: 124451536-124451536 on Build GRCh38

View Full Product Details