Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 18163146-18163675 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEND2
Entrez Gene ID
139105
Gene Name
BEN domain containing 2
Gene Aliases
CXorf20
UniGene
Hs.403802
Gene Chromosome Location
Chr.X: 18162931-18220904 on Build GRCh38
Transcripts
NM_153346.4

SNP Detail

SNP ID Chromosome Location
rs1016426994 Chr.X: 18163501-18163501 on Build GRCh38
rs992870193 Chr.X: 18163583-18163583 on Build GRCh38
rs900863459 Chr.X: 18163333-18163333 on Build GRCh38
rs769031888 Chr.X: 18163567-18163567 on Build GRCh38
rs904431944 Chr.X: 18163297-18163297 on Build GRCh38
rs905091739 Chr.X: 18163613-18163613 on Build GRCh38
rs764306290 Chr.X: 18163394-18163394 on Build GRCh38
rs894287139 Chr.X: 18163279-18163279 on Build GRCh38
rs763098891 Chr.X: 18163173-18163173 on Build GRCh38
rs919276376 Chr.X: 18163211-18163211 on Build GRCh38
rs191708133 Chr.X: 18163309-18163309 on Build GRCh38
rs893629511 Chr.X: 18163559-18163559 on Build GRCh38
rs972756377 Chr.X: 18163525-18163525 on Build GRCh38
rs983659355 Chr.X: 18163189-18163189 on Build GRCh38
rs938769392 Chr.X: 18163233-18163233 on Build GRCh38
rs972089274 Chr.X: 18163180-18163180 on Build GRCh38
rs1031316513 Chr.X: 18163310-18163310 on Build GRCh38
rs182994225 Chr.X: 18163316-18163316 on Build GRCh38
rs953512988 Chr.X: 18163433-18163433 on Build GRCh38
rs986254992 Chr.X: 18163438-18163438 on Build GRCh38
rs187344824 Chr.X: 18163194-18163194 on Build GRCh38
rs1028032227 Chr.X: 18163395-18163395 on Build GRCh38
rs1057466596 Chr.X: 18163267-18163267 on Build GRCh38
rs931643890 Chr.X: 18163383-18163383 on Build GRCh38
rs963646411 Chr.X: 18163519-18163519 on Build GRCh38
rs5955941 Chr.X: 18163179-18163179 on Build GRCh38
rs183017800 Chr.X: 18163190-18163190 on Build GRCh38
rs558580853 Chr.X: 18163601-18163601 on Build GRCh38
rs867913884 Chr.X: 18163427-18163427 on Build GRCh38
rs1001506751 Chr.X: 18163616-18163616 on Build GRCh38
rs1042959749 Chr.X: 18163296-18163296 on Build GRCh38
rs767673099 Chr.X: 18163302-18163302 on Build GRCh38
rs1052312107 Chr.X: 18163520-18163520 on Build GRCh38
rs61256762 Chr.X: 18163309-18163309 on Build GRCh38
rs919238185 Chr.X: 18163534-18163534 on Build GRCh38
rs948536042 Chr.X: 18163294-18163294 on Build GRCh38
rs997916352 Chr.X: 18163344-18163344 on Build GRCh38
rs916003675 Chr.X: 18163642-18163642 on Build GRCh38
rs963694775 Chr.X: 18163287-18163287 on Build GRCh38
rs973648551 Chr.X: 18163301-18163302 on Build GRCh38

View Full Product Details