Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 46573913-46574414 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHST7
Entrez Gene ID
56548
Gene Name
carbohydrate sulfotransferase 7
Gene Aliases
C6ST-2, GST-5
UniGene
Hs.129955
Gene Chromosome Location
Chr.X: 46573687-46598496 on Build GRCh38
Transcripts
NM_019886.3

SNP Detail

SNP ID Chromosome Location
rs1048253650 Chr.X: 46574020-46574020 on Build GRCh38
rs777553106 Chr.X: 46574369-46574369 on Build GRCh38
rs912192592 Chr.X: 46573989-46573989 on Build GRCh38
rs931122047 Chr.X: 46574013-46574013 on Build GRCh38
rs777555910 Chr.X: 46574161-46574161 on Build GRCh38
rs1057072866 Chr.X: 46574121-46574121 on Build GRCh38
rs993207253 Chr.X: 46574015-46574015 on Build GRCh38
rs754570107 Chr.X: 46573961-46573961 on Build GRCh38
rs749031627 Chr.X: 46574372-46574372 on Build GRCh38
rs773167607 Chr.X: 46574057-46574057 on Build GRCh38
rs376196221 Chr.X: 46574387-46574387 on Build GRCh38
rs762118180 Chr.X: 46574246-46574246 on Build GRCh38
rs770884974 Chr.X: 46574217-46574217 on Build GRCh38
rs1006895013 Chr.X: 46574179-46574179 on Build GRCh38
rs778401924 Chr.X: 46573991-46573991 on Build GRCh38
rs775250094 Chr.X: 46574244-46574244 on Build GRCh38
rs753313217 Chr.X: 46573957-46573957 on Build GRCh38
rs202021438 Chr.X: 46574222-46574222 on Build GRCh38
rs61575462 Chr.X: 46573994-46573994 on Build GRCh38
rs773884876 Chr.X: 46574091-46574091 on Build GRCh38
rs771837285 Chr.X: 46574042-46574042 on Build GRCh38
rs763455298 Chr.X: 46574276-46574276 on Build GRCh38
rs369680575 Chr.X: 46574144-46574144 on Build GRCh38
rs751788961 Chr.X: 46574188-46574188 on Build GRCh38
rs762398181 Chr.X: 46574058-46574058 on Build GRCh38
rs754310332 Chr.X: 46574134-46574134 on Build GRCh38
rs747935476 Chr.X: 46574035-46574035 on Build GRCh38
rs770347970 Chr.X: 46574221-46574221 on Build GRCh38
rs755895426 Chr.X: 46574213-46574213 on Build GRCh38
rs770422083 Chr.X: 46574068-46574068 on Build GRCh38
rs200081268 Chr.X: 46574310-46574310 on Build GRCh38
rs750516329 Chr.X: 46573983-46573983 on Build GRCh38
rs766424462 Chr.X: 46574182-46574182 on Build GRCh38
rs895785535 Chr.X: 46574140-46574140 on Build GRCh38
rs747186047 Chr.X: 46574001-46574001 on Build GRCh38
rs921040844 Chr.X: 46574007-46574007 on Build GRCh38
rs750903362 Chr.X: 46574160-46574160 on Build GRCh38
rs751698930 Chr.X: 46574315-46574315 on Build GRCh38
rs765590326 Chr.X: 46574273-46574273 on Build GRCh38
rs781382088 Chr.X: 46574210-46574210 on Build GRCh38
rs374103732 Chr.X: 46574220-46574220 on Build GRCh38
rs752864505 Chr.X: 46574350-46574350 on Build GRCh38
rs756606455 Chr.X: 46574232-46574232 on Build GRCh38
rs755870456 Chr.X: 46574365-46574365 on Build GRCh38
rs373483256 Chr.X: 46574319-46574319 on Build GRCh38
rs759196964 Chr.X: 46574117-46574117 on Build GRCh38
rs767273988 Chr.X: 46574120-46574120 on Build GRCh38
rs765705791 Chr.X: 46574150-46574150 on Build GRCh38
rs773605240 Chr.X: 46574274-46574274 on Build GRCh38
rs769680098 Chr.X: 46574055-46574055 on Build GRCh38
rs771898341 Chr.X: 46574231-46574231 on Build GRCh38
rs956147595 Chr.X: 46574340-46574340 on Build GRCh38
rs200180964 Chr.X: 46574201-46574201 on Build GRCh38
rs377001713 Chr.X: 46574229-46574229 on Build GRCh38
rs1023191728 Chr.X: 46574211-46574211 on Build GRCh38
rs768457634 Chr.X: 46574014-46574014 on Build GRCh38
rs761449271 Chr.X: 46574335-46574335 on Build GRCh38
rs781021631 Chr.X: 46574109-46574128 on Build GRCh38
rs904627226 Chr.X: 46574181-46574181 on Build GRCh38
rs780902051 Chr.X: 46574032-46574032 on Build GRCh38
rs1031871173 Chr.X: 46574207-46574207 on Build GRCh38

View Full Product Details