Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101623697-101624202 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMCX3
Entrez Gene ID
51566
Gene Name
armadillo repeat containing, X-linked 3
Gene Aliases
ALEX3, GASP6, dJ545K15.2
UniGene
Hs.592225
Gene Chromosome Location
Chr.X: 101623127-101631910 on Build GRCh38
Transcripts
NM_016607.3 NM_177947.2 NM_177948.2 XM_005262141.2

SNP Detail

SNP ID Chromosome Location
rs936309090 Chr.X: 101623734-101623734 on Build GRCh38
rs781806986 Chr.X: 101623936-101623936 on Build GRCh38
rs55791402 Chr.X: 101623938-101623938 on Build GRCh38
rs1034329665 Chr.X: 101624180-101624180 on Build GRCh38
rs933047327 Chr.X: 101623898-101623898 on Build GRCh38
rs782561900 Chr.X: 101623748-101623748 on Build GRCh38
rs782400305 Chr.X: 101624110-101624110 on Build GRCh38
rs1038669759 Chr.X: 101623865-101623865 on Build GRCh38
rs900186152 Chr.X: 101623868-101623868 on Build GRCh38
rs782668939 Chr.X: 101623935-101623935 on Build GRCh38
rs189332677 Chr.X: 101624029-101624029 on Build GRCh38
rs1016115364 Chr.X: 101624061-101624061 on Build GRCh38
rs782682855 Chr.X: 101624117-101624117 on Build GRCh38
rs782564475 Chr.X: 101623744-101623744 on Build GRCh38
rs56035524 Chr.X: 101623927-101623927 on Build GRCh38
rs1001870399 Chr.X: 101624131-101624131 on Build GRCh38
rs782253775 Chr.X: 101624109-101624109 on Build GRCh38
rs1051960450 Chr.X: 101623944-101623944 on Build GRCh38
rs782621248 Chr.X: 101624047-101624047 on Build GRCh38

View Full Product Details