Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 16834928-16835438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUBN
Entrez Gene ID
8029
Gene Name
cubilin
Gene Aliases
IFCR, MGA1, gp280
UniGene
Hs.166206
Gene Chromosome Location
Chr.10: 16823966-17130492 on Build GRCh38
Transcripts
NM_001081.3 XM_011519709.2 XM_011519710.2 XM_011519711.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1236001 c.10238A>G p.D3413G Chr.10: 16835138-16835138 on Build GRCh38
1702097 c.10351G>A p.D3451N Chr.10: 16835025-16835025 on Build GRCh38
5998840 c.10192G>T p.D3398Y Chr.10: 16835184-16835184 on Build GRCh38
916763 c.10362+2T>C p.? Chr.10: 16835012-16835012 on Build GRCh38
1702098 c.10301C>T p.S3434F Chr.10: 16835075-16835075 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145647391 Chr.10: 16835126-16835126 on Build GRCh38
rs566428134 Chr.10: 16835089-16835089 on Build GRCh38
rs752570411 Chr.10: 16835164-16835164 on Build GRCh38
rs566692005 Chr.10: 16835340-16835340 on Build GRCh38
rs703061 Chr.10: 16835392-16835392 on Build GRCh38
rs7898873 Chr.10: 16835081-16835081 on Build GRCh38
rs117311278 Chr.10: 16835225-16835225 on Build GRCh38
rs761048540 Chr.10: 16835220-16835220 on Build GRCh38
rs141890166 Chr.10: 16835120-16835120 on Build GRCh38
rs780850360 Chr.10: 16835189-16835189 on Build GRCh38
rs142537000 Chr.10: 16835227-16835227 on Build GRCh38
rs749126611 Chr.10: 16835020-16835020 on Build GRCh38
rs760807440 Chr.10: 16835138-16835138 on Build GRCh38
rs777539576 Chr.10: 16835177-16835177 on Build GRCh38
rs143066121 Chr.10: 16835099-16835099 on Build GRCh38
rs542073853 Chr.10: 16835182-16835182 on Build GRCh38
rs577492421 Chr.10: 16835090-16835090 on Build GRCh38
rs776263089 Chr.10: 16835049-16835049 on Build GRCh38
rs755380998 Chr.10: 16835331-16835331 on Build GRCh38
rs778538546 Chr.10: 16835026-16835026 on Build GRCh38
rs377579746 Chr.10: 16835213-16835213 on Build GRCh38
rs574703032 Chr.10: 16835180-16835180 on Build GRCh38
rs556303931 Chr.10: 16835141-16835141 on Build GRCh38
rs1801230 Chr.10: 16835111-16835111 on Build GRCh38
rs372592269 Chr.10: 16835095-16835095 on Build GRCh38
rs760969787 Chr.10: 16835091-16835091 on Build GRCh38
rs117570588 Chr.10: 16835094-16835094 on Build GRCh38
rs1034347553 Chr.10: 16835000-16835000 on Build GRCh38
rs769284377 Chr.10: 16835195-16835195 on Build GRCh38
rs772809239 Chr.10: 16835037-16835037 on Build GRCh38
rs780537200 Chr.10: 16835118-16835118 on Build GRCh38
rs1014926675 Chr.10: 16835282-16835282 on Build GRCh38
rs746666327 Chr.10: 16835047-16835047 on Build GRCh38
rs148314512 Chr.10: 16835276-16835276 on Build GRCh38
rs749145773 Chr.10: 16835080-16835080 on Build GRCh38
rs182428546 Chr.10: 16835323-16835323 on Build GRCh38
rs780190067 Chr.10: 16835008-16835008 on Build GRCh38
rs752029852 Chr.10: 16834985-16834985 on Build GRCh38
rs562169898 Chr.10: 16835403-16835403 on Build GRCh38
rs753236784 Chr.10: 16835416-16835416 on Build GRCh38
rs750697916 Chr.10: 16834994-16834994 on Build GRCh38
rs759618283 Chr.10: 16835150-16835150 on Build GRCh38
rs373977491 Chr.10: 16835161-16835161 on Build GRCh38
rs190546478 Chr.10: 16835211-16835211 on Build GRCh38
rs957796034 Chr.10: 16835354-16835354 on Build GRCh38
rs758121501 Chr.10: 16835108-16835108 on Build GRCh38
rs754704005 Chr.10: 16835090-16835091 on Build GRCh38
rs762417121 Chr.10: 16835127-16835127 on Build GRCh38
rs1056224479 Chr.10: 16835258-16835258 on Build GRCh38
rs745539078 Chr.10: 16835194-16835194 on Build GRCh38
rs747982383 Chr.10: 16835027-16835027 on Build GRCh38
rs111573959 Chr.10: 16835229-16835229 on Build GRCh38
rs533766298 Chr.10: 16835345-16835345 on Build GRCh38
rs145661638 Chr.10: 16835025-16835025 on Build GRCh38
rs145872906 Chr.10: 16835109-16835109 on Build GRCh38
rs377414794 Chr.10: 16835154-16835154 on Build GRCh38
rs754845243 Chr.10: 16835096-16835096 on Build GRCh38
rs144484373 Chr.10: 16835143-16835143 on Build GRCh38
rs1020507679 Chr.10: 16835284-16835284 on Build GRCh38
rs752660097 Chr.10: 16835105-16835105 on Build GRCh38
rs770286772 Chr.10: 16835048-16835048 on Build GRCh38
rs745433438 Chr.10: 16835124-16835124 on Build GRCh38
rs763365999 Chr.10: 16835069-16835069 on Build GRCh38
rs1018371339 Chr.10: 16835384-16835384 on Build GRCh38
rs758386041 Chr.10: 16835171-16835171 on Build GRCh38
rs370985411 Chr.10: 16835196-16835196 on Build GRCh38
rs773774080 Chr.10: 16835137-16835137 on Build GRCh38
rs186426453 Chr.10: 16834992-16834992 on Build GRCh38
rs769211365 Chr.10: 16835125-16835125 on Build GRCh38
rs547284067 Chr.10: 16834977-16834977 on Build GRCh38
rs778111776 Chr.10: 16835248-16835248 on Build GRCh38
rs937430832 Chr.10: 16835230-16835230 on Build GRCh38
rs886046864 Chr.10: 16835209-16835209 on Build GRCh38
rs1006616618 Chr.10: 16835372-16835372 on Build GRCh38
rs771874206 Chr.10: 16835034-16835034 on Build GRCh38
rs762846233 Chr.10: 16835243-16835243 on Build GRCh38
rs115686202 Chr.10: 16835226-16835226 on Build GRCh38
rs766669502 Chr.10: 16835139-16835139 on Build GRCh38
rs45520337 Chr.10: 16835322-16835322 on Build GRCh38
rs61844025 Chr.10: 16835353-16835353 on Build GRCh38
rs147730705 Chr.10: 16835131-16835131 on Build GRCh38
rs369136480 Chr.10: 16834969-16834969 on Build GRCh38
rs370911251 Chr.10: 16834993-16834993 on Build GRCh38
rs943972833 Chr.10: 16835178-16835178 on Build GRCh38
rs977078064 Chr.10: 16835393-16835393 on Build GRCh38
rs776574207 Chr.10: 16835007-16835007 on Build GRCh38
rs746649985 Chr.10: 16835110-16835110 on Build GRCh38

View Full Product Details