Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 27012466-27013010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD26
Entrez Gene ID
22852
Gene Name
ankyrin repeat domain 26
Gene Aliases
THC2, bA145E8.1
UniGene
Hs.361041
Gene Chromosome Location
Chr.10: 26938195-27100498 on Build GRCh38
Transcripts
NM_001256053.1 NM_014915.2 XM_006717423.2 XM_006717425.3 XM_011519416.2 XM_017015928.1 XM_017015929.1 XM_017015930.1 XM_017015931.1 XM_017015932.1 XM_017015933.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7349393 c.4907A>C p.D1636A Chr.10: 27012976-27012976 on Build GRCh38
6600506 c.4965G>A p.R1655R Chr.10: 27012918-27012918 on Build GRCh38
6569484 c.314delA p.N105fs*3 Chr.10: 27012983-27012983 on Build GRCh38
7349395 c.321A>C p.L107F Chr.10: 27012976-27012976 on Build GRCh38
5209722 c.4924A>G p.I1642V Chr.10: 27012959-27012959 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021398369 Chr.10: 27012547-27012547 on Build GRCh38
rs765009874 Chr.10: 27012984-27012984 on Build GRCh38
rs750690783 Chr.10: 27012550-27012550 on Build GRCh38
rs769629179 Chr.10: 27012977-27012977 on Build GRCh38
rs775782993 Chr.10: 27012942-27012942 on Build GRCh38
rs1044944425 Chr.10: 27012717-27012717 on Build GRCh38
rs780083874 Chr.10: 27012882-27012882 on Build GRCh38
rs563039964 Chr.10: 27012655-27012655 on Build GRCh38
rs926308913 Chr.10: 27012969-27012969 on Build GRCh38
rs771318907 Chr.10: 27012931-27012931 on Build GRCh38
rs763617426 Chr.10: 27012855-27012855 on Build GRCh38
rs755065312 Chr.10: 27012894-27012894 on Build GRCh38
rs920703334 Chr.10: 27012711-27012711 on Build GRCh38
rs564602409 Chr.10: 27012567-27012567 on Build GRCh38
rs891459230 Chr.10: 27012539-27012539 on Build GRCh38
rs531966650 Chr.10: 27012664-27012664 on Build GRCh38
rs570760382 Chr.10: 27012810-27012810 on Build GRCh38
rs756909530 Chr.10: 27012873-27012873 on Build GRCh38
rs745539207 Chr.10: 27012933-27012933 on Build GRCh38
rs892692254 Chr.10: 27012840-27012840 on Build GRCh38
rs753449722 Chr.10: 27012869-27012869 on Build GRCh38
rs572143624 Chr.10: 27012956-27012956 on Build GRCh38
rs1028820059 Chr.10: 27012573-27012573 on Build GRCh38
rs192307552 Chr.10: 27012917-27012917 on Build GRCh38
rs756778765 Chr.10: 27012926-27012926 on Build GRCh38
rs770197413 Chr.10: 27012915-27012915 on Build GRCh38
rs980940005 Chr.10: 27012748-27012748 on Build GRCh38
rs781421980 Chr.10: 27012898-27012898 on Build GRCh38
rs747042493 Chr.10: 27012891-27012891 on Build GRCh38
rs1009882823 Chr.10: 27012542-27012542 on Build GRCh38
rs556054076 Chr.10: 27012819-27012819 on Build GRCh38
rs568576373 Chr.10: 27012712-27012712 on Build GRCh38
rs368545607 Chr.10: 27012879-27012879 on Build GRCh38
rs374752369 Chr.10: 27012859-27012859 on Build GRCh38
rs764884515 Chr.10: 27012875-27012875 on Build GRCh38
rs758267677 Chr.10: 27012881-27012881 on Build GRCh38
rs548680982 Chr.10: 27012704-27012704 on Build GRCh38
rs776133728 Chr.10: 27012964-27012964 on Build GRCh38
rs374056886 Chr.10: 27012572-27012572 on Build GRCh38
rs752424161 Chr.10: 27012761-27012761 on Build GRCh38
rs954533068 Chr.10: 27012609-27012609 on Build GRCh38
rs987796385 Chr.10: 27012626-27012626 on Build GRCh38
rs1056201240 Chr.10: 27012775-27012775 on Build GRCh38
rs915022204 Chr.10: 27012934-27012934 on Build GRCh38
rs539678467 Chr.10: 27012818-27012818 on Build GRCh38
rs758485048 Chr.10: 27012743-27012743 on Build GRCh38
rs973364433 Chr.10: 27012695-27012695 on Build GRCh38
rs199806046 Chr.10: 27012980-27012980 on Build GRCh38
rs533441985 Chr.10: 27012754-27012754 on Build GRCh38
rs912083737 Chr.10: 27012585-27012585 on Build GRCh38
rs899158751 Chr.10: 27012820-27012820 on Build GRCh38
rs939283009 Chr.10: 27012752-27012752 on Build GRCh38
rs547817560 Chr.10: 27012807-27012807 on Build GRCh38
rs774359126 Chr.10: 27012842-27012842 on Build GRCh38
rs765360047 Chr.10: 27012544-27012544 on Build GRCh38
rs184369322 Chr.10: 27012919-27012919 on Build GRCh38
rs372371136 Chr.10: 27012959-27012959 on Build GRCh38
rs11015453 Chr.10: 27012513-27012513 on Build GRCh38
rs1005340156 Chr.10: 27012795-27012795 on Build GRCh38
rs760226729 Chr.10: 27012852-27012852 on Build GRCh38
rs201953836 Chr.10: 27012889-27012889 on Build GRCh38
rs780649837 Chr.10: 27012780-27012780 on Build GRCh38
rs533931215 Chr.10: 27012765-27012765 on Build GRCh38
rs199699340 Chr.10: 27012911-27012911 on Build GRCh38
rs368507197 Chr.10: 27012920-27012920 on Build GRCh38
rs1053137311 Chr.10: 27012839-27012839 on Build GRCh38

View Full Product Details