Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73253936-73254440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP70
Entrez Gene ID
118491
Gene Name
cilia and flagella associated protein 70
Gene Aliases
TTC18
UniGene
Hs.591367
Gene Chromosome Location
Chr.10: 73253758-73361064 on Build GRCh38
Transcripts
NM_145170.3 XM_005269489.2 XM_006717604.2 XM_006717605.2 XM_006717609.2 XM_006717610.2 XM_006717611.3 XM_011539212.2 XM_017015620.1 XM_017015621.1 XM_017015622.1 XM_017015623.1 XM_017015624.1 XM_017015625.1 XM_017015626.1 XM_017015627.1 XM_017015628.1 XM_017015629.1 XM_017015630.1 XM_017015631.1 XM_017015632.1 NR_038373.1 NR_134458.1 NR_134459.1 NR_134460.1

Gene Symbol
DNAJC9-AS1
Entrez Gene ID
414245
Gene Name
DNAJC9 antisense RNA 1
Gene Aliases
C10orf103, bA537A6.3
UniGene
Hs.568860
Gene Chromosome Location
Chr.10: 73247367-73276984 on Build GRCh38
Transcripts
NM_145170.3 XM_005269489.2 XM_006717604.2 XM_006717605.2 XM_006717609.2 XM_006717610.2 XM_006717611.3 XM_011539212.2 XM_017015620.1 XM_017015621.1 XM_017015622.1 XM_017015623.1 XM_017015624.1 XM_017015625.1 XM_017015626.1 XM_017015627.1 XM_017015628.1 XM_017015629.1 XM_017015630.1 XM_017015631.1 XM_017015632.1 NR_038373.1 NR_134458.1 NR_134459.1 NR_134460.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4923211 c.3341T>C p.V1114A Chr.10: 73254000-73254000 on Build GRCh38
685389 c.3356C>T p.P1119L Chr.10: 73253985-73253985 on Build GRCh38
6194118 c.3303G>T p.E1101D Chr.10: 73254038-73254038 on Build GRCh38
1297414 c.3359C>G p.S1120C Chr.10: 73253982-73253982 on Build GRCh38
7162181 c.3288G>T p.L1096L Chr.10: 73254053-73254053 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772688587 Chr.10: 73254101-73254102 on Build GRCh38
rs944671135 Chr.10: 73254372-73254372 on Build GRCh38
rs1132907 Chr.10: 73254284-73254284 on Build GRCh38
rs979004743 Chr.10: 73254175-73254175 on Build GRCh38
rs769547174 Chr.10: 73254020-73254020 on Build GRCh38
rs920723914 Chr.10: 73254332-73254332 on Build GRCh38
rs530006768 Chr.10: 73254165-73254165 on Build GRCh38
rs3830554 Chr.10: 73254127-73254128 on Build GRCh38
rs142119729 Chr.10: 73254066-73254066 on Build GRCh38
rs899078038 Chr.10: 73254407-73254407 on Build GRCh38
rs762304745 Chr.10: 73254086-73254086 on Build GRCh38
rs1028191289 Chr.10: 73254266-73254266 on Build GRCh38
rs758044655 Chr.10: 73253978-73253978 on Build GRCh38
rs1044610517 Chr.10: 73254125-73254125 on Build GRCh38
rs776887790 Chr.10: 73254072-73254072 on Build GRCh38
rs765890679 Chr.10: 73253972-73253972 on Build GRCh38
rs1046678293 Chr.10: 73254308-73254308 on Build GRCh38
rs765688676 Chr.10: 73254095-73254095 on Build GRCh38
rs992532386 Chr.10: 73254310-73254310 on Build GRCh38
rs778776847 Chr.10: 73253970-73253970 on Build GRCh38
rs866972075 Chr.10: 73254356-73254356 on Build GRCh38
rs113752339 Chr.10: 73254079-73254079 on Build GRCh38
rs371914735 Chr.10: 73254042-73254042 on Build GRCh38
rs772156332 Chr.10: 73254049-73254049 on Build GRCh38
rs564953754 Chr.10: 73253986-73253986 on Build GRCh38
rs999861548 Chr.10: 73254283-73254283 on Build GRCh38
rs60209004 Chr.10: 73254126-73254127 on Build GRCh38
rs539894199 Chr.10: 73253982-73253982 on Build GRCh38
rs754079843 Chr.10: 73254409-73254409 on Build GRCh38
rs886378050 Chr.10: 73254061-73254061 on Build GRCh38
rs767482369 Chr.10: 73254069-73254071 on Build GRCh38
rs1037582262 Chr.10: 73254379-73254379 on Build GRCh38
rs968529323 Chr.10: 73254269-73254269 on Build GRCh38
rs767275806 Chr.10: 73254319-73254319 on Build GRCh38
rs950145415 Chr.10: 73254293-73254293 on Build GRCh38
rs773913116 Chr.10: 73254096-73254096 on Build GRCh38
rs974491483 Chr.10: 73254131-73254131 on Build GRCh38
rs879842721 Chr.10: 73254268-73254268 on Build GRCh38
rs930817869 Chr.10: 73254337-73254337 on Build GRCh38
rs907987818 Chr.10: 73254366-73254366 on Build GRCh38
rs750926559 Chr.10: 73253976-73253976 on Build GRCh38
rs779021509 Chr.10: 73253974-73253974 on Build GRCh38
rs574249327 Chr.10: 73254143-73254143 on Build GRCh38
rs1055715559 Chr.10: 73254279-73254279 on Build GRCh38
rs777754433 Chr.10: 73254032-73254032 on Build GRCh38
rs917489955 Chr.10: 73254313-73254313 on Build GRCh38
rs1013871440 Chr.10: 73254018-73254018 on Build GRCh38
rs797020006 Chr.10: 73254128-73254129 on Build GRCh38
rs145830698 Chr.10: 73254133-73254133 on Build GRCh38
rs576935090 Chr.10: 73254062-73254062 on Build GRCh38
rs1005214229 Chr.10: 73254344-73254344 on Build GRCh38
rs937382695 Chr.10: 73254226-73254226 on Build GRCh38
rs747986672 Chr.10: 73254002-73254002 on Build GRCh38
rs775368453 Chr.10: 73254059-73254059 on Build GRCh38
rs760130493 Chr.10: 73254104-73254105 on Build GRCh38
rs561222108 Chr.10: 73254352-73254352 on Build GRCh38
rs781065003 Chr.10: 73254000-73254000 on Build GRCh38
rs369369028 Chr.10: 73254358-73254358 on Build GRCh38
rs560852529 Chr.10: 73254328-73254328 on Build GRCh38
rs1008916963 Chr.10: 73254210-73254210 on Build GRCh38
rs913378763 Chr.10: 73254160-73254160 on Build GRCh38
rs925959681 Chr.10: 73254212-73254212 on Build GRCh38
rs1041347533 Chr.10: 73254223-73254223 on Build GRCh38
rs955674531 Chr.10: 73254300-73254300 on Build GRCh38
rs1028783925 Chr.10: 73254132-73254132 on Build GRCh38
rs963004288 Chr.10: 73254116-73254116 on Build GRCh38
rs146234051 Chr.10: 73253981-73253981 on Build GRCh38
rs901113039 Chr.10: 73254227-73254227 on Build GRCh38
rs997080891 Chr.10: 73254242-73254242 on Build GRCh38
rs375715915 Chr.10: 73254102-73254102 on Build GRCh38

View Full Product Details