Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 76875373-76875894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNMA1
Entrez Gene ID
3778
Gene Name
potassium calcium-activated channel subfamily M alpha 1
Gene Aliases
BKTM, KCa1.1, MaxiK, SAKCA, SLO, SLO-ALPHA, SLO1, bA205K10.1, hSlo, mSLO1
UniGene
Hs.144795
Gene Chromosome Location
Chr.10: 76869601-77637819 on Build GRCh38
Transcripts
NM_001014797.2 NM_001271518.1 NM_001322829.1 NM_001322830.1 NM_001322832.1 NM_001322835.1 NM_001322838.1 XM_005269776.3 XM_005269778.1 XM_005269781.2 XM_005269787.3 XM_005269789.2 XM_005269792.2 XM_005269796.2 XM_011539773.1 XM_011539775.1 XM_011539776.1 XM_011539780.1 XM_011539781.2 XM_011539785.1 XM_017016207.1 XM_017016208.1 XM_017016210.1 XM_017016211.1 XM_017016212.1 XM_017016213.1 XM_017016216.1 XM_017016217.1 XM_017016223.1

SNP Detail

SNP ID Chromosome Location
rs11001915 Chr.10: 76875733-76875733 on Build GRCh38
rs1055968136 Chr.10: 76875445-76875445 on Build GRCh38
rs566576268 Chr.10: 76875626-76875626 on Build GRCh38
rs11398145 Chr.10: 76875753-76875754 on Build GRCh38
rs181681812 Chr.10: 76875590-76875590 on Build GRCh38
rs190220738 Chr.10: 76875796-76875796 on Build GRCh38
rs562631346 Chr.10: 76875868-76875868 on Build GRCh38
rs963818765 Chr.10: 76875691-76875691 on Build GRCh38
rs894751219 Chr.10: 76875482-76875482 on Build GRCh38
rs956163542 Chr.10: 76875857-76875857 on Build GRCh38
rs947516622 Chr.10: 76875817-76875817 on Build GRCh38
rs903201378 Chr.10: 76875576-76875576 on Build GRCh38
rs938391921 Chr.10: 76875770-76875770 on Build GRCh38
rs182112758 Chr.10: 76875842-76875842 on Build GRCh38
rs978885646 Chr.10: 76875461-76875461 on Build GRCh38
rs1046815527 Chr.10: 76875416-76875416 on Build GRCh38
rs185412098 Chr.10: 76875786-76875786 on Build GRCh38
rs1053260075 Chr.10: 76875567-76875567 on Build GRCh38
rs928411082 Chr.10: 76875758-76875758 on Build GRCh38
rs1030423562 Chr.10: 76875620-76875620 on Build GRCh38
rs548509661 Chr.10: 76875421-76875421 on Build GRCh38
rs529694625 Chr.10: 76875538-76875538 on Build GRCh38
rs950337337 Chr.10: 76875595-76875595 on Build GRCh38
rs141667052 Chr.10: 76875672-76875672 on Build GRCh38
rs903398448 Chr.10: 76875837-76875837 on Build GRCh38
rs974037066 Chr.10: 76875699-76875699 on Build GRCh38
rs143856102 Chr.10: 76875420-76875421 on Build GRCh38
rs905551188 Chr.10: 76875646-76875646 on Build GRCh38
rs1018050179 Chr.10: 76875690-76875690 on Build GRCh38
rs577148641 Chr.10: 76875849-76875849 on Build GRCh38
rs186559818 Chr.10: 76875420-76875420 on Build GRCh38
rs919800832 Chr.10: 76875700-76875700 on Build GRCh38
rs563213441 Chr.10: 76875824-76875824 on Build GRCh38
rs954249142 Chr.10: 76875433-76875433 on Build GRCh38
rs965335789 Chr.10: 76875413-76875413 on Build GRCh38
rs1018883163 Chr.10: 76875412-76875413 on Build GRCh38
rs1008039526 Chr.10: 76875684-76875684 on Build GRCh38
rs758363456 Chr.10: 76875400-76875403 on Build GRCh38
rs7908954 Chr.10: 76875847-76875847 on Build GRCh38
rs150674677 Chr.10: 76875795-76875795 on Build GRCh38
rs999221142 Chr.10: 76875604-76875604 on Build GRCh38
rs1897594 Chr.10: 76875520-76875520 on Build GRCh38
rs145752207 Chr.10: 76875427-76875428 on Build GRCh38
rs774551718 Chr.10: 76875707-76875707 on Build GRCh38
rs886241499 Chr.10: 76875693-76875693 on Build GRCh38
rs1043670668 Chr.10: 76875558-76875558 on Build GRCh38
rs545938425 Chr.10: 76875852-76875852 on Build GRCh38
rs538771786 Chr.10: 76875654-76875654 on Build GRCh38
rs190236757 Chr.10: 76875444-76875444 on Build GRCh38

View Full Product Details