Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 102854131-102854633 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BORCS7
Entrez Gene ID
119032
Gene Name
BLOC-1 related complex subunit 7
Gene Aliases
C10orf32
UniGene
Hs.34492
Gene Chromosome Location
Chr.10: 102854210-102864961 on Build GRCh38
Transcripts
NM_001136200.1 NM_144591.3 NR_037644.1

Gene Symbol
BORCS7-ASMT
Entrez Gene ID
100528007
Gene Name
BORCS7-ASMT readthrough (NMD candidate)
Gene Aliases
C10orf32-AS3MT, C10orf32-ASMT
UniGene
Hs.720370
Gene Chromosome Location
Chr.10: 102854210-102901899 on Build GRCh38
Transcripts
NM_001136200.1 NM_144591.3 NR_037644.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6265383 c.117G>T p.V39V Chr.10: 102854403-102854403 on Build GRCh38
6366690 c.74T>C p.V25A Chr.10: 102854360-102854360 on Build GRCh38
3806364 c.51G>A p.V17V Chr.10: 102854337-102854337 on Build GRCh38
5646180 c.75G>A p.V25V Chr.10: 102854361-102854361 on Build GRCh38
5132653 c.103C>T p.L35F Chr.10: 102854389-102854389 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1009454988 Chr.10: 102854586-102854587 on Build GRCh38
rs568353653 Chr.10: 102854428-102854428 on Build GRCh38
rs535263629 Chr.10: 102854430-102854430 on Build GRCh38
rs780883999 Chr.10: 102854247-102854247 on Build GRCh38
rs753910123 Chr.10: 102854265-102854265 on Build GRCh38
rs777983313 Chr.10: 102854242-102854242 on Build GRCh38
rs746420401 Chr.10: 102854327-102854327 on Build GRCh38
rs190693137 Chr.10: 102854199-102854199 on Build GRCh38
rs899662368 Chr.10: 102854405-102854405 on Build GRCh38
rs767065954 Chr.10: 102854300-102854300 on Build GRCh38
rs753668246 Chr.10: 102854352-102854352 on Build GRCh38
rs1053596316 Chr.10: 102854555-102854555 on Build GRCh38
rs34951268 Chr.10: 102854222-102854223 on Build GRCh38
rs375123320 Chr.10: 102854447-102854447 on Build GRCh38
rs767070482 Chr.10: 102854366-102854366 on Build GRCh38
rs371203672 Chr.10: 102854284-102854284 on Build GRCh38
rs765943098 Chr.10: 102854351-102854351 on Build GRCh38
rs756062269 Chr.10: 102854304-102854304 on Build GRCh38
rs991700164 Chr.10: 102854329-102854329 on Build GRCh38
rs758625979 Chr.10: 102854320-102854320 on Build GRCh38
rs543408645 Chr.10: 102854544-102854544 on Build GRCh38
rs888911532 Chr.10: 102854448-102854448 on Build GRCh38
rs376197535 Chr.10: 102854249-102854249 on Build GRCh38
rs775998610 Chr.10: 102854335-102854335 on Build GRCh38
rs996756389 Chr.10: 102854223-102854223 on Build GRCh38
rs566816775 Chr.10: 102854303-102854303 on Build GRCh38
rs199826074 Chr.10: 102854357-102854357 on Build GRCh38
rs908191922 Chr.10: 102854485-102854485 on Build GRCh38
rs960107141 Chr.10: 102854527-102854527 on Build GRCh38
rs1047234485 Chr.10: 102854173-102854173 on Build GRCh38
rs1011604313 Chr.10: 102854419-102854419 on Build GRCh38
rs919618551 Chr.10: 102854337-102854337 on Build GRCh38
rs972487756 Chr.10: 102854298-102854298 on Build GRCh38
rs530184624 Chr.10: 102854276-102854276 on Build GRCh38
rs903181729 Chr.10: 102854600-102854600 on Build GRCh38
rs564047911 Chr.10: 102854197-102854197 on Build GRCh38
rs893731866 Chr.10: 102854568-102854568 on Build GRCh38
rs993495623 Chr.10: 102854484-102854484 on Build GRCh38
rs1002951538 Chr.10: 102854455-102854455 on Build GRCh38
rs1036308354 Chr.10: 102854471-102854471 on Build GRCh38
rs1040779397 Chr.10: 102854529-102854529 on Build GRCh38
rs778699886 Chr.10: 102854315-102854315 on Build GRCh38
rs3824754 Chr.10: 102854593-102854593 on Build GRCh38
rs749094031 Chr.10: 102854266-102854266 on Build GRCh38
rs763774245 Chr.10: 102854293-102854293 on Build GRCh38
rs1029036235 Chr.10: 102854229-102854229 on Build GRCh38
rs763714380 Chr.10: 102854171-102854171 on Build GRCh38
rs749714949 Chr.10: 102854450-102854450 on Build GRCh38
rs768900369 Chr.10: 102854271-102854271 on Build GRCh38
rs138404285 Chr.10: 102854424-102854424 on Build GRCh38
rs148731942 Chr.10: 102854490-102854490 on Build GRCh38
rs901588590 Chr.10: 102854416-102854416 on Build GRCh38
rs772699694 Chr.10: 102854478-102854478 on Build GRCh38
rs140388364 Chr.10: 102854403-102854403 on Build GRCh38
rs374746283 Chr.10: 102854474-102854474 on Build GRCh38
rs749889707 Chr.10: 102854381-102854381 on Build GRCh38
rs982023156 Chr.10: 102854279-102854279 on Build GRCh38
rs200327590 Chr.10: 102854308-102854308 on Build GRCh38
rs1036140184 Chr.10: 102854490-102854490 on Build GRCh38
rs201323817 Chr.10: 102854283-102854283 on Build GRCh38
rs1052552430 Chr.10: 102854415-102854415 on Build GRCh38
rs913326392 Chr.10: 102854185-102854210 on Build GRCh38
rs180715493 Chr.10: 102854201-102854201 on Build GRCh38
rs760142332 Chr.10: 102854345-102854345 on Build GRCh38
rs899752726 Chr.10: 102854209-102854209 on Build GRCh38
rs373428917 Chr.10: 102854473-102854473 on Build GRCh38
rs371914756 Chr.10: 102854434-102854434 on Build GRCh38
rs367688173 Chr.10: 102854339-102854339 on Build GRCh38
rs572188161 Chr.10: 102854165-102854165 on Build GRCh38
rs759643167 Chr.10: 102854281-102854282 on Build GRCh38
rs553957031 Chr.10: 102854433-102854433 on Build GRCh38
rs932487993 Chr.10: 102854553-102854553 on Build GRCh38
rs757300174 Chr.10: 102854542-102854542 on Build GRCh38
rs957338860 Chr.10: 102854230-102854230 on Build GRCh38
rs111610360 Chr.10: 102854367-102854367 on Build GRCh38
rs747116496 Chr.10: 102854321-102854321 on Build GRCh38
rs570505102 Chr.10: 102854365-102854365 on Build GRCh38
rs368899901 Chr.10: 102854277-102854277 on Build GRCh38
rs17523050 Chr.10: 102854188-102854188 on Build GRCh38
rs770008075 Chr.10: 102854334-102854334 on Build GRCh38
rs1052169046 Chr.10: 102854167-102854167 on Build GRCh38
rs867612369 Chr.10: 102854389-102854389 on Build GRCh38
rs542382839 Chr.10: 102854481-102854481 on Build GRCh38
rs777048317 Chr.10: 102854325-102854325 on Build GRCh38
rs759872142 Chr.10: 102854286-102854286 on Build GRCh38
rs776990316 Chr.10: 102854596-102854597 on Build GRCh38
rs377473812 Chr.10: 102854465-102854465 on Build GRCh38
rs754687640 Chr.10: 102854312-102854312 on Build GRCh38
rs375146221 Chr.10: 102854388-102854388 on Build GRCh38
rs962046337 Chr.10: 102854295-102854295 on Build GRCh38
rs201180336 Chr.10: 102854319-102854319 on Build GRCh38
rs774110787 Chr.10: 102854294-102854294 on Build GRCh38
rs761568723 Chr.10: 102854296-102854296 on Build GRCh38
rs141320276 Chr.10: 102854482-102854482 on Build GRCh38
rs779890664 Chr.10: 102854263-102854263 on Build GRCh38
rs771450267 Chr.10: 102854322-102854322 on Build GRCh38
rs576101338 Chr.10: 102854532-102854532 on Build GRCh38
rs937629606 Chr.10: 102854307-102854307 on Build GRCh38
rs34926154 Chr.10: 102854340-102854340 on Build GRCh38
rs748399407 Chr.10: 102854275-102854275 on Build GRCh38
rs1053688462 Chr.10: 102854418-102854418 on Build GRCh38
rs1028867539 Chr.10: 102854234-102854234 on Build GRCh38
rs991731426 Chr.10: 102854477-102854477 on Build GRCh38
rs772822661 Chr.10: 102854343-102854343 on Build GRCh38

View Full Product Details