Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 116868558-116869053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENO4
Entrez Gene ID
387712
Gene Name
enolase family member 4
Gene Aliases
C10orf134
UniGene
Hs.693248
Gene Chromosome Location
Chr.10: 116848230-116911803 on Build GRCh38
Transcripts
NM_001242699.1 XM_005269815.3 XM_006717835.3 XM_006717836.3 XM_011539797.2 XM_011539799.2 XM_017016241.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3685972 c.1022_1023insA p.G344fs*25 Chr.10: 116868672-116868673 on Build GRCh38
3686421 c.1000-1G>A p.? Chr.10: 116868649-116868649 on Build GRCh38
5477112 c.999+4A>C p.? Chr.10: 116868583-116868583 on Build GRCh38
3686423 c.1036G>T p.D346Y Chr.10: 116868686-116868686 on Build GRCh38
3686422 c.1025A>C p.K342T Chr.10: 116868675-116868675 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1030189915 Chr.10: 116868872-116868872 on Build GRCh38
rs556527257 Chr.10: 116868841-116868841 on Build GRCh38
rs979604559 Chr.10: 116868906-116868906 on Build GRCh38
rs897688449 Chr.10: 116869009-116869009 on Build GRCh38
rs1016071318 Chr.10: 116869002-116869002 on Build GRCh38
rs536998451 Chr.10: 116868898-116868898 on Build GRCh38
rs190354965 Chr.10: 116868945-116868945 on Build GRCh38
rs949117439 Chr.10: 116868954-116868954 on Build GRCh38
rs774543012 Chr.10: 116868878-116868878 on Build GRCh38
rs545335745 Chr.10: 116868907-116868907 on Build GRCh38
rs769733077 Chr.10: 116868747-116868747 on Build GRCh38
rs1052118468 Chr.10: 116868753-116868753 on Build GRCh38
rs947516592 Chr.10: 116868840-116868840 on Build GRCh38
rs942916593 Chr.10: 116868727-116868727 on Build GRCh38
rs552011958 Chr.10: 116868746-116868746 on Build GRCh38
rs186731151 Chr.10: 116868659-116868659 on Build GRCh38
rs961895308 Chr.10: 116868594-116868594 on Build GRCh38
rs772292260 Chr.10: 116868667-116868668 on Build GRCh38
rs960302873 Chr.10: 116869012-116869013 on Build GRCh38
rs904108604 Chr.10: 116868745-116868745 on Build GRCh38
rs12245758 Chr.10: 116868839-116868839 on Build GRCh38
rs1043467447 Chr.10: 116868842-116868843 on Build GRCh38
rs201883348 Chr.10: 116868857-116868857 on Build GRCh38
rs1637572 Chr.10: 116868874-116868874 on Build GRCh38
rs911349441 Chr.10: 116868881-116868881 on Build GRCh38
rs773301233 Chr.10: 116868965-116868965 on Build GRCh38
rs112460241 Chr.10: 116868899-116868899 on Build GRCh38
rs961947253 Chr.10: 116868868-116868868 on Build GRCh38
rs761964048 Chr.10: 116868926-116868926 on Build GRCh38
rs918561096 Chr.10: 116868617-116868617 on Build GRCh38
rs904947366 Chr.10: 116868984-116868984 on Build GRCh38
rs1051215086 Chr.10: 116868682-116868682 on Build GRCh38
rs1023417579 Chr.10: 116868799-116868799 on Build GRCh38
rs548328784 Chr.10: 116868871-116868871 on Build GRCh38
rs1013656642 Chr.10: 116868777-116868777 on Build GRCh38
rs896176301 Chr.10: 116868765-116868765 on Build GRCh38
rs972714586 Chr.10: 116868602-116868602 on Build GRCh38
rs148569690 Chr.10: 116868824-116868824 on Build GRCh38
rs1027081167 Chr.10: 116868955-116868955 on Build GRCh38
rs774729350 Chr.10: 116868605-116868605 on Build GRCh38
rs768026642 Chr.10: 116868686-116868686 on Build GRCh38
rs993279190 Chr.10: 116869021-116869021 on Build GRCh38
rs995614566 Chr.10: 116868678-116868678 on Build GRCh38
rs532126992 Chr.10: 116868685-116868685 on Build GRCh38
rs144769178 Chr.10: 116868636-116868636 on Build GRCh38
rs1005896909 Chr.10: 116869000-116869000 on Build GRCh38
rs935466876 Chr.10: 116868920-116868920 on Build GRCh38
rs1001610580 Chr.10: 116868987-116868987 on Build GRCh38
rs745376022 Chr.10: 116868716-116868716 on Build GRCh38
rs905910651 Chr.10: 116868979-116868979 on Build GRCh38
rs878972910 Chr.10: 116868860-116868861 on Build GRCh38
rs535595959 Chr.10: 116868743-116868743 on Build GRCh38
rs750887648 Chr.10: 116868688-116868688 on Build GRCh38
rs970060687 Chr.10: 116868694-116868694 on Build GRCh38

View Full Product Details