Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 34916023-34916564 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APIP
Entrez Gene ID
51074
Gene Name
APAF1 interacting protein
Gene Aliases
APIP2, CGI-29, CGI29, MMRP19, hAPIP
UniGene
Hs.447794
Gene Chromosome Location
Chr.11: 34882295-34916411 on Build GRCh38
Transcripts
NM_015957.3 XM_011520154.2 XM_017017875.1 NM_001135024.1 NM_001166158.1 NM_003477.2 XM_011520390.1

Gene Symbol
PDHX
Entrez Gene ID
8050
Gene Name
pyruvate dehydrogenase complex component X
Gene Aliases
DLDBP, E3BP, OPDX, PDX1, proX
UniGene
Hs.502315
Gene Chromosome Location
Chr.11: 34915829-34996128 on Build GRCh38
Transcripts
NM_015957.3 XM_011520154.2 XM_017017875.1 NM_001135024.1 NM_001166158.1 NM_003477.2 XM_011520390.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5773054 c.57+9G>A p.? Chr.11: 34916219-34916219 on Build GRCh38
6068500 c.13G>T p.V5L Chr.11: 34916228-34916228 on Build GRCh38
5582082 c.41G>A p.R14Q Chr.11: 34916412-34916412 on Build GRCh38
4543554 c.33G>A p.G11G Chr.11: 34916404-34916404 on Build GRCh38
5773055 c.13+9G>A p.? Chr.11: 34916219-34916219 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs556253723 Chr.11: 34916043-34916043 on Build GRCh38
rs762615901 Chr.11: 34916350-34916350 on Build GRCh38
rs890108854 Chr.11: 34916097-34916097 on Build GRCh38
rs988410408 Chr.11: 34916435-34916435 on Build GRCh38
rs753853669 Chr.11: 34916335-34916335 on Build GRCh38
rs759895240 Chr.11: 34916211-34916211 on Build GRCh38
rs567329509 Chr.11: 34916314-34916314 on Build GRCh38
rs777780994 Chr.11: 34916275-34916275 on Build GRCh38
rs747005576 Chr.11: 34916263-34916263 on Build GRCh38
rs890401987 Chr.11: 34916303-34916303 on Build GRCh38
rs767922429 Chr.11: 34916516-34916522 on Build GRCh38
rs746202661 Chr.11: 34916291-34916291 on Build GRCh38
rs779100145 Chr.11: 34916336-34916336 on Build GRCh38
rs572118408 Chr.11: 34916407-34916407 on Build GRCh38
rs1003040156 Chr.11: 34916481-34916481 on Build GRCh38
rs982080557 Chr.11: 34916091-34916091 on Build GRCh38
rs757293776 Chr.11: 34916374-34916374 on Build GRCh38
rs753674694 Chr.11: 34916526-34916527 on Build GRCh38
rs767425029 Chr.11: 34916356-34916356 on Build GRCh38
rs2016814 Chr.11: 34916150-34916150 on Build GRCh38
rs886048245 Chr.11: 34916298-34916298 on Build GRCh38
rs758466385 Chr.11: 34916533-34916533 on Build GRCh38
rs374157413 Chr.11: 34916166-34916166 on Build GRCh38
rs774655472 Chr.11: 34916210-34916210 on Build GRCh38
rs756933277 Chr.11: 34916243-34916243 on Build GRCh38
rs574902434 Chr.11: 34916464-34916464 on Build GRCh38
rs760074931 Chr.11: 34916228-34916228 on Build GRCh38
rs113073242 Chr.11: 34916454-34916454 on Build GRCh38
rs555955830 Chr.11: 34916348-34916348 on Build GRCh38
rs897512132 Chr.11: 34916346-34916346 on Build GRCh38
rs868532786 Chr.11: 34916432-34916432 on Build GRCh38
rs992171757 Chr.11: 34916309-34916309 on Build GRCh38
rs886048246 Chr.11: 34916501-34916501 on Build GRCh38
rs749401268 Chr.11: 34916277-34916277 on Build GRCh38
rs761199870 Chr.11: 34916302-34916302 on Build GRCh38
rs764336742 Chr.11: 34916263-34916263 on Build GRCh38
rs766242750 Chr.11: 34916326-34916326 on Build GRCh38
rs761422179 Chr.11: 34916316-34916316 on Build GRCh38
rs202241286 Chr.11: 34916226-34916226 on Build GRCh38
rs770555008 Chr.11: 34916486-34916486 on Build GRCh38
rs375130125 Chr.11: 34916532-34916532 on Build GRCh38
rs759318998 Chr.11: 34916494-34916494 on Build GRCh38
rs921397166 Chr.11: 34916480-34916480 on Build GRCh38
rs375386462 Chr.11: 34916176-34916176 on Build GRCh38
rs779852629 Chr.11: 34916259-34916259 on Build GRCh38
rs946156648 Chr.11: 34916504-34916522 on Build GRCh38
rs920198909 Chr.11: 34916501-34916501 on Build GRCh38
rs552157217 Chr.11: 34916184-34916184 on Build GRCh38
rs904375023 Chr.11: 34916053-34916053 on Build GRCh38
rs1008256125 Chr.11: 34916158-34916158 on Build GRCh38
rs868091494 Chr.11: 34916237-34916237 on Build GRCh38
rs771070943 Chr.11: 34916280-34916280 on Build GRCh38
rs2016821 Chr.11: 34916054-34916054 on Build GRCh38
rs763711108 Chr.11: 34916231-34916231 on Build GRCh38
rs899848978 Chr.11: 34916351-34916351 on Build GRCh38
rs760569642 Chr.11: 34916519-34916519 on Build GRCh38
rs769228399 Chr.11: 34916341-34916341 on Build GRCh38
rs778548879 Chr.11: 34916528-34916532 on Build GRCh38
rs2956111 Chr.11: 34916384-34916384 on Build GRCh38
rs752007834 Chr.11: 34916400-34916401 on Build GRCh38
rs762116656 Chr.11: 34916512-34916515 on Build GRCh38
rs773929855 Chr.11: 34916491-34916491 on Build GRCh38
rs988171497 Chr.11: 34916171-34916171 on Build GRCh38
rs764971293 Chr.11: 34916319-34916319 on Build GRCh38
rs188206670 Chr.11: 34916082-34916082 on Build GRCh38
rs764543470 Chr.11: 34916307-34916307 on Build GRCh38
rs774565620 Chr.11: 34916285-34916285 on Build GRCh38
rs772737420 Chr.11: 34916456-34916456 on Build GRCh38
rs1027753975 Chr.11: 34916145-34916145 on Build GRCh38
rs754980282 Chr.11: 34916525-34916526 on Build GRCh38
rs563586329 Chr.11: 34916499-34916499 on Build GRCh38
rs937496575 Chr.11: 34916117-34916117 on Build GRCh38
rs957214303 Chr.11: 34916227-34916227 on Build GRCh38
rs911726687 Chr.11: 34916538-34916538 on Build GRCh38
rs958630928 Chr.11: 34916497-34916497 on Build GRCh38
rs751333457 Chr.11: 34916327-34916327 on Build GRCh38
rs765979127 Chr.11: 34916352-34916352 on Build GRCh38
rs750683422 Chr.11: 34916521-34916522 on Build GRCh38
rs771164728 Chr.11: 34916205-34916205 on Build GRCh38
rs749560956 Chr.11: 34916204-34916204 on Build GRCh38
rs761405893 Chr.11: 34916344-34916344 on Build GRCh38
rs772530666 Chr.11: 34916293-34916293 on Build GRCh38
rs371074122 Chr.11: 34916527-34916529 on Build GRCh38
rs928296244 Chr.11: 34916284-34916284 on Build GRCh38
rs760871034 Chr.11: 34916101-34916101 on Build GRCh38
rs780153979 Chr.11: 34916385-34916385 on Build GRCh38
rs2956113 Chr.11: 34916337-34916337 on Build GRCh38
rs563821039 Chr.11: 34916156-34916156 on Build GRCh38
rs112032820 Chr.11: 34916527-34916527 on Build GRCh38
rs386752391 Chr.11: 34916150-34916151 on Build GRCh38
rs989876829 Chr.11: 34916045-34916045 on Build GRCh38
rs756796889 Chr.11: 34916417-34916417 on Build GRCh38
rs756201233 Chr.11: 34916524-34916527 on Build GRCh38
rs773273824 Chr.11: 34916198-34916198 on Build GRCh38
rs769211875 Chr.11: 34916399-34916399 on Build GRCh38
rs369975644 Chr.11: 34916249-34916249 on Build GRCh38
rs545009828 Chr.11: 34916079-34916079 on Build GRCh38
rs967545063 Chr.11: 34916114-34916114 on Build GRCh38
rs867851814 Chr.11: 34916391-34916391 on Build GRCh38
rs769982440 Chr.11: 34916190-34916190 on Build GRCh38
rs16926448 Chr.11: 34916151-34916151 on Build GRCh38
rs780056321 Chr.11: 34916177-34916177 on Build GRCh38
rs772976341 Chr.11: 34916349-34916349 on Build GRCh38
rs146420175 Chr.11: 34916234-34916234 on Build GRCh38
rs764026434 Chr.11: 34916524-34916524 on Build GRCh38
rs750504158 Chr.11: 34916381-34916381 on Build GRCh38
rs139123979 Chr.11: 34916245-34916245 on Build GRCh38
rs758274248 Chr.11: 34916253-34916253 on Build GRCh38
rs762622673 Chr.11: 34916325-34916325 on Build GRCh38
rs2956114 Chr.11: 34916266-34916266 on Build GRCh38
rs35924042 Chr.11: 34916134-34916134 on Build GRCh38
rs565528504 Chr.11: 34916081-34916081 on Build GRCh38
rs555380381 Chr.11: 34916448-34916448 on Build GRCh38
rs753186558 Chr.11: 34916331-34916331 on Build GRCh38
rs775289563 Chr.11: 34916515-34916515 on Build GRCh38
rs775840738 Chr.11: 34916297-34916297 on Build GRCh38
rs561463212 Chr.11: 34916137-34916137 on Build GRCh38
rs979504930 Chr.11: 34916475-34916475 on Build GRCh38
rs750168506 Chr.11: 34916321-34916321 on Build GRCh38
rs544017835 Chr.11: 34916469-34916469 on Build GRCh38
rs898669437 Chr.11: 34916135-34916135 on Build GRCh38
rs538595954 Chr.11: 34916379-34916379 on Build GRCh38
rs199893242 Chr.11: 34916223-34916223 on Build GRCh38
rs1047160307 Chr.11: 34916317-34916317 on Build GRCh38
rs747343062 Chr.11: 34916338-34916338 on Build GRCh38
rs2956112 Chr.11: 34916354-34916354 on Build GRCh38
rs968049518 Chr.11: 34916459-34916459 on Build GRCh38
rs1019278252 Chr.11: 34916174-34916174 on Build GRCh38
rs759259296 Chr.11: 34916355-34916355 on Build GRCh38

View Full Product Details